Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81526832-81527319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP12
Entrez Gene ID
646486
Gene Name
fatty acid binding protein 12
Gene Aliases
-
UniGene
Hs.586370
Gene Chromosome Location
Chr.8: 81524673-81590664 on Build GRCh38
Transcripts
NM_001105281.1 XM_005251285.4 XM_006716465.3 XM_011517577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569540 c.291C>A p.D97E Chr.8: 81527077-81527077 on Build GRCh38
4600428 c.313A>C p.I105L Chr.8: 81527055-81527055 on Build GRCh38
3651315 c.318G>A p.T106T Chr.8: 81527050-81527050 on Build GRCh38
3651313 c.324G>A p.K108K Chr.8: 81527044-81527044 on Build GRCh38
3835066 c.304G>A p.E102K Chr.8: 81527064-81527064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929959320 Chr.8: 81527207-81527207 on Build GRCh38
rs755811880 Chr.8: 81526972-81526972 on Build GRCh38
rs371573132 Chr.8: 81527054-81527054 on Build GRCh38
rs540524895 Chr.8: 81527293-81527293 on Build GRCh38
rs568288088 Chr.8: 81527051-81527051 on Build GRCh38
rs74730211 Chr.8: 81526955-81526955 on Build GRCh38
rs902063246 Chr.8: 81527290-81527290 on Build GRCh38
rs917463647 Chr.8: 81526884-81526884 on Build GRCh38
rs185223281 Chr.8: 81527105-81527105 on Build GRCh38
rs956359181 Chr.8: 81527033-81527033 on Build GRCh38
rs561314204 Chr.8: 81527246-81527246 on Build GRCh38
rs1007753955 Chr.8: 81526892-81526892 on Build GRCh38
rs766179971 Chr.8: 81527032-81527032 on Build GRCh38
rs868782483 Chr.8: 81527037-81527037 on Build GRCh38
rs1039538672 Chr.8: 81527244-81527244 on Build GRCh38
rs980285877 Chr.8: 81527084-81527084 on Build GRCh38
rs1044530410 Chr.8: 81526909-81526909 on Build GRCh38
rs574975609 Chr.8: 81527088-81527088 on Build GRCh38
rs1006737238 Chr.8: 81527236-81527236 on Build GRCh38
rs752950072 Chr.8: 81527073-81527073 on Build GRCh38
rs147223804 Chr.8: 81526943-81526943 on Build GRCh38
rs371779914 Chr.8: 81527154-81527154 on Build GRCh38
rs190323068 Chr.8: 81526867-81526867 on Build GRCh38
rs941463615 Chr.8: 81527089-81527089 on Build GRCh38
rs535737231 Chr.8: 81527120-81527120 on Build GRCh38
rs781066160 Chr.8: 81527171-81527171 on Build GRCh38
rs781419689 Chr.8: 81527111-81527111 on Build GRCh38
rs968145641 Chr.8: 81526861-81526861 on Build GRCh38
rs770085173 Chr.8: 81527116-81527116 on Build GRCh38
rs745824048 Chr.8: 81527272-81527272 on Build GRCh38
rs768829609 Chr.8: 81527001-81527001 on Build GRCh38
rs748244972 Chr.8: 81527115-81527115 on Build GRCh38
rs780381787 Chr.8: 81527098-81527098 on Build GRCh38
rs1000662577 Chr.8: 81527288-81527288 on Build GRCh38
rs763400246 Chr.8: 81527111-81527112 on Build GRCh38
rs16909306 Chr.8: 81526986-81526986 on Build GRCh38
rs773076749 Chr.8: 81527022-81527022 on Build GRCh38
rs752261434 Chr.8: 81527075-81527075 on Build GRCh38
rs181841840 Chr.8: 81526959-81526959 on Build GRCh38
rs934929205 Chr.8: 81526871-81526871 on Build GRCh38
rs770211701 Chr.8: 81527014-81527014 on Build GRCh38
rs569387483 Chr.8: 81526887-81526887 on Build GRCh38
rs891053824 Chr.8: 81527223-81527223 on Build GRCh38
rs766991525 Chr.8: 81527069-81527069 on Build GRCh38
rs1050992613 Chr.8: 81527208-81527208 on Build GRCh38
rs539028086 Chr.8: 81526870-81526870 on Build GRCh38
rs368067152 Chr.8: 81527050-81527050 on Build GRCh38
rs773505573 Chr.8: 81527170-81527170 on Build GRCh38
rs114393507 Chr.8: 81526979-81526979 on Build GRCh38
rs368473552 Chr.8: 81527004-81527004 on Build GRCh38
rs758809104 Chr.8: 81527096-81527096 on Build GRCh38
rs11995518 Chr.8: 81526864-81526864 on Build GRCh38
rs866183763 Chr.8: 81527123-81527123 on Build GRCh38
rs979589265 Chr.8: 81527000-81527000 on Build GRCh38
rs375883973 Chr.8: 81527085-81527085 on Build GRCh38
rs372512967 Chr.8: 81527008-81527008 on Build GRCh38
rs11995519 Chr.8: 81526876-81526876 on Build GRCh38
rs897114788 Chr.8: 81526976-81526976 on Build GRCh38
rs950180971 Chr.8: 81526885-81526885 on Build GRCh38
rs564575673 Chr.8: 81527086-81527086 on Build GRCh38
rs752936870 Chr.8: 81527012-81527013 on Build GRCh38
rs746307941 Chr.8: 81526978-81526978 on Build GRCh38
rs779067892 Chr.8: 81527070-81527070 on Build GRCh38
rs747532400 Chr.8: 81527104-81527104 on Build GRCh38
rs538375305 Chr.8: 81526921-81526921 on Build GRCh38
rs780619714 Chr.8: 81526984-81526984 on Build GRCh38
rs375525557 Chr.8: 81527025-81527025 on Build GRCh38
rs752314681 Chr.8: 81526970-81526970 on Build GRCh38
rs776903473 Chr.8: 81527002-81527002 on Build GRCh38
rs11992576 Chr.8: 81527286-81527286 on Build GRCh38
rs566712459 Chr.8: 81526869-81526869 on Build GRCh38
rs905960966 Chr.8: 81526914-81526914 on Build GRCh38
rs779233701 Chr.8: 81526975-81526975 on Build GRCh38

View Full Product Details