Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85202707-85203213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F5
Entrez Gene ID
1875
Gene Name
E2F transcription factor 5
Gene Aliases
E2F-5
UniGene
Hs.445758
Gene Chromosome Location
Chr.8: 85177384-85214518 on Build GRCh38
Transcripts
NM_001083588.1 NM_001083589.1 NM_001951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799462 c.345-5T>C p.? Chr.8: 85203089-85203089 on Build GRCh38
6304528 c.398T>A p.L133H Chr.8: 85203147-85203147 on Build GRCh38
3651385 c.429G>A p.K143K Chr.8: 85203178-85203178 on Build GRCh38
7365776 c.376G>A p.V126I Chr.8: 85203125-85203125 on Build GRCh38
7317383 c.406G>A p.E136K Chr.8: 85203155-85203155 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs954569323 Chr.8: 85202969-85202969 on Build GRCh38
rs770438498 Chr.8: 85203146-85203146 on Build GRCh38
rs887314127 Chr.8: 85202817-85202817 on Build GRCh38
rs552560126 Chr.8: 85202982-85202982 on Build GRCh38
rs755993431 Chr.8: 85203084-85203084 on Build GRCh38
rs543811621 Chr.8: 85202801-85202801 on Build GRCh38
rs926009125 Chr.8: 85203072-85203072 on Build GRCh38
rs530703879 Chr.8: 85202763-85202763 on Build GRCh38
rs752051603 Chr.8: 85202730-85202730 on Build GRCh38
rs948309096 Chr.8: 85202812-85202812 on Build GRCh38
rs368524242 Chr.8: 85203089-85203089 on Build GRCh38
rs1020106336 Chr.8: 85203043-85203043 on Build GRCh38
rs898933068 Chr.8: 85202843-85202843 on Build GRCh38
rs566425334 Chr.8: 85202932-85202932 on Build GRCh38
rs958797162 Chr.8: 85203085-85203085 on Build GRCh38
rs997244089 Chr.8: 85203024-85203024 on Build GRCh38
rs778592679 Chr.8: 85203149-85203149 on Build GRCh38
rs375519002 Chr.8: 85203062-85203062 on Build GRCh38
rs554914900 Chr.8: 85202955-85202955 on Build GRCh38
rs1008856506 Chr.8: 85202972-85202972 on Build GRCh38
rs991415588 Chr.8: 85203096-85203096 on Build GRCh38
rs528732056 Chr.8: 85202948-85202948 on Build GRCh38
rs745462728 Chr.8: 85203157-85203157 on Build GRCh38
rs901608040 Chr.8: 85202936-85202936 on Build GRCh38
rs4150941 Chr.8: 85203005-85203005 on Build GRCh38
rs950310511 Chr.8: 85202758-85202758 on Build GRCh38
rs4150940 Chr.8: 85202961-85202961 on Build GRCh38
rs767025378 Chr.8: 85203057-85203057 on Build GRCh38
rs546710828 Chr.8: 85202872-85202872 on Build GRCh38
rs371244692 Chr.8: 85203056-85203056 on Build GRCh38
rs1050092784 Chr.8: 85203049-85203049 on Build GRCh38
rs11988348 Chr.8: 85202824-85202824 on Build GRCh38
rs529769201 Chr.8: 85202842-85202842 on Build GRCh38
rs750095835 Chr.8: 85203058-85203058 on Build GRCh38
rs780896098 Chr.8: 85203069-85203069 on Build GRCh38
rs771857458 Chr.8: 85203165-85203165 on Build GRCh38
rs917251917 Chr.8: 85203132-85203132 on Build GRCh38
rs751056904 Chr.8: 85203081-85203082 on Build GRCh38
rs777120076 Chr.8: 85203185-85203185 on Build GRCh38
rs917472989 Chr.8: 85202731-85202731 on Build GRCh38
rs748716655 Chr.8: 85203133-85203133 on Build GRCh38
rs752557784 Chr.8: 85203080-85203080 on Build GRCh38
rs939382497 Chr.8: 85202737-85202737 on Build GRCh38
rs551994994 Chr.8: 85202960-85202960 on Build GRCh38

View Full Product Details