Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85214453-85214983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf59
Entrez Gene ID
401466
Gene Name
chromosome 8 open reading frame 59
Gene Aliases
-
UniGene
Hs.443072
Gene Chromosome Location
Chr.8: 85214053-85220422 on Build GRCh38
Transcripts
NM_001099670.2 NM_001099671.2 NM_001099672.2 NM_001099673.2 NM_001293320.1 XM_011517528.2 NM_001083588.1 NM_001083589.1 NM_001951.3

Gene Symbol
E2F5
Entrez Gene ID
1875
Gene Name
E2F transcription factor 5
Gene Aliases
E2F-5
UniGene
Hs.445758
Gene Chromosome Location
Chr.8: 85177384-85214518 on Build GRCh38
Transcripts
NM_001099670.2 NM_001099671.2 NM_001099672.2 NM_001099673.2 NM_001293320.1 XM_011517528.2 NM_001083588.1 NM_001083589.1 NM_001951.3

SNP Detail

SNP ID Chromosome Location
rs748866461 Chr.8: 85214582-85214582 on Build GRCh38
rs561464143 Chr.8: 85214739-85214739 on Build GRCh38
rs777994267 Chr.8: 85214666-85214668 on Build GRCh38
rs965076656 Chr.8: 85214682-85214682 on Build GRCh38
rs373760171 Chr.8: 85214908-85214908 on Build GRCh38
rs776860201 Chr.8: 85214618-85214618 on Build GRCh38
rs556208081 Chr.8: 85214515-85214515 on Build GRCh38
rs952666030 Chr.8: 85214836-85214836 on Build GRCh38
rs191018956 Chr.8: 85214476-85214476 on Build GRCh38
rs975051539 Chr.8: 85214783-85214783 on Build GRCh38
rs750041883 Chr.8: 85214899-85214899 on Build GRCh38
rs992604486 Chr.8: 85214876-85214876 on Build GRCh38
rs376027637 Chr.8: 85214650-85214650 on Build GRCh38
rs774418869 Chr.8: 85214939-85214939 on Build GRCh38
rs202090035 Chr.8: 85214588-85214589 on Build GRCh38
rs61703408 Chr.8: 85214601-85214602 on Build GRCh38
rs918351038 Chr.8: 85214879-85214879 on Build GRCh38
rs1027912816 Chr.8: 85214811-85214811 on Build GRCh38
rs970051042 Chr.8: 85214707-85214707 on Build GRCh38
rs763785999 Chr.8: 85214529-85214530 on Build GRCh38
rs745550311 Chr.8: 85214606-85214606 on Build GRCh38
rs756066698 Chr.8: 85214881-85214881 on Build GRCh38
rs183133372 Chr.8: 85214901-85214901 on Build GRCh38
rs908395779 Chr.8: 85214857-85214858 on Build GRCh38
rs540254303 Chr.8: 85214826-85214826 on Build GRCh38
rs200451411 Chr.8: 85214570-85214570 on Build GRCh38
rs370757686 Chr.8: 85214807-85214807 on Build GRCh38
rs758702544 Chr.8: 85214660-85214661 on Build GRCh38
rs377353626 Chr.8: 85214546-85214546 on Build GRCh38
rs984444120 Chr.8: 85214838-85214838 on Build GRCh38
rs998644271 Chr.8: 85214632-85214632 on Build GRCh38
rs763145927 Chr.8: 85214535-85214535 on Build GRCh38
rs777335121 Chr.8: 85214580-85214580 on Build GRCh38
rs201945771 Chr.8: 85214920-85214920 on Build GRCh38
rs1022505867 Chr.8: 85214701-85214701 on Build GRCh38
rs998249753 Chr.8: 85214590-85214590 on Build GRCh38
rs373044712 Chr.8: 85214614-85214614 on Build GRCh38
rs748508230 Chr.8: 85214918-85214918 on Build GRCh38
rs573602987 Chr.8: 85214959-85214959 on Build GRCh38
rs781604003 Chr.8: 85214914-85214914 on Build GRCh38
rs747403753 Chr.8: 85214892-85214893 on Build GRCh38
rs766474573 Chr.8: 85214635-85214635 on Build GRCh38
rs564799502 Chr.8: 85214765-85214765 on Build GRCh38
rs763059450 Chr.8: 85214628-85214628 on Build GRCh38
rs749816393 Chr.8: 85214936-85214936 on Build GRCh38
rs376907630 Chr.8: 85214518-85214518 on Build GRCh38
rs759799870 Chr.8: 85214940-85214940 on Build GRCh38
rs770259878 Chr.8: 85214622-85214622 on Build GRCh38
rs867424952 Chr.8: 85214602-85214602 on Build GRCh38
rs757702699 Chr.8: 85214894-85214894 on Build GRCh38
rs560083275 Chr.8: 85214858-85214858 on Build GRCh38
rs759766631 Chr.8: 85214646-85214646 on Build GRCh38
rs112505588 Chr.8: 85214592-85214592 on Build GRCh38
rs1039547769 Chr.8: 85214560-85214560 on Build GRCh38
rs774272223 Chr.8: 85214580-85214580 on Build GRCh38
rs772242693 Chr.8: 85214941-85214941 on Build GRCh38
rs773228144 Chr.8: 85214532-85214532 on Build GRCh38
rs765290215 Chr.8: 85214528-85214528 on Build GRCh38
rs370313519 Chr.8: 85214892-85214892 on Build GRCh38
rs752501727 Chr.8: 85214663-85214663 on Build GRCh38
rs138606099 Chr.8: 85214590-85214591 on Build GRCh38
rs774506628 Chr.8: 85214639-85214639 on Build GRCh38
rs756790167 Chr.8: 85214894-85214894 on Build GRCh38
rs530285532 Chr.8: 85214744-85214744 on Build GRCh38
rs980955380 Chr.8: 85214791-85214791 on Build GRCh38
rs753371291 Chr.8: 85214893-85214893 on Build GRCh38
rs778592930 Chr.8: 85214895-85214895 on Build GRCh38
rs767238372 Chr.8: 85214563-85214563 on Build GRCh38
rs201160424 Chr.8: 85214604-85214604 on Build GRCh38
rs746414634 Chr.8: 85214896-85214897 on Build GRCh38
rs35310594 Chr.8: 85214823-85214823 on Build GRCh38
rs544787591 Chr.8: 85214623-85214623 on Build GRCh38
rs766633951 Chr.8: 85214539-85214539 on Build GRCh38
rs1011342632 Chr.8: 85214693-85214693 on Build GRCh38
rs141613857 Chr.8: 85214599-85214600 on Build GRCh38
rs781680215 Chr.8: 85214893-85214894 on Build GRCh38
rs575774428 Chr.8: 85214619-85214619 on Build GRCh38
rs752622993 Chr.8: 85214564-85214564 on Build GRCh38
rs754726127 Chr.8: 85214557-85214557 on Build GRCh38
rs867967451 Chr.8: 85214603-85214603 on Build GRCh38
rs778211260 Chr.8: 85214927-85214927 on Build GRCh38
rs1035224506 Chr.8: 85214828-85214828 on Build GRCh38
rs752837008 Chr.8: 85214602-85214603 on Build GRCh38
rs770940141 Chr.8: 85214938-85214938 on Build GRCh38

View Full Product Details