Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85218951-85219472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf59
Entrez Gene ID
401466
Gene Name
chromosome 8 open reading frame 59
Gene Aliases
-
UniGene
Hs.443072
Gene Chromosome Location
Chr.8: 85214053-85220422 on Build GRCh38
Transcripts
NM_001099670.2 NM_001099671.2 NM_001099672.2 NM_001099673.2 NM_001293320.1 NR_120681.1 XM_011517528.2

SNP Detail

SNP ID Chromosome Location
rs920302181 Chr.8: 85219200-85219200 on Build GRCh38
rs36053988 Chr.8: 85219299-85219299 on Build GRCh38
rs370332598 Chr.8: 85219074-85219074 on Build GRCh38
rs1038771609 Chr.8: 85219138-85219138 on Build GRCh38
rs188735798 Chr.8: 85219266-85219266 on Build GRCh38
rs964913183 Chr.8: 85219433-85219433 on Build GRCh38
rs911756694 Chr.8: 85219034-85219034 on Build GRCh38
rs544592159 Chr.8: 85219051-85219051 on Build GRCh38
rs912041247 Chr.8: 85219100-85219100 on Build GRCh38
rs760933176 Chr.8: 85219256-85219256 on Build GRCh38
rs191169278 Chr.8: 85219011-85219011 on Build GRCh38
rs67573812 Chr.8: 85219234-85219234 on Build GRCh38
rs1001741224 Chr.8: 85219294-85219294 on Build GRCh38
rs1045918507 Chr.8: 85219242-85219242 on Build GRCh38
rs985910724 Chr.8: 85219029-85219029 on Build GRCh38
rs994461596 Chr.8: 85219298-85219298 on Build GRCh38
rs966955409 Chr.8: 85219395-85219395 on Build GRCh38
rs375928081 Chr.8: 85219280-85219280 on Build GRCh38
rs886385866 Chr.8: 85219247-85219247 on Build GRCh38
rs958340450 Chr.8: 85219322-85219322 on Build GRCh38
rs894726803 Chr.8: 85219186-85219186 on Build GRCh38
rs149110219 Chr.8: 85219406-85219406 on Build GRCh38
rs953285663 Chr.8: 85219381-85219381 on Build GRCh38
rs920506590 Chr.8: 85218976-85218976 on Build GRCh38
rs551443384 Chr.8: 85219296-85219296 on Build GRCh38
rs985983430 Chr.8: 85219417-85219417 on Build GRCh38
rs563696127 Chr.8: 85219118-85219118 on Build GRCh38
rs1054494481 Chr.8: 85219142-85219142 on Build GRCh38
rs961855128 Chr.8: 85219297-85219297 on Build GRCh38
rs191387565 Chr.8: 85219422-85219422 on Build GRCh38
rs930346275 Chr.8: 85219206-85219206 on Build GRCh38
rs35620009 Chr.8: 85219367-85219367 on Build GRCh38
rs530477025 Chr.8: 85219088-85219088 on Build GRCh38
rs183259071 Chr.8: 85219148-85219148 on Build GRCh38
rs944568180 Chr.8: 85219054-85219054 on Build GRCh38
rs531567684 Chr.8: 85219345-85219345 on Build GRCh38
rs1042168895 Chr.8: 85219062-85219062 on Build GRCh38
rs529489138 Chr.8: 85219139-85219139 on Build GRCh38
rs550370079 Chr.8: 85219113-85219113 on Build GRCh38

View Full Product Details