Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85448250-85448750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA3
Entrez Gene ID
761
Gene Name
carbonic anhydrase 3
Gene Aliases
CAIII, Car3
UniGene
Hs.82129
Gene Chromosome Location
Chr.8: 85438827-85449040 on Build GRCh38
Transcripts
NM_005181.3 NR_121630.1 NR_121631.1 NR_121632.1

Gene Symbol
CA3-AS1
Entrez Gene ID
100996348
Gene Name
CA3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.606624
Gene Chromosome Location
Chr.8: 85441806-85464915 on Build GRCh38
Transcripts
NM_005181.3 NR_121630.1 NR_121631.1 NR_121632.1

SNP Detail

SNP ID Chromosome Location
rs1054712082 Chr.8: 85448538-85448538 on Build GRCh38
rs976597076 Chr.8: 85448387-85448387 on Build GRCh38
rs1003258422 Chr.8: 85448690-85448690 on Build GRCh38
rs557692970 Chr.8: 85448494-85448494 on Build GRCh38
rs1013089709 Chr.8: 85448578-85448578 on Build GRCh38
rs3087782 Chr.8: 85448544-85448544 on Build GRCh38
rs904797252 Chr.8: 85448608-85448608 on Build GRCh38
rs187100077 Chr.8: 85448449-85448449 on Build GRCh38
rs576869280 Chr.8: 85448292-85448293 on Build GRCh38
rs192012127 Chr.8: 85448543-85448543 on Build GRCh38
rs762174004 Chr.8: 85448435-85448435 on Build GRCh38
rs773181917 Chr.8: 85448328-85448328 on Build GRCh38
rs944785913 Chr.8: 85448492-85448492 on Build GRCh38
rs973283883 Chr.8: 85448298-85448298 on Build GRCh38
rs1036724767 Chr.8: 85448507-85448507 on Build GRCh38
rs970884599 Chr.8: 85448605-85448605 on Build GRCh38
rs1024450078 Chr.8: 85448604-85448604 on Build GRCh38
rs903150484 Chr.8: 85448519-85448519 on Build GRCh38
rs920467671 Chr.8: 85448303-85448303 on Build GRCh38
rs547649946 Chr.8: 85448720-85448720 on Build GRCh38
rs978750634 Chr.8: 85448327-85448327 on Build GRCh38
rs766783694 Chr.8: 85448721-85448721 on Build GRCh38
rs535781821 Chr.8: 85448345-85448345 on Build GRCh38
rs956148094 Chr.8: 85448641-85448641 on Build GRCh38
rs985334562 Chr.8: 85448482-85448482 on Build GRCh38
rs531914525 Chr.8: 85448573-85448573 on Build GRCh38
rs911976370 Chr.8: 85448454-85448454 on Build GRCh38
rs572116879 Chr.8: 85448421-85448421 on Build GRCh38
rs1000139317 Chr.8: 85448617-85448617 on Build GRCh38
rs182570902 Chr.8: 85448422-85448422 on Build GRCh38
rs962274782 Chr.8: 85448295-85448295 on Build GRCh38
rs762394244 Chr.8: 85448462-85448462 on Build GRCh38
rs577689535 Chr.8: 85448514-85448514 on Build GRCh38
rs956349808 Chr.8: 85448359-85448359 on Build GRCh38
rs1031658898 Chr.8: 85448634-85448634 on Build GRCh38
rs183290622 Chr.8: 85448635-85448635 on Build GRCh38
rs569797933 Chr.8: 85448311-85448311 on Build GRCh38
rs555677527 Chr.8: 85448406-85448406 on Build GRCh38
rs767656204 Chr.8: 85448531-85448531 on Build GRCh38
rs761187519 Chr.8: 85448692-85448692 on Build GRCh38
rs1023182287 Chr.8: 85448307-85448307 on Build GRCh38

View Full Product Details