Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85448629-85449174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA3
Entrez Gene ID
761
Gene Name
carbonic anhydrase 3
Gene Aliases
CAIII, Car3
UniGene
Hs.82129
Gene Chromosome Location
Chr.8: 85438827-85449040 on Build GRCh38
Transcripts
NM_005181.3 NR_121630.1 NR_121631.1 NR_121632.1

Gene Symbol
CA3-AS1
Entrez Gene ID
100996348
Gene Name
CA3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.606624
Gene Chromosome Location
Chr.8: 85441806-85464915 on Build GRCh38
Transcripts
NM_005181.3 NR_121630.1 NR_121631.1 NR_121632.1

SNP Detail

SNP ID Chromosome Location
rs935970952 Chr.8: 85448880-85448880 on Build GRCh38
rs894744898 Chr.8: 85449041-85449041 on Build GRCh38
rs1003258422 Chr.8: 85448690-85448690 on Build GRCh38
rs4566 Chr.8: 85448853-85448853 on Build GRCh38
rs1046327041 Chr.8: 85449047-85449047 on Build GRCh38
rs918281137 Chr.8: 85448838-85448838 on Build GRCh38
rs985301992 Chr.8: 85448761-85448761 on Build GRCh38
rs752300472 Chr.8: 85449042-85449042 on Build GRCh38
rs569777443 Chr.8: 85448833-85448833 on Build GRCh38
rs547649946 Chr.8: 85448720-85448720 on Build GRCh38
rs527938602 Chr.8: 85448756-85448756 on Build GRCh38
rs766783694 Chr.8: 85448721-85448721 on Build GRCh38
rs897524615 Chr.8: 85449083-85449083 on Build GRCh38
rs11545785 Chr.8: 85448946-85448946 on Build GRCh38
rs1003741139 Chr.8: 85449070-85449070 on Build GRCh38
rs758082603 Chr.8: 85449073-85449073 on Build GRCh38
rs907359010 Chr.8: 85449061-85449061 on Build GRCh38
rs190056888 Chr.8: 85448784-85448784 on Build GRCh38
rs1044571977 Chr.8: 85449023-85449023 on Build GRCh38
rs1030130161 Chr.8: 85449093-85449093 on Build GRCh38
rs977525759 Chr.8: 85448852-85448852 on Build GRCh38
rs973316762 Chr.8: 85448753-85448753 on Build GRCh38
rs994474394 Chr.8: 85449149-85449149 on Build GRCh38
rs948861857 Chr.8: 85448969-85448969 on Build GRCh38
rs954181224 Chr.8: 85449097-85449100 on Build GRCh38
rs187244388 Chr.8: 85448758-85448758 on Build GRCh38
rs754437685 Chr.8: 85448806-85448806 on Build GRCh38
rs201132723 Chr.8: 85448888-85448888 on Build GRCh38
rs533281997 Chr.8: 85448790-85448790 on Build GRCh38
rs1014658117 Chr.8: 85449071-85449071 on Build GRCh38
rs761187519 Chr.8: 85448692-85448692 on Build GRCh38
rs550231914 Chr.8: 85448809-85448809 on Build GRCh38

View Full Product Details