Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85463625-85464119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA2
Entrez Gene ID
760
Gene Name
carbonic anhydrase 2
Gene Aliases
CA-II, CAC, CAII, Car2, HEL-76, HEL-S-282
UniGene
Hs.155097
Gene Chromosome Location
Chr.8: 85463902-85481492 on Build GRCh38
Transcripts
NM_000067.2 NM_001293675.1 NR_121630.1 NR_121631.1

Gene Symbol
CA3-AS1
Entrez Gene ID
100996348
Gene Name
CA3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.606624
Gene Chromosome Location
Chr.8: 85441806-85464915 on Build GRCh38
Transcripts
NM_000067.2 NM_001293675.1 NR_121630.1 NR_121631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686273 c.21C>A p.Y7* Chr.8: 85464102-85464102 on Build GRCh38
1132685 c.22G>T p.G8C Chr.8: 85464103-85464103 on Build GRCh38
6381782 c.26A>T p.K9I Chr.8: 85464107-85464107 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901835844 Chr.8: 85463700-85463700 on Build GRCh38
rs1031886124 Chr.8: 85463754-85463754 on Build GRCh38
rs1048976134 Chr.8: 85463997-85463998 on Build GRCh38
rs886927195 Chr.8: 85464003-85464003 on Build GRCh38
rs780289062 Chr.8: 85464046-85464046 on Build GRCh38
rs901885492 Chr.8: 85464006-85464006 on Build GRCh38
rs77895131 Chr.8: 85463769-85463770 on Build GRCh38
rs11261477 Chr.8: 85463901-85463901 on Build GRCh38
rs889232543 Chr.8: 85463659-85463659 on Build GRCh38
rs886063147 Chr.8: 85464000-85464001 on Build GRCh38
rs776620660 Chr.8: 85464071-85464071 on Build GRCh38
rs1033318567 Chr.8: 85464056-85464056 on Build GRCh38
rs953637967 Chr.8: 85463828-85463828 on Build GRCh38
rs937489581 Chr.8: 85463653-85463653 on Build GRCh38
rs980539798 Chr.8: 85463927-85463927 on Build GRCh38
rs982695053 Chr.8: 85463857-85463866 on Build GRCh38
rs557422827 Chr.8: 85463944-85463944 on Build GRCh38
rs960408531 Chr.8: 85463947-85463947 on Build GRCh38
rs1053036058 Chr.8: 85464049-85464049 on Build GRCh38
rs548979293 Chr.8: 85463755-85463755 on Build GRCh38
rs986352027 Chr.8: 85463856-85463856 on Build GRCh38
rs893365560 Chr.8: 85463782-85463782 on Build GRCh38
rs943446423 Chr.8: 85463665-85463665 on Build GRCh38
rs914725254 Chr.8: 85463918-85463918 on Build GRCh38
rs969583527 Chr.8: 85463812-85463812 on Build GRCh38
rs1041962608 Chr.8: 85463931-85463931 on Build GRCh38
rs370317596 Chr.8: 85464035-85464035 on Build GRCh38
rs556311734 Chr.8: 85464039-85464039 on Build GRCh38
rs746755362 Chr.8: 85464061-85464061 on Build GRCh38
rs957711602 Chr.8: 85464070-85464070 on Build GRCh38
rs1025334692 Chr.8: 85463862-85463862 on Build GRCh38
rs891112016 Chr.8: 85463760-85463760 on Build GRCh38
rs372555587 Chr.8: 85464096-85464096 on Build GRCh38
rs1002130884 Chr.8: 85464055-85464055 on Build GRCh38
rs1026785469 Chr.8: 85463838-85463838 on Build GRCh38
rs886063146 Chr.8: 85463906-85463906 on Build GRCh38
rs1023072498 Chr.8: 85463810-85463810 on Build GRCh38
rs946231839 Chr.8: 85463930-85463930 on Build GRCh38
rs868721474 Chr.8: 85463740-85463740 on Build GRCh38
rs989651022 Chr.8: 85463907-85463907 on Build GRCh38
rs912838940 Chr.8: 85463937-85463937 on Build GRCh38
rs1018304924 Chr.8: 85463794-85463794 on Build GRCh38
rs529147093 Chr.8: 85463648-85463648 on Build GRCh38
rs761725568 Chr.8: 85464081-85464081 on Build GRCh38
rs911912552 Chr.8: 85463865-85463865 on Build GRCh38
rs772237919 Chr.8: 85464040-85464040 on Build GRCh38
rs1042381274 Chr.8: 85463745-85463745 on Build GRCh38
rs984645370 Chr.8: 85463970-85463970 on Build GRCh38
rs769398095 Chr.8: 85464098-85464098 on Build GRCh38
rs910381596 Chr.8: 85463974-85463974 on Build GRCh38
rs539649032 Chr.8: 85463991-85463991 on Build GRCh38
rs989228844 Chr.8: 85464093-85464093 on Build GRCh38
rs1040813945 Chr.8: 85463687-85463687 on Build GRCh38
rs934679925 Chr.8: 85464011-85464011 on Build GRCh38
rs995549547 Chr.8: 85463829-85463830 on Build GRCh38
rs768352706 Chr.8: 85464066-85464066 on Build GRCh38
rs191493709 Chr.8: 85463852-85463852 on Build GRCh38
rs886063148 Chr.8: 85464029-85464029 on Build GRCh38
rs1011730557 Chr.8: 85463809-85463809 on Build GRCh38
rs980473651 Chr.8: 85463894-85463894 on Build GRCh38
rs943482809 Chr.8: 85463984-85463984 on Build GRCh38
rs930531676 Chr.8: 85463966-85463966 on Build GRCh38
rs746064476 Chr.8: 85464038-85464038 on Build GRCh38
rs1024891279 Chr.8: 85464037-85464037 on Build GRCh38
rs570970117 Chr.8: 85463952-85463952 on Build GRCh38
rs1008209971 Chr.8: 85463793-85463793 on Build GRCh38
rs971702156 Chr.8: 85463949-85463949 on Build GRCh38

View Full Product Details