Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90058833-90059329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALB1
Entrez Gene ID
793
Gene Name
calbindin 1
Gene Aliases
CALB, D-28K
UniGene
Hs.65425
Gene Chromosome Location
Chr.8: 90058608-90082881 on Build GRCh38
Transcripts
NM_004929.3

SNP Detail

SNP ID Chromosome Location
rs375011102 Chr.8: 90059292-90059292 on Build GRCh38
rs949705214 Chr.8: 90059234-90059234 on Build GRCh38
rs1054680655 Chr.8: 90059304-90059304 on Build GRCh38
rs879251782 Chr.8: 90059010-90059010 on Build GRCh38
rs997271342 Chr.8: 90058908-90058909 on Build GRCh38
rs1019685510 Chr.8: 90058868-90058868 on Build GRCh38
rs1041452346 Chr.8: 90059261-90059262 on Build GRCh38
rs994230945 Chr.8: 90059287-90059287 on Build GRCh38
rs976931864 Chr.8: 90059005-90059005 on Build GRCh38
rs972544772 Chr.8: 90059176-90059176 on Build GRCh38
rs571767388 Chr.8: 90059302-90059302 on Build GRCh38
rs1033744272 Chr.8: 90058862-90058862 on Build GRCh38
rs901229274 Chr.8: 90059276-90059276 on Build GRCh38
rs1026119102 Chr.8: 90058924-90058927 on Build GRCh38
rs1043316773 Chr.8: 90059108-90059109 on Build GRCh38
rs918256240 Chr.8: 90059199-90059199 on Build GRCh38
rs913418495 Chr.8: 90059051-90059051 on Build GRCh38
rs965883788 Chr.8: 90058887-90058887 on Build GRCh38
rs1045647259 Chr.8: 90059269-90059269 on Build GRCh38
rs561236064 Chr.8: 90058900-90058900 on Build GRCh38
rs954183344 Chr.8: 90059035-90059035 on Build GRCh38
rs950613405 Chr.8: 90058966-90058966 on Build GRCh38
rs1006889574 Chr.8: 90059251-90059251 on Build GRCh38
rs1053248327 Chr.8: 90059049-90059050 on Build GRCh38
rs569398579 Chr.8: 90058985-90058985 on Build GRCh38
rs909152676 Chr.8: 90058997-90058997 on Build GRCh38
rs1020903138 Chr.8: 90058963-90058963 on Build GRCh38
rs910075426 Chr.8: 90059093-90059093 on Build GRCh38
rs547469638 Chr.8: 90059296-90059296 on Build GRCh38
rs376824982 Chr.8: 90059055-90059059 on Build GRCh38
rs962764894 Chr.8: 90059103-90059103 on Build GRCh38
rs890067816 Chr.8: 90059113-90059113 on Build GRCh38
rs1001785413 Chr.8: 90058857-90058857 on Build GRCh38
rs980216461 Chr.8: 90059016-90059016 on Build GRCh38
rs187862991 Chr.8: 90058957-90058957 on Build GRCh38
rs947557686 Chr.8: 90059098-90059098 on Build GRCh38
rs1805896 Chr.8: 90059014-90059014 on Build GRCh38
rs780656928 Chr.8: 90058893-90058893 on Build GRCh38
rs749833620 Chr.8: 90058970-90058970 on Build GRCh38
rs986051334 Chr.8: 90059092-90059092 on Build GRCh38
rs905658567 Chr.8: 90058856-90058856 on Build GRCh38
rs148552007 Chr.8: 90059002-90059002 on Build GRCh38
rs938436932 Chr.8: 90059049-90059053 on Build GRCh38
rs1025708251 Chr.8: 90059288-90059288 on Build GRCh38

View Full Product Details