Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167919679-167920210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC2
Entrez Gene ID
25874
Gene Name
mitochondrial pyruvate carrier 2
Gene Aliases
BRP44
UniGene
Hs.517768
Gene Chromosome Location
Chr.1: 167916675-167937069 on Build GRCh38
Transcripts
NM_001143674.3 NM_015415.3 NR_026550.2 XM_006711266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666191 c.236-2A>G p.? Chr.1: 167920092-167920092 on Build GRCh38
2155541 c.338G>A p.R113H Chr.1: 167919988-167919988 on Build GRCh38
6714550 c.279T>A p.N93K Chr.1: 167920047-167920047 on Build GRCh38
207639 c.303C>A p.F101L Chr.1: 167920023-167920023 on Build GRCh38
6567006 c.281G>T p.W94L Chr.1: 167920045-167920045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17556487 Chr.1: 167919856-167919856 on Build GRCh38
rs753860575 Chr.1: 167920000-167920000 on Build GRCh38
rs370288807 Chr.1: 167919965-167919965 on Build GRCh38
rs199537893 Chr.1: 167919908-167919908 on Build GRCh38
rs1006980785 Chr.1: 167920175-167920175 on Build GRCh38
rs17343090 Chr.1: 167919989-167919989 on Build GRCh38
rs1054523447 Chr.1: 167919784-167919784 on Build GRCh38
rs750120214 Chr.1: 167920012-167920012 on Build GRCh38
rs190995994 Chr.1: 167919888-167919888 on Build GRCh38
rs764237612 Chr.1: 167919991-167919991 on Build GRCh38
rs565346745 Chr.1: 167919845-167919845 on Build GRCh38
rs924626145 Chr.1: 167919765-167919765 on Build GRCh38
rs531435728 Chr.1: 167919737-167919737 on Build GRCh38
rs895003865 Chr.1: 167919962-167919962 on Build GRCh38
rs34646997 Chr.1: 167920155-167920155 on Build GRCh38
rs559127162 Chr.1: 167920152-167920152 on Build GRCh38
rs76483034 Chr.1: 167919923-167919923 on Build GRCh38
rs533622874 Chr.1: 167919945-167919945 on Build GRCh38
rs1054971501 Chr.1: 167919911-167919911 on Build GRCh38
rs765025318 Chr.1: 167920110-167920111 on Build GRCh38
rs113323575 Chr.1: 167920039-167920039 on Build GRCh38
rs973595755 Chr.1: 167919725-167919725 on Build GRCh38
rs376267824 Chr.1: 167920033-167920033 on Build GRCh38
rs746166200 Chr.1: 167920138-167920138 on Build GRCh38
rs928992466 Chr.1: 167919776-167919776 on Build GRCh38
rs138442408 Chr.1: 167919785-167919785 on Build GRCh38
rs780660875 Chr.1: 167920057-167920057 on Build GRCh38
rs936111157 Chr.1: 167919778-167919778 on Build GRCh38
rs548627012 Chr.1: 167919905-167919905 on Build GRCh38
rs201446823 Chr.1: 167920166-167920166 on Build GRCh38
rs200893061 Chr.1: 167919902-167919902 on Build GRCh38
rs993743147 Chr.1: 167919864-167919864 on Build GRCh38
rs761642173 Chr.1: 167920003-167920003 on Build GRCh38
rs887433515 Chr.1: 167919894-167919894 on Build GRCh38
rs943518489 Chr.1: 167919853-167919853 on Build GRCh38
rs549964224 Chr.1: 167919839-167919839 on Build GRCh38
rs749868549 Chr.1: 167920068-167920068 on Build GRCh38
rs75981218 Chr.1: 167919921-167919921 on Build GRCh38
rs537213051 Chr.1: 167920047-167920047 on Build GRCh38
rs186190544 Chr.1: 167919857-167919857 on Build GRCh38
rs765145098 Chr.1: 167920008-167920008 on Build GRCh38
rs983338372 Chr.1: 167919838-167919838 on Build GRCh38
rs894979236 Chr.1: 167920114-167920114 on Build GRCh38
rs924937976 Chr.1: 167919756-167919757 on Build GRCh38
rs758060310 Chr.1: 167920017-167920017 on Build GRCh38
rs536946046 Chr.1: 167919895-167919895 on Build GRCh38
rs113507981 Chr.1: 167919978-167919978 on Build GRCh38
rs771341294 Chr.1: 167920079-167920079 on Build GRCh38
rs758021542 Chr.1: 167919843-167919843 on Build GRCh38
rs779345466 Chr.1: 167920136-167920136 on Build GRCh38
rs1044220055 Chr.1: 167920010-167920010 on Build GRCh38
rs902136501 Chr.1: 167919857-167919857 on Build GRCh38
rs920271701 Chr.1: 167919735-167919735 on Build GRCh38
rs751101699 Chr.1: 167920041-167920041 on Build GRCh38
rs570213717 Chr.1: 167919935-167919935 on Build GRCh38
rs112286447 Chr.1: 167920123-167920123 on Build GRCh38

View Full Product Details