Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167935648-167936143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC2
Entrez Gene ID
25874
Gene Name
mitochondrial pyruvate carrier 2
Gene Aliases
BRP44
UniGene
Hs.517768
Gene Chromosome Location
Chr.1: 167916675-167937069 on Build GRCh38
Transcripts
NM_001143674.3 NM_015415.3 NR_026550.2 XM_006711266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3802736 c.76G>A p.E26K Chr.1: 167935766-167935766 on Build GRCh38
6395327 c.64C>A p.L22M Chr.1: 167935778-167935778 on Build GRCh38
1601191 c.38_39insA p.Y13fs*1 Chr.1: 167935803-167935804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011205080 Chr.1: 167936014-167936014 on Build GRCh38
rs956662196 Chr.1: 167936013-167936013 on Build GRCh38
rs770882889 Chr.1: 167935737-167935737 on Build GRCh38
rs757373217 Chr.1: 167935856-167935856 on Build GRCh38
rs202065715 Chr.1: 167935699-167935699 on Build GRCh38
rs778857293 Chr.1: 167935739-167935739 on Build GRCh38
rs531303902 Chr.1: 167936107-167936107 on Build GRCh38
rs760077800 Chr.1: 167935829-167935829 on Build GRCh38
rs756306815 Chr.1: 167935844-167935844 on Build GRCh38
rs906267243 Chr.1: 167935973-167935973 on Build GRCh38
rs189051947 Chr.1: 167936043-167936043 on Build GRCh38
rs1030949231 Chr.1: 167935726-167935726 on Build GRCh38
rs559971510 Chr.1: 167936028-167936028 on Build GRCh38
rs758665226 Chr.1: 167935681-167935681 on Build GRCh38
rs936807930 Chr.1: 167935808-167935808 on Build GRCh38
rs964580115 Chr.1: 167935672-167935672 on Build GRCh38
rs987620113 Chr.1: 167936106-167936106 on Build GRCh38
rs956815605 Chr.1: 167935756-167935756 on Build GRCh38
rs763675160 Chr.1: 167936035-167936035 on Build GRCh38
rs777928479 Chr.1: 167935846-167935846 on Build GRCh38
rs12746154 Chr.1: 167935820-167935820 on Build GRCh38
rs898935354 Chr.1: 167935689-167935689 on Build GRCh38
rs1003399152 Chr.1: 167935981-167935981 on Build GRCh38
rs375999580 Chr.1: 167935706-167935706 on Build GRCh38
rs984861856 Chr.1: 167935698-167935698 on Build GRCh38
rs548357867 Chr.1: 167936066-167936066 on Build GRCh38
rs374785748 Chr.1: 167936063-167936063 on Build GRCh38
rs536422844 Chr.1: 167935777-167935777 on Build GRCh38
rs947893646 Chr.1: 167935779-167935779 on Build GRCh38
rs761366753 Chr.1: 167935811-167935811 on Build GRCh38
rs1037386764 Chr.1: 167935908-167935908 on Build GRCh38
rs745793010 Chr.1: 167935762-167935762 on Build GRCh38
rs969164372 Chr.1: 167936036-167936036 on Build GRCh38
rs1011915106 Chr.1: 167935960-167935960 on Build GRCh38
rs77188947 Chr.1: 167935790-167935790 on Build GRCh38
rs545944343 Chr.1: 167935836-167935850 on Build GRCh38
rs12032727 Chr.1: 167935696-167935696 on Build GRCh38
rs764810377 Chr.1: 167935812-167935812 on Build GRCh38
rs745857752 Chr.1: 167935859-167935859 on Build GRCh38
rs984514219 Chr.1: 167936047-167936047 on Build GRCh38
rs1014505210 Chr.1: 167936062-167936062 on Build GRCh38
rs143230699 Chr.1: 167935774-167935774 on Build GRCh38
rs753043678 Chr.1: 167935833-167935833 on Build GRCh38
rs1027195140 Chr.1: 167936008-167936008 on Build GRCh38
rs1031228775 Chr.1: 167936000-167936000 on Build GRCh38
rs565129308 Chr.1: 167935813-167935813 on Build GRCh38
rs948019576 Chr.1: 167936112-167936112 on Build GRCh38
rs752004373 Chr.1: 167935701-167935701 on Build GRCh38
rs779725806 Chr.1: 167935891-167935891 on Build GRCh38
rs541071028 Chr.1: 167936016-167936016 on Build GRCh38
rs900113676 Chr.1: 167935956-167935956 on Build GRCh38
rs766738676 Chr.1: 167935692-167935694 on Build GRCh38
rs754024630 Chr.1: 167935851-167935851 on Build GRCh38
rs771977419 Chr.1: 167935863-167935863 on Build GRCh38
rs769339266 Chr.1: 167935975-167935975 on Build GRCh38
rs11553557 Chr.1: 167935868-167935868 on Build GRCh38
rs865791987 Chr.1: 167935757-167935757 on Build GRCh38
rs372760188 Chr.1: 167935691-167935691 on Build GRCh38
rs185287770 Chr.1: 167935782-167935782 on Build GRCh38
rs577207194 Chr.1: 167935921-167935921 on Build GRCh38
rs961705456 Chr.1: 167936091-167936091 on Build GRCh38
rs373391367 Chr.1: 167935718-167935718 on Build GRCh38
rs376625019 Chr.1: 167935830-167935830 on Build GRCh38
rs898794214 Chr.1: 167935957-167935957 on Build GRCh38
rs150529062 Chr.1: 167935786-167935786 on Build GRCh38
rs1021628595 Chr.1: 167936019-167936019 on Build GRCh38
rs778948843 Chr.1: 167935858-167935858 on Build GRCh38
rs563568591 Chr.1: 167936075-167936075 on Build GRCh38
rs772674652 Chr.1: 167936093-167936093 on Build GRCh38
rs368734933 Chr.1: 167935707-167935707 on Build GRCh38
rs750724509 Chr.1: 167935677-167935677 on Build GRCh38
rs771817573 Chr.1: 167935765-167935765 on Build GRCh38
rs9943195 Chr.1: 167935965-167935965 on Build GRCh38
rs748379648 Chr.1: 167935710-167935710 on Build GRCh38
rs748551016 Chr.1: 167935932-167935932 on Build GRCh38

View Full Product Details