Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167991065-167991516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
UniGene
Hs.435741
Gene Chromosome Location
Chr.1: 167936247-168075843 on Build GRCh38
Transcripts
NM_001017977.2 NM_001198956.1 NM_001198957.1 NM_018442.3 XM_005245331.4 XM_005245332.4 XM_005245333.4 XM_017001779.1 XM_017001780.1 XM_017001781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477858 c.581C>T p.A194V Chr.1: 167991232-167991232 on Build GRCh38
6640546 c.571C>T p.R191* Chr.1: 167991222-167991222 on Build GRCh38
6640545 c.571C>T p.R191* Chr.1: 167991222-167991222 on Build GRCh38
1203070 c.572G>A p.R191Q Chr.1: 167991223-167991223 on Build GRCh38
7180407 c.644C>T p.S215L Chr.1: 167991295-167991295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761876903 Chr.1: 167991313-167991313 on Build GRCh38
rs981482476 Chr.1: 167991484-167991484 on Build GRCh38
rs769457841 Chr.1: 167991232-167991232 on Build GRCh38
rs778943974 Chr.1: 167991168-167991168 on Build GRCh38
rs754022376 Chr.1: 167991215-167991215 on Build GRCh38
rs182295643 Chr.1: 167991356-167991356 on Build GRCh38
rs144780485 Chr.1: 167991206-167991206 on Build GRCh38
rs768963114 Chr.1: 167991303-167991303 on Build GRCh38
rs776926593 Chr.1: 167991312-167991312 on Build GRCh38
rs537452883 Chr.1: 167991183-167991183 on Build GRCh38
rs756258272 Chr.1: 167991154-167991154 on Build GRCh38
rs777076741 Chr.1: 167991271-167991271 on Build GRCh38
rs776330085 Chr.1: 167991184-167991184 on Build GRCh38
rs907036158 Chr.1: 167991257-167991257 on Build GRCh38
rs747645638 Chr.1: 167991150-167991150 on Build GRCh38
rs370331379 Chr.1: 167991236-167991236 on Build GRCh38
rs1026159715 Chr.1: 167991439-167991439 on Build GRCh38
rs764220841 Chr.1: 167991157-167991157 on Build GRCh38
rs575785493 Chr.1: 167991424-167991424 on Build GRCh38
rs541147287 Chr.1: 167991400-167991400 on Build GRCh38
rs962880654 Chr.1: 167991405-167991405 on Build GRCh38
rs754841555 Chr.1: 167991338-167991338 on Build GRCh38
rs141191160 Chr.1: 167991270-167991270 on Build GRCh38
rs780053774 Chr.1: 167991254-167991254 on Build GRCh38
rs544056995 Chr.1: 167991172-167991172 on Build GRCh38
rs376255164 Chr.1: 167991178-167991178 on Build GRCh38
rs746450901 Chr.1: 167991274-167991274 on Build GRCh38
rs539692470 Chr.1: 167991430-167991430 on Build GRCh38
rs557404859 Chr.1: 167991420-167991420 on Build GRCh38
rs370286423 Chr.1: 167991190-167991190 on Build GRCh38
rs780705724 Chr.1: 167991352-167991354 on Build GRCh38
rs201922591 Chr.1: 167991166-167991166 on Build GRCh38
rs776156549 Chr.1: 167991140-167991140 on Build GRCh38
rs754717069 Chr.1: 167991258-167991258 on Build GRCh38
rs774144475 Chr.1: 167991408-167991409 on Build GRCh38
rs775876588 Chr.1: 167991299-167991299 on Build GRCh38
rs946508292 Chr.1: 167991225-167991225 on Build GRCh38
rs765173203 Chr.1: 167991235-167991235 on Build GRCh38
rs374086980 Chr.1: 167991325-167991325 on Build GRCh38
rs766387347 Chr.1: 167991327-167991327 on Build GRCh38
rs750457636 Chr.1: 167991323-167991323 on Build GRCh38
rs747704216 Chr.1: 167991267-167991267 on Build GRCh38
rs772934981 Chr.1: 167991260-167991260 on Build GRCh38
rs1056006796 Chr.1: 167991187-167991187 on Build GRCh38
rs902147015 Chr.1: 167991247-167991247 on Build GRCh38
rs922806032 Chr.1: 167991403-167991403 on Build GRCh38
rs758263884 Chr.1: 167991253-167991253 on Build GRCh38
rs962857921 Chr.1: 167991089-167991089 on Build GRCh38
rs751501475 Chr.1: 167991336-167991336 on Build GRCh38
rs781021091 Chr.1: 167991263-167991263 on Build GRCh38
rs754589200 Chr.1: 167991186-167991190 on Build GRCh38
rs372070991 Chr.1: 167991384-167991384 on Build GRCh38
rs536135552 Chr.1: 167991365-167991365 on Build GRCh38
rs894872834 Chr.1: 167991189-167991189 on Build GRCh38
rs761062262 Chr.1: 167991300-167991300 on Build GRCh38
rs781072446 Chr.1: 167991344-167991344 on Build GRCh38
rs548210459 Chr.1: 167991391-167991394 on Build GRCh38
rs747661098 Chr.1: 167991185-167991185 on Build GRCh38
rs375920232 Chr.1: 167991174-167991174 on Build GRCh38
rs1047634625 Chr.1: 167991309-167991309 on Build GRCh38
rs533166219 Chr.1: 167991427-167991428 on Build GRCh38
rs535125415 Chr.1: 167991314-167991314 on Build GRCh38
rs757382904 Chr.1: 167991223-167991223 on Build GRCh38
rs760939923 Chr.1: 167991205-167991205 on Build GRCh38
rs1012603324 Chr.1: 167991230-167991230 on Build GRCh38
rs983064410 Chr.1: 167991429-167991429 on Build GRCh38
rs928865758 Chr.1: 167991487-167991487 on Build GRCh38
rs775825049 Chr.1: 167991204-167991204 on Build GRCh38
rs772549706 Chr.1: 167991298-167991298 on Build GRCh38
rs762742078 Chr.1: 167991374-167991374 on Build GRCh38
rs748889182 Chr.1: 167991393-167991394 on Build GRCh38
rs769144204 Chr.1: 167991188-167991188 on Build GRCh38
rs552628179 Chr.1: 167991107-167991107 on Build GRCh38
rs767925052 Chr.1: 167991197-167991197 on Build GRCh38
rs964145992 Chr.1: 167991389-167991389 on Build GRCh38
rs951323832 Chr.1: 167991469-167991469 on Build GRCh38
rs746777537 Chr.1: 167991256-167991256 on Build GRCh38

View Full Product Details