Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169367617-169368126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320972.1 NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1 NM_013330.4 NM_197972.2 NR_104229.1 XM_005245106.4 XM_017001099.1

Gene Symbol
NME7
Entrez Gene ID
29922
Gene Name
NME/NM23 family member 7
Gene Aliases
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
UniGene
Hs.706952
Gene Chromosome Location
Chr.1: 169132385-169367963 on Build GRCh38
Transcripts
NM_001320972.1 NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1 NM_013330.4 NM_197972.2 NR_104229.1 XM_005245106.4 XM_017001099.1

SNP Detail

SNP ID Chromosome Location
rs1039266274 Chr.1: 169367937-169367937 on Build GRCh38
rs759866194 Chr.1: 169367744-169367744 on Build GRCh38
rs376146232 Chr.1: 169367694-169367694 on Build GRCh38
rs182440832 Chr.1: 169367825-169367825 on Build GRCh38
rs574988276 Chr.1: 169367866-169367866 on Build GRCh38
rs1023327588 Chr.1: 169367823-169367823 on Build GRCh38
rs866912095 Chr.1: 169367654-169367654 on Build GRCh38
rs1045122897 Chr.1: 169368099-169368099 on Build GRCh38
rs528625051 Chr.1: 169367973-169367973 on Build GRCh38
rs1039767734 Chr.1: 169367673-169367673 on Build GRCh38
rs781440758 Chr.1: 169367665-169367666 on Build GRCh38
rs540871211 Chr.1: 169367759-169367760 on Build GRCh38
rs753968466 Chr.1: 169367755-169367755 on Build GRCh38
rs896041101 Chr.1: 169368031-169368031 on Build GRCh38
rs780155209 Chr.1: 169367718-169367718 on Build GRCh38
rs965379842 Chr.1: 169367918-169367918 on Build GRCh38
rs902275300 Chr.1: 169367899-169367899 on Build GRCh38
rs890744196 Chr.1: 169367707-169367707 on Build GRCh38
rs951202988 Chr.1: 169367881-169367881 on Build GRCh38
rs781725684 Chr.1: 169367754-169367754 on Build GRCh38
rs903403567 Chr.1: 169367754-169367754 on Build GRCh38
rs748272513 Chr.1: 169367689-169367689 on Build GRCh38
rs757152108 Chr.1: 169367705-169367705 on Build GRCh38
rs886227069 Chr.1: 169368007-169368007 on Build GRCh38
rs1034856377 Chr.1: 169367809-169367809 on Build GRCh38
rs575200562 Chr.1: 169367954-169367954 on Build GRCh38
rs778841633 Chr.1: 169367709-169367709 on Build GRCh38
rs79305322 Chr.1: 169367972-169367972 on Build GRCh38
rs1023537568 Chr.1: 169368082-169368082 on Build GRCh38
rs370315923 Chr.1: 169367675-169367675 on Build GRCh38
rs868727526 Chr.1: 169367978-169367978 on Build GRCh38
rs1004627526 Chr.1: 169368014-169368014 on Build GRCh38
rs983712779 Chr.1: 169367807-169367807 on Build GRCh38
rs948813558 Chr.1: 169367643-169367643 on Build GRCh38
rs564420690 Chr.1: 169367968-169367968 on Build GRCh38
rs1035272052 Chr.1: 169368045-169368045 on Build GRCh38
rs1013681096 Chr.1: 169367785-169367785 on Build GRCh38
rs1053731711 Chr.1: 169367894-169367894 on Build GRCh38
rs761246597 Chr.1: 169367726-169367726 on Build GRCh38
rs960940593 Chr.1: 169367819-169367819 on Build GRCh38
rs991158196 Chr.1: 169368077-169368077 on Build GRCh38
rs755215348 Chr.1: 169367739-169367739 on Build GRCh38
rs774874724 Chr.1: 169367732-169367732 on Build GRCh38
rs569190245 Chr.1: 169367777-169367777 on Build GRCh38
rs554755904 Chr.1: 169367668-169367668 on Build GRCh38
rs375459194 Chr.1: 169367714-169367714 on Build GRCh38
rs201280194 Chr.1: 169367745-169367745 on Build GRCh38
rs1042043357 Chr.1: 169367867-169367867 on Build GRCh38
rs921299327 Chr.1: 169367934-169367934 on Build GRCh38
rs753034821 Chr.1: 169367749-169367749 on Build GRCh38
rs924740627 Chr.1: 169367999-169367999 on Build GRCh38
rs770453293 Chr.1: 169368062-169368062 on Build GRCh38
rs777995054 Chr.1: 169367693-169367693 on Build GRCh38
rs896127786 Chr.1: 169367768-169367768 on Build GRCh38
rs750332506 Chr.1: 169367774-169367775 on Build GRCh38
rs967920712 Chr.1: 169368098-169368098 on Build GRCh38
rs371164733 Chr.1: 169367711-169367711 on Build GRCh38
rs17345517 Chr.1: 169367882-169367882 on Build GRCh38
rs138292594 Chr.1: 169367953-169367953 on Build GRCh38
rs904713714 Chr.1: 169367658-169367658 on Build GRCh38
rs934745251 Chr.1: 169367884-169367884 on Build GRCh38
rs915914381 Chr.1: 169367854-169367854 on Build GRCh38
rs576920432 Chr.1: 169367791-169367791 on Build GRCh38
rs948694251 Chr.1: 169367860-169367860 on Build GRCh38
rs771881486 Chr.1: 169367712-169367712 on Build GRCh38
rs371770204 Chr.1: 169367758-169367758 on Build GRCh38
rs751659915 Chr.1: 169367728-169367728 on Build GRCh38
rs563506635 Chr.1: 169367902-169367902 on Build GRCh38
rs199701543 Chr.1: 169367676-169367676 on Build GRCh38
rs975807965 Chr.1: 169367930-169367930 on Build GRCh38
rs559311599 Chr.1: 169367862-169367862 on Build GRCh38
rs757317205 Chr.1: 169367757-169367757 on Build GRCh38
rs530994165 Chr.1: 169367932-169367932 on Build GRCh38
rs546636382 Chr.1: 169368009-169368009 on Build GRCh38

View Full Product Details