Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169382025-169382527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336063 c.923C>A p.S308Y Chr.1: 169382187-169382187 on Build GRCh38
3477945 c.923C>T p.S308F Chr.1: 169382187-169382187 on Build GRCh38
1295444 c.798-1G>A p.? Chr.1: 169382061-169382061 on Build GRCh38
3477944 c.850T>G p.S284A Chr.1: 169382114-169382114 on Build GRCh38
6409464 c.992C>A p.T331N Chr.1: 169382256-169382256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763773221 Chr.1: 169382214-169382214 on Build GRCh38
rs770880796 Chr.1: 169382132-169382132 on Build GRCh38
rs749415821 Chr.1: 169382273-169382273 on Build GRCh38
rs151227755 Chr.1: 169382241-169382241 on Build GRCh38
rs901286172 Chr.1: 169382122-169382122 on Build GRCh38
rs771987442 Chr.1: 169382286-169382286 on Build GRCh38
rs748305816 Chr.1: 169382118-169382118 on Build GRCh38
rs3905328 Chr.1: 169382358-169382358 on Build GRCh38
rs752007228 Chr.1: 169382237-169382237 on Build GRCh38
rs759375514 Chr.1: 169382281-169382281 on Build GRCh38
rs745870460 Chr.1: 169382073-169382073 on Build GRCh38
rs760258475 Chr.1: 169382205-169382205 on Build GRCh38
rs776979549 Chr.1: 169382115-169382115 on Build GRCh38
rs773489397 Chr.1: 169382445-169382445 on Build GRCh38
rs771930138 Chr.1: 169382104-169382104 on Build GRCh38
rs1016134664 Chr.1: 169382105-169382105 on Build GRCh38
rs770036329 Chr.1: 169382246-169382246 on Build GRCh38
rs755712576 Chr.1: 169382059-169382059 on Build GRCh38
rs770964640 Chr.1: 169382275-169382275 on Build GRCh38
rs367863339 Chr.1: 169382053-169382053 on Build GRCh38
rs371736116 Chr.1: 169382280-169382280 on Build GRCh38
rs756777650 Chr.1: 169382231-169382231 on Build GRCh38
rs931199738 Chr.1: 169382443-169382443 on Build GRCh38
rs141688688 Chr.1: 169382391-169382391 on Build GRCh38
rs544110336 Chr.1: 169382446-169382446 on Build GRCh38
rs7538157 Chr.1: 169382306-169382306 on Build GRCh38
rs774130419 Chr.1: 169382135-169382135 on Build GRCh38
rs768874021 Chr.1: 169382113-169382113 on Build GRCh38
rs769948503 Chr.1: 169382119-169382119 on Build GRCh38
rs764826652 Chr.1: 169382319-169382319 on Build GRCh38
rs940001793 Chr.1: 169382494-169382494 on Build GRCh38
rs781427328 Chr.1: 169382240-169382240 on Build GRCh38
rs762983684 Chr.1: 169382123-169382123 on Build GRCh38
rs759413842 Chr.1: 169382149-169382149 on Build GRCh38
rs879775106 Chr.1: 169382487-169382489 on Build GRCh38
rs752256487 Chr.1: 169382057-169382057 on Build GRCh38
rs980977008 Chr.1: 169382402-169382402 on Build GRCh38
rs970544396 Chr.1: 169382399-169382399 on Build GRCh38
rs116716791 Chr.1: 169382442-169382442 on Build GRCh38
rs911638202 Chr.1: 169382350-169382350 on Build GRCh38
rs1023102436 Chr.1: 169382294-169382294 on Build GRCh38
rs767246103 Chr.1: 169382056-169382056 on Build GRCh38
rs773288637 Chr.1: 169382120-169382120 on Build GRCh38
rs543538093 Chr.1: 169382103-169382103 on Build GRCh38
rs1039297285 Chr.1: 169382389-169382389 on Build GRCh38
rs761483125 Chr.1: 169382311-169382311 on Build GRCh38
rs760505274 Chr.1: 169382303-169382303 on Build GRCh38
rs749827673 Chr.1: 169382182-169382182 on Build GRCh38
rs751389885 Chr.1: 169382051-169382051 on Build GRCh38
rs777926369 Chr.1: 169382257-169382257 on Build GRCh38
rs538865634 Chr.1: 169382299-169382299 on Build GRCh38
rs757746401 Chr.1: 169382266-169382267 on Build GRCh38
rs1048990758 Chr.1: 169382422-169382422 on Build GRCh38
rs780747450 Chr.1: 169382235-169382235 on Build GRCh38
rs886356938 Chr.1: 169382425-169382425 on Build GRCh38
rs200364171 Chr.1: 169382095-169382095 on Build GRCh38
rs752449008 Chr.1: 169382204-169382204 on Build GRCh38
rs753428598 Chr.1: 169382222-169382222 on Build GRCh38
rs758977043 Chr.1: 169382092-169382092 on Build GRCh38
rs577753234 Chr.1: 169382071-169382071 on Build GRCh38
rs930466284 Chr.1: 169382405-169382405 on Build GRCh38
rs747451163 Chr.1: 169382107-169382107 on Build GRCh38
rs767207875 Chr.1: 169382152-169382152 on Build GRCh38
rs755429516 Chr.1: 169382238-169382238 on Build GRCh38
rs577700573 Chr.1: 169382064-169382064 on Build GRCh38
rs748749868 Chr.1: 169382066-169382066 on Build GRCh38
rs114660556 Chr.1: 169382320-169382320 on Build GRCh38

View Full Product Details