Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169386830-169387345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336065 c.1041T>C p.V347V Chr.1: 169387020-169387020 on Build GRCh38
6442703 c.1025G>A p.R342K Chr.1: 169387004-169387004 on Build GRCh38
5480930 c.1027A>G p.I343V Chr.1: 169387006-169387006 on Build GRCh38
4458014 c.1069C>T p.P357S Chr.1: 169387048-169387048 on Build GRCh38
7154697 c.1142G>C p.R381T Chr.1: 169387121-169387121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144655565 Chr.1: 169387071-169387071 on Build GRCh38
rs962605791 Chr.1: 169386996-169386996 on Build GRCh38
rs372941684 Chr.1: 169386955-169386955 on Build GRCh38
rs752618372 Chr.1: 169386893-169386893 on Build GRCh38
rs767962455 Chr.1: 169387102-169387102 on Build GRCh38
rs764427263 Chr.1: 169387117-169387117 on Build GRCh38
rs779453114 Chr.1: 169387041-169387043 on Build GRCh38
rs778869599 Chr.1: 169387016-169387016 on Build GRCh38
rs754141029 Chr.1: 169387124-169387124 on Build GRCh38
rs776031512 Chr.1: 169387200-169387200 on Build GRCh38
rs756533987 Chr.1: 169387115-169387115 on Build GRCh38
rs11544428 Chr.1: 169387111-169387111 on Build GRCh38
rs567056442 Chr.1: 169387261-169387261 on Build GRCh38
rs765905577 Chr.1: 169387190-169387190 on Build GRCh38
rs1026569894 Chr.1: 169387314-169387314 on Build GRCh38
rs749505610 Chr.1: 169386980-169386980 on Build GRCh38
rs550591205 Chr.1: 169387177-169387177 on Build GRCh38
rs746930700 Chr.1: 169387158-169387158 on Build GRCh38
rs769356495 Chr.1: 169387180-169387180 on Build GRCh38
rs776574218 Chr.1: 169387160-169387160 on Build GRCh38
rs768568248 Chr.1: 169387063-169387063 on Build GRCh38
rs756058398 Chr.1: 169386952-169386952 on Build GRCh38
rs762480944 Chr.1: 169387097-169387097 on Build GRCh38
rs775447641 Chr.1: 169387025-169387025 on Build GRCh38
rs1006617698 Chr.1: 169386928-169386928 on Build GRCh38
rs779153670 Chr.1: 169387142-169387142 on Build GRCh38
rs781698324 Chr.1: 169386953-169386953 on Build GRCh38
rs746276207 Chr.1: 169387216-169387216 on Build GRCh38
rs778777571 Chr.1: 169386894-169386894 on Build GRCh38
rs1029255977 Chr.1: 169386903-169386903 on Build GRCh38
rs575507575 Chr.1: 169386922-169386922 on Build GRCh38
rs761571132 Chr.1: 169387076-169387076 on Build GRCh38
rs994677538 Chr.1: 169387280-169387280 on Build GRCh38
rs138679316 Chr.1: 169387084-169387084 on Build GRCh38
rs999209801 Chr.1: 169386888-169386888 on Build GRCh38
rs757591575 Chr.1: 169387225-169387225 on Build GRCh38
rs189043990 Chr.1: 169387188-169387188 on Build GRCh38
rs768658214 Chr.1: 169387159-169387159 on Build GRCh38
rs757473451 Chr.1: 169387125-169387125 on Build GRCh38
rs201383109 Chr.1: 169386978-169386978 on Build GRCh38
rs764515306 Chr.1: 169387207-169387207 on Build GRCh38
rs950732754 Chr.1: 169386908-169386908 on Build GRCh38
rs556966143 Chr.1: 169386920-169386920 on Build GRCh38
rs753168847 Chr.1: 169387103-169387103 on Build GRCh38
rs544642079 Chr.1: 169386886-169386886 on Build GRCh38
rs148163554 Chr.1: 169387089-169387089 on Build GRCh38
rs369740623 Chr.1: 169387022-169387022 on Build GRCh38
rs755449024 Chr.1: 169386950-169386950 on Build GRCh38
rs74383380 Chr.1: 169387060-169387060 on Build GRCh38
rs200837980 Chr.1: 169387151-169387151 on Build GRCh38
rs545549685 Chr.1: 169386945-169386945 on Build GRCh38
rs761181111 Chr.1: 169387205-169387205 on Build GRCh38
rs373643357 Chr.1: 169387154-169387154 on Build GRCh38
rs551974736 Chr.1: 169387210-169387210 on Build GRCh38
rs1044021757 Chr.1: 169386885-169386885 on Build GRCh38
rs771052582 Chr.1: 169386998-169386998 on Build GRCh38
rs116192263 Chr.1: 169387189-169387189 on Build GRCh38
rs1044112526 Chr.1: 169387193-169387193 on Build GRCh38
rs778008272 Chr.1: 169386979-169386979 on Build GRCh38
rs192424034 Chr.1: 169387310-169387310 on Build GRCh38

View Full Product Details