Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170983331-170983836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH9
Entrez Gene ID
80133
Gene Name
maestro heat like repeat family member 9
Gene Aliases
ARMC11, C1orf129
UniGene
Hs.591489
Gene Chromosome Location
Chr.1: 170935471-171066341 on Build GRCh38
Transcripts
NM_001163629.1 NM_025063.2 XM_011510005.2 XM_011510006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478211 c.667C>T p.P223S Chr.1: 170983472-170983472 on Build GRCh38
5818132 c.726T>C p.A242A Chr.1: 170983531-170983531 on Build GRCh38
3478212 c.667C>T p.P223S Chr.1: 170983472-170983472 on Build GRCh38
6715250 c.729+2T>C p.? Chr.1: 170983536-170983536 on Build GRCh38
5567709 c.656G>T p.S219I Chr.1: 170983461-170983461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376392385 Chr.1: 170983403-170983403 on Build GRCh38
rs1003932493 Chr.1: 170983503-170983503 on Build GRCh38
rs567301071 Chr.1: 170983777-170983777 on Build GRCh38
rs774743916 Chr.1: 170983680-170983680 on Build GRCh38
rs201549824 Chr.1: 170983485-170983485 on Build GRCh38
rs370652735 Chr.1: 170983381-170983381 on Build GRCh38
rs548706826 Chr.1: 170983764-170983764 on Build GRCh38
rs759554597 Chr.1: 170983532-170983532 on Build GRCh38
rs752904080 Chr.1: 170983458-170983458 on Build GRCh38
rs1036065965 Chr.1: 170983634-170983634 on Build GRCh38
rs371793612 Chr.1: 170983547-170983547 on Build GRCh38
rs536309064 Chr.1: 170983482-170983482 on Build GRCh38
rs931767140 Chr.1: 170983635-170983636 on Build GRCh38
rs781476670 Chr.1: 170983561-170983561 on Build GRCh38
rs561437742 Chr.1: 170983517-170983517 on Build GRCh38
rs527244324 Chr.1: 170983693-170983693 on Build GRCh38
rs762495463 Chr.1: 170983390-170983390 on Build GRCh38
rs560306816 Chr.1: 170983656-170983656 on Build GRCh38
rs779204268 Chr.1: 170983457-170983457 on Build GRCh38
rs746306461 Chr.1: 170983563-170983563 on Build GRCh38
rs940531009 Chr.1: 170983426-170983426 on Build GRCh38
rs764687608 Chr.1: 170983404-170983404 on Build GRCh38
rs373670994 Chr.1: 170983435-170983435 on Build GRCh38
rs367575706 Chr.1: 170983447-170983447 on Build GRCh38
rs768149166 Chr.1: 170983504-170983504 on Build GRCh38
rs988218807 Chr.1: 170983521-170983521 on Build GRCh38
rs746433019 Chr.1: 170983497-170983497 on Build GRCh38
rs760479378 Chr.1: 170983438-170983438 on Build GRCh38
rs774824147 Chr.1: 170983384-170983384 on Build GRCh38
rs761315275 Chr.1: 170983404-170983405 on Build GRCh38
rs757628549 Chr.1: 170983558-170983558 on Build GRCh38
rs199994744 Chr.1: 170983487-170983487 on Build GRCh38
rs773855378 Chr.1: 170983508-170983508 on Build GRCh38
rs533599844 Chr.1: 170983557-170983557 on Build GRCh38
rs766369095 Chr.1: 170983443-170983443 on Build GRCh38
rs758710250 Chr.1: 170983468-170983468 on Build GRCh38
rs765354128 Chr.1: 170983535-170983535 on Build GRCh38
rs115503255 Chr.1: 170983544-170983544 on Build GRCh38
rs112394419 Chr.1: 170983511-170983511 on Build GRCh38
rs756627786 Chr.1: 170983577-170983577 on Build GRCh38
rs377151868 Chr.1: 170983516-170983516 on Build GRCh38
rs116251875 Chr.1: 170983456-170983456 on Build GRCh38
rs778343986 Chr.1: 170983583-170983583 on Build GRCh38
rs1045008229 Chr.1: 170983739-170983739 on Build GRCh38
rs761520213 Chr.1: 170983421-170983421 on Build GRCh38
rs1040069428 Chr.1: 170983635-170983638 on Build GRCh38
rs77952906 Chr.1: 170983635-170983635 on Build GRCh38
rs898993551 Chr.1: 170983455-170983455 on Build GRCh38
rs558605019 Chr.1: 170983621-170983621 on Build GRCh38
rs371155414 Chr.1: 170983425-170983425 on Build GRCh38
rs762206507 Chr.1: 170983473-170983475 on Build GRCh38
rs766133020 Chr.1: 170983518-170983518 on Build GRCh38
rs773980157 Chr.1: 170983404-170983404 on Build GRCh38
rs776614693 Chr.1: 170983523-170983523 on Build GRCh38
rs770070716 Chr.1: 170983537-170983537 on Build GRCh38
rs767232814 Chr.1: 170983422-170983422 on Build GRCh38
rs911653245 Chr.1: 170983620-170983620 on Build GRCh38
rs903670962 Chr.1: 170983636-170983636 on Build GRCh38
rs1043920919 Chr.1: 170983636-170983636 on Build GRCh38
rs1031364738 Chr.1: 170983480-170983480 on Build GRCh38
rs754322656 Chr.1: 170983455-170983455 on Build GRCh38
rs930463374 Chr.1: 170983639-170983639 on Build GRCh38
rs970125039 Chr.1: 170983499-170983499 on Build GRCh38
rs373241096 Chr.1: 170983391-170983391 on Build GRCh38

View Full Product Details