Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170989776-170990278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH9
Entrez Gene ID
80133
Gene Name
maestro heat like repeat family member 9
Gene Aliases
ARMC11, C1orf129
UniGene
Hs.591489
Gene Chromosome Location
Chr.1: 170935471-171066341 on Build GRCh38
Transcripts
NM_001163629.1 NM_025063.2 XM_011510005.2 XM_011510006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
677202 c.901A>C p.I301L Chr.1: 170989876-170989876 on Build GRCh38
5796907 c.977C>T p.S326L Chr.1: 170989952-170989952 on Build GRCh38
6059617 c.987G>T p.K329N Chr.1: 170989962-170989962 on Build GRCh38
2074281 c.1018C>T p.P340S Chr.1: 170989993-170989993 on Build GRCh38
2074275 c.892G>A p.V298M Chr.1: 170989867-170989867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760952083 Chr.1: 170989836-170989836 on Build GRCh38
rs913951879 Chr.1: 170990111-170990111 on Build GRCh38
rs754591722 Chr.1: 170990106-170990106 on Build GRCh38
rs28634500 Chr.1: 170989947-170989947 on Build GRCh38
rs570669210 Chr.1: 170990096-170990096 on Build GRCh38
rs1043936283 Chr.1: 170990138-170990138 on Build GRCh38
rs556055352 Chr.1: 170990152-170990152 on Build GRCh38
rs150307319 Chr.1: 170990086-170990086 on Build GRCh38
rs201767264 Chr.1: 170990097-170990097 on Build GRCh38
rs200783486 Chr.1: 170990015-170990015 on Build GRCh38
rs375294245 Chr.1: 170990051-170990051 on Build GRCh38
rs753574467 Chr.1: 170990042-170990042 on Build GRCh38
rs763561147 Chr.1: 170990012-170990012 on Build GRCh38
rs766660433 Chr.1: 170989855-170989855 on Build GRCh38
rs980671603 Chr.1: 170990056-170990056 on Build GRCh38
rs574389769 Chr.1: 170990156-170990156 on Build GRCh38
rs748171829 Chr.1: 170989952-170989952 on Build GRCh38
rs756598818 Chr.1: 170989974-170989974 on Build GRCh38
rs927332047 Chr.1: 170990233-170990233 on Build GRCh38
rs1015210881 Chr.1: 170990061-170990061 on Build GRCh38
rs935266539 Chr.1: 170989840-170989840 on Build GRCh38
rs945442503 Chr.1: 170990132-170990132 on Build GRCh38
rs925174155 Chr.1: 170989801-170989801 on Build GRCh38
rs376992226 Chr.1: 170989921-170989921 on Build GRCh38
rs115940585 Chr.1: 170989926-170989926 on Build GRCh38
rs759800504 Chr.1: 170990002-170990002 on Build GRCh38
rs747187726 Chr.1: 170989976-170989976 on Build GRCh38
rs962187415 Chr.1: 170990003-170990003 on Build GRCh38
rs935192178 Chr.1: 170990140-170990140 on Build GRCh38
rs780916786 Chr.1: 170989904-170989904 on Build GRCh38
rs749974210 Chr.1: 170990018-170990018 on Build GRCh38
rs905949585 Chr.1: 170989954-170989954 on Build GRCh38
rs374949615 Chr.1: 170989942-170989942 on Build GRCh38
rs754543717 Chr.1: 170990009-170990010 on Build GRCh38
rs1056562886 Chr.1: 170990088-170990088 on Build GRCh38
rs16863926 Chr.1: 170990167-170990167 on Build GRCh38
rs200507519 Chr.1: 170989996-170989996 on Build GRCh38
rs753299518 Chr.1: 170989870-170989870 on Build GRCh38
rs140210961 Chr.1: 170990084-170990084 on Build GRCh38
rs776956950 Chr.1: 170990001-170990001 on Build GRCh38
rs755786681 Chr.1: 170990035-170990035 on Build GRCh38
rs772184998 Chr.1: 170989809-170989809 on Build GRCh38
rs775856958 Chr.1: 170990011-170990011 on Build GRCh38
rs765634568 Chr.1: 170989867-170989867 on Build GRCh38
rs765691368 Chr.1: 170990009-170990009 on Build GRCh38
rs551877320 Chr.1: 170990038-170990038 on Build GRCh38
rs754179012 Chr.1: 170989857-170989857 on Build GRCh38
rs371280477 Chr.1: 170989906-170989906 on Build GRCh38
rs746471353 Chr.1: 170990103-170990103 on Build GRCh38
rs746079987 Chr.1: 170989975-170989975 on Build GRCh38
rs1045737820 Chr.1: 170990217-170990217 on Build GRCh38
rs966097381 Chr.1: 170990072-170990072 on Build GRCh38
rs771267291 Chr.1: 170990216-170990216 on Build GRCh38
rs145863395 Chr.1: 170990192-170990192 on Build GRCh38
rs772129951 Chr.1: 170989968-170989968 on Build GRCh38
rs16863922 Chr.1: 170989888-170989888 on Build GRCh38
rs756708951 Chr.1: 170989886-170989886 on Build GRCh38
rs751120860 Chr.1: 170989889-170989890 on Build GRCh38
rs755845428 Chr.1: 170989925-170989925 on Build GRCh38
rs773405697 Chr.1: 170989822-170989822 on Build GRCh38
rs749679455 Chr.1: 170990186-170990186 on Build GRCh38
rs760051072 Chr.1: 170989861-170989861 on Build GRCh38

View Full Product Details