Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172551382-172551880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
677128 c.689C>G p.S230* Chr.1: 172551550-172551550 on Build GRCh38
677126 c.689C>T p.S230L Chr.1: 172551550-172551550 on Build GRCh38
3802869 c.718G>C p.D240H Chr.1: 172551579-172551579 on Build GRCh38
69925 c.154G>A p.E52K Chr.1: 172551603-172551603 on Build GRCh38
677124 c.725A>G p.N242S Chr.1: 172551586-172551586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977464259 Chr.1: 172551830-172551830 on Build GRCh38
rs775685143 Chr.1: 172551659-172551659 on Build GRCh38
rs556430269 Chr.1: 172551817-172551817 on Build GRCh38
rs770144062 Chr.1: 172551491-172551491 on Build GRCh38
rs376930360 Chr.1: 172551590-172551590 on Build GRCh38
rs779387200 Chr.1: 172551488-172551488 on Build GRCh38
rs748018623 Chr.1: 172551612-172551612 on Build GRCh38
rs781670721 Chr.1: 172551495-172551496 on Build GRCh38
rs74124237 Chr.1: 172551444-172551444 on Build GRCh38
rs956598306 Chr.1: 172551674-172551674 on Build GRCh38
rs148137461 Chr.1: 172551559-172551559 on Build GRCh38
rs3213552 Chr.1: 172551825-172551825 on Build GRCh38
rs992627441 Chr.1: 172551595-172551595 on Build GRCh38
rs765528399 Chr.1: 172551556-172551556 on Build GRCh38
rs111691518 Chr.1: 172551657-172551657 on Build GRCh38
rs114783081 Chr.1: 172551526-172551526 on Build GRCh38
rs929273332 Chr.1: 172551715-172551715 on Build GRCh38
rs754177821 Chr.1: 172551475-172551475 on Build GRCh38
rs969129718 Chr.1: 172551617-172551617 on Build GRCh38
rs1047734584 Chr.1: 172551752-172551752 on Build GRCh38
rs1017682400 Chr.1: 172551785-172551785 on Build GRCh38
rs115406795 Chr.1: 172551540-172551540 on Build GRCh38
rs770927729 Chr.1: 172551649-172551649 on Build GRCh38
rs779716477 Chr.1: 172551588-172551588 on Build GRCh38
rs758953678 Chr.1: 172551494-172551494 on Build GRCh38
rs780516653 Chr.1: 172551497-172551497 on Build GRCh38
rs113997299 Chr.1: 172551552-172551552 on Build GRCh38
rs867334442 Chr.1: 172551773-172551773 on Build GRCh38
rs964890111 Chr.1: 172551504-172551504 on Build GRCh38
rs374074256 Chr.1: 172551611-172551611 on Build GRCh38
rs531645449 Chr.1: 172551414-172551414 on Build GRCh38
rs755393213 Chr.1: 172551480-172551480 on Build GRCh38
rs574102791 Chr.1: 172551792-172551792 on Build GRCh38
rs748397904 Chr.1: 172551672-172551673 on Build GRCh38
rs765219307 Chr.1: 172551730-172551730 on Build GRCh38
rs75755648 Chr.1: 172551516-172551516 on Build GRCh38
rs376480955 Chr.1: 172551742-172551742 on Build GRCh38
rs200545553 Chr.1: 172551501-172551501 on Build GRCh38
rs2981376 Chr.1: 172551807-172551807 on Build GRCh38
rs773596355 Chr.1: 172551507-172551507 on Build GRCh38
rs924334958 Chr.1: 172551591-172551591 on Build GRCh38
rs116719025 Chr.1: 172551578-172551578 on Build GRCh38
rs1037670775 Chr.1: 172551408-172551408 on Build GRCh38
rs760855663 Chr.1: 172551530-172551530 on Build GRCh38
rs779914580 Chr.1: 172551511-172551511 on Build GRCh38
rs764699418 Chr.1: 172551576-172551576 on Build GRCh38
rs954809338 Chr.1: 172551456-172551456 on Build GRCh38
rs142695292 Chr.1: 172551656-172551656 on Build GRCh38
rs763435159 Chr.1: 172551673-172551673 on Build GRCh38
rs768503138 Chr.1: 172551517-172551517 on Build GRCh38
rs773257366 Chr.1: 172551633-172551633 on Build GRCh38
rs890504909 Chr.1: 172551757-172551757 on Build GRCh38
rs774301527 Chr.1: 172551518-172551518 on Build GRCh38
rs761607303 Chr.1: 172551519-172551519 on Build GRCh38
rs915173371 Chr.1: 172551784-172551784 on Build GRCh38
rs754704496 Chr.1: 172551592-172551592 on Build GRCh38
rs138683830 Chr.1: 172551409-172551409 on Build GRCh38
rs1031350226 Chr.1: 172551835-172551835 on Build GRCh38
rs969087499 Chr.1: 172551786-172551786 on Build GRCh38
rs771884822 Chr.1: 172551619-172551619 on Build GRCh38
rs565786532 Chr.1: 172551650-172551650 on Build GRCh38
rs776904538 Chr.1: 172551546-172551546 on Build GRCh38
rs761823545 Chr.1: 172551510-172551510 on Build GRCh38
rs758790849 Chr.1: 172551560-172551560 on Build GRCh38
rs536189965 Chr.1: 172551435-172551435 on Build GRCh38
rs201186467 Chr.1: 172551525-172551525 on Build GRCh38
rs899113540 Chr.1: 172551701-172551701 on Build GRCh38
rs978785844 Chr.1: 172551801-172551801 on Build GRCh38
rs747084478 Chr.1: 172551647-172551647 on Build GRCh38
rs145307638 Chr.1: 172551580-172551580 on Build GRCh38
rs922016110 Chr.1: 172551803-172551803 on Build GRCh38

View Full Product Details