Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172557123-172557669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848973 c.1052C>T p.P351L Chr.1: 172557411-172557411 on Build GRCh38
1185652 c.1084G>T p.V362L Chr.1: 172557669-172557669 on Build GRCh38
3478397 c.1044C>T p.V348V Chr.1: 172557403-172557403 on Build GRCh38
899645 c.931A>C p.K311Q Chr.1: 172557290-172557290 on Build GRCh38
6413469 c.583C>T p.L195F Chr.1: 172557645-172557645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774327744 Chr.1: 172557571-172557571 on Build GRCh38
rs753654115 Chr.1: 172557300-172557300 on Build GRCh38
rs942498224 Chr.1: 172557584-172557584 on Build GRCh38
rs1044499681 Chr.1: 172557415-172557415 on Build GRCh38
rs996298244 Chr.1: 172557625-172557625 on Build GRCh38
rs73034011 Chr.1: 172557569-172557569 on Build GRCh38
rs973675270 Chr.1: 172557496-172557496 on Build GRCh38
rs374512283 Chr.1: 172557366-172557366 on Build GRCh38
rs1053744808 Chr.1: 172557510-172557510 on Build GRCh38
rs771704327 Chr.1: 172557392-172557392 on Build GRCh38
rs893027768 Chr.1: 172557624-172557624 on Build GRCh38
rs764196099 Chr.1: 172557627-172557627 on Build GRCh38
rs771446183 Chr.1: 172557234-172557234 on Build GRCh38
rs145242307 Chr.1: 172557297-172557297 on Build GRCh38
rs555030690 Chr.1: 172557394-172557394 on Build GRCh38
rs764371218 Chr.1: 172557623-172557623 on Build GRCh38
rs575899039 Chr.1: 172557592-172557592 on Build GRCh38
rs371813863 Chr.1: 172557405-172557405 on Build GRCh38
rs893068911 Chr.1: 172557245-172557245 on Build GRCh38
rs991685966 Chr.1: 172557402-172557402 on Build GRCh38
rs761415512 Chr.1: 172557284-172557284 on Build GRCh38
rs765302682 Chr.1: 172557452-172557452 on Build GRCh38
rs558031111 Chr.1: 172557181-172557181 on Build GRCh38
rs1039555319 Chr.1: 172557621-172557622 on Build GRCh38
rs200787187 Chr.1: 172557306-172557306 on Build GRCh38
rs760382528 Chr.1: 172557421-172557421 on Build GRCh38
rs752850850 Chr.1: 172557308-172557308 on Build GRCh38
rs767198141 Chr.1: 172557288-172557288 on Build GRCh38
rs781630987 Chr.1: 172557363-172557363 on Build GRCh38
rs759503320 Chr.1: 172557447-172557447 on Build GRCh38
rs747709918 Chr.1: 172557373-172557373 on Build GRCh38
rs770570819 Chr.1: 172557433-172557433 on Build GRCh38
rs566374577 Chr.1: 172557210-172557210 on Build GRCh38
rs778364413 Chr.1: 172557372-172557372 on Build GRCh38
rs573702217 Chr.1: 172557513-172557513 on Build GRCh38
rs907342245 Chr.1: 172557417-172557417 on Build GRCh38
rs748825932 Chr.1: 172557250-172557250 on Build GRCh38
rs778188388 Chr.1: 172557334-172557334 on Build GRCh38
rs963599700 Chr.1: 172557469-172557469 on Build GRCh38
rs544278293 Chr.1: 172557514-172557514 on Build GRCh38
rs751737570 Chr.1: 172557340-172557340 on Build GRCh38
rs983340190 Chr.1: 172557641-172557641 on Build GRCh38
rs1003047241 Chr.1: 172557431-172557431 on Build GRCh38
rs1011546647 Chr.1: 172557247-172557247 on Build GRCh38
rs758659135 Chr.1: 172557309-172557309 on Build GRCh38
rs763183210 Chr.1: 172557607-172557607 on Build GRCh38
rs573728513 Chr.1: 172557511-172557511 on Build GRCh38
rs777060446 Chr.1: 172557235-172557235 on Build GRCh38
rs765356073 Chr.1: 172557307-172557307 on Build GRCh38
rs773875676 Chr.1: 172557266-172557266 on Build GRCh38
rs757628124 Chr.1: 172557354-172557354 on Build GRCh38
rs768098011 Chr.1: 172557257-172557257 on Build GRCh38
rs533766789 Chr.1: 172557371-172557371 on Build GRCh38
rs1040012088 Chr.1: 172557200-172557200 on Build GRCh38
rs116807645 Chr.1: 172557410-172557410 on Build GRCh38
rs781372435 Chr.1: 172557631-172557631 on Build GRCh38
rs757522527 Chr.1: 172557628-172557628 on Build GRCh38
rs776505827 Chr.1: 172557437-172557437 on Build GRCh38
rs140836766 Chr.1: 172557348-172557348 on Build GRCh38
rs752801742 Chr.1: 172557604-172557604 on Build GRCh38
rs760448939 Chr.1: 172557299-172557299 on Build GRCh38

View Full Product Details