Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94755590-94756125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L4
Entrez Gene ID
286148
Gene Name
dpy-19 like 4 (C. elegans)
Gene Aliases
-
UniGene
Hs.567828
Gene Chromosome Location
Chr.8: 94719708-94793848 on Build GRCh38
Transcripts
NM_181787.2 XM_005250894.4 XM_005250895.3 XM_011516983.2 XM_011516984.2 XM_017013351.1 XM_017013352.1 XM_017013353.1 XM_017013354.1 XM_017013355.1 XM_017013356.1 XM_017013357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652004 c.638C>T p.S213F Chr.8: 94756062-94756062 on Build GRCh38
6649095 c.639C>A p.S213S Chr.8: 94756063-94756063 on Build GRCh38
7427794 c.669A>G p.P223P Chr.8: 94756093-94756093 on Build GRCh38
5885415 c.626G>A p.R209K Chr.8: 94756050-94756050 on Build GRCh38
244140 c.613G>T p.V205L Chr.8: 94756037-94756037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032816261 Chr.8: 94755661-94755661 on Build GRCh38
rs144933141 Chr.8: 94755667-94755667 on Build GRCh38
rs779172302 Chr.8: 94755950-94755950 on Build GRCh38
rs891560308 Chr.8: 94755796-94755796 on Build GRCh38
rs762233810 Chr.8: 94756053-94756053 on Build GRCh38
rs771734380 Chr.8: 94756042-94756042 on Build GRCh38
rs541448166 Chr.8: 94755868-94755868 on Build GRCh38
rs909291331 Chr.8: 94755755-94755755 on Build GRCh38
rs1036226426 Chr.8: 94755789-94755789 on Build GRCh38
rs373311763 Chr.8: 94756013-94756013 on Build GRCh38
rs575423779 Chr.8: 94755879-94755879 on Build GRCh38
rs780926679 Chr.8: 94755742-94755742 on Build GRCh38
rs555652654 Chr.8: 94755871-94755871 on Build GRCh38
rs1047736354 Chr.8: 94755704-94755704 on Build GRCh38
rs766480385 Chr.8: 94756068-94756068 on Build GRCh38
rs976631580 Chr.8: 94755876-94755876 on Build GRCh38
rs188046478 Chr.8: 94756000-94756000 on Build GRCh38
rs762448729 Chr.8: 94755776-94755776 on Build GRCh38
rs771975592 Chr.8: 94755872-94755872 on Build GRCh38
rs949647631 Chr.8: 94755820-94755820 on Build GRCh38
rs1013952410 Chr.8: 94755623-94755623 on Build GRCh38
rs751614882 Chr.8: 94756085-94756085 on Build GRCh38
rs938192798 Chr.8: 94755758-94755758 on Build GRCh38
rs906119197 Chr.8: 94755769-94755769 on Build GRCh38
rs79870348 Chr.8: 94756019-94756019 on Build GRCh38
rs756068996 Chr.8: 94755995-94755995 on Build GRCh38
rs765458064 Chr.8: 94756055-94756055 on Build GRCh38
rs745558151 Chr.8: 94756023-94756023 on Build GRCh38
rs1055418258 Chr.8: 94755618-94755618 on Build GRCh38
rs140869521 Chr.8: 94755637-94755637 on Build GRCh38
rs147304313 Chr.8: 94755622-94755622 on Build GRCh38
rs570569172 Chr.8: 94755757-94755757 on Build GRCh38
rs902960518 Chr.8: 94755638-94755638 on Build GRCh38
rs544399354 Chr.8: 94755895-94755895 on Build GRCh38
rs149045017 Chr.8: 94755839-94755839 on Build GRCh38
rs369859623 Chr.8: 94755990-94755990 on Build GRCh38
rs904265420 Chr.8: 94756092-94756092 on Build GRCh38
rs892879842 Chr.8: 94755918-94755918 on Build GRCh38
rs140964341 Chr.8: 94756064-94756064 on Build GRCh38
rs938244886 Chr.8: 94755617-94755617 on Build GRCh38
rs369769582 Chr.8: 94756093-94756093 on Build GRCh38
rs111452771 Chr.8: 94755702-94755702 on Build GRCh38
rs375038094 Chr.8: 94755753-94755753 on Build GRCh38
rs938911521 Chr.8: 94755931-94755931 on Build GRCh38
rs529248861 Chr.8: 94755665-94755665 on Build GRCh38
rs927549567 Chr.8: 94755901-94755901 on Build GRCh38
rs1003002540 Chr.8: 94755787-94755787 on Build GRCh38
rs563899032 Chr.8: 94755924-94755924 on Build GRCh38
rs773726403 Chr.8: 94756061-94756061 on Build GRCh38
rs184952946 Chr.8: 94755863-94755863 on Build GRCh38
rs1018238195 Chr.8: 94755871-94755872 on Build GRCh38
rs918387615 Chr.8: 94755880-94755880 on Build GRCh38
rs375681274 Chr.8: 94756047-94756047 on Build GRCh38
rs1017832338 Chr.8: 94755930-94755930 on Build GRCh38
rs558333451 Chr.8: 94755756-94755756 on Build GRCh38
rs919222199 Chr.8: 94755696-94755696 on Build GRCh38
rs147953831 Chr.8: 94755814-94755814 on Build GRCh38
rs891987105 Chr.8: 94755994-94755994 on Build GRCh38
rs957163509 Chr.8: 94755663-94755663 on Build GRCh38
rs200949180 Chr.8: 94756049-94756049 on Build GRCh38
rs942072385 Chr.8: 94755887-94755887 on Build GRCh38
rs984832986 Chr.8: 94755754-94755754 on Build GRCh38
rs867934496 Chr.8: 94756058-94756058 on Build GRCh38
rs749993212 Chr.8: 94755775-94755775 on Build GRCh38
rs368414296 Chr.8: 94755986-94755986 on Build GRCh38
rs1044339051 Chr.8: 94755890-94755890 on Build GRCh38
rs776919310 Chr.8: 94756052-94756052 on Build GRCh38
rs777769828 Chr.8: 94755997-94755997 on Build GRCh38
rs951234759 Chr.8: 94755882-94755882 on Build GRCh38
rs983966658 Chr.8: 94755884-94755885 on Build GRCh38
rs1007416536 Chr.8: 94755921-94755921 on Build GRCh38
rs372064939 Chr.8: 94755669-94755669 on Build GRCh38
rs773956232 Chr.8: 94756014-94756014 on Build GRCh38
rs909300460 Chr.8: 94755885-94755885 on Build GRCh38
rs998534298 Chr.8: 94755647-94755647 on Build GRCh38
rs966336350 Chr.8: 94755668-94755668 on Build GRCh38

View Full Product Details