Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 95244991-95245493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf37
Entrez Gene ID
157657
Gene Name
chromosome 8 open reading frame 37
Gene Aliases
BBS21, CORD16, RP64, smalltalk
UniGene
Hs.548157
Gene Chromosome Location
Chr.8: 95244913-95270602 on Build GRCh38
Transcripts
NM_177965.3 XM_005250799.3

SNP Detail

SNP ID Chromosome Location
rs187795112 Chr.8: 95245339-95245339 on Build GRCh38
rs202112088 Chr.8: 95245173-95245173 on Build GRCh38
rs79305170 Chr.8: 95245405-95245405 on Build GRCh38
rs980935659 Chr.8: 95245151-95245151 on Build GRCh38
rs569744735 Chr.8: 95245255-95245256 on Build GRCh38
rs1048179 Chr.8: 95245183-95245183 on Build GRCh38
rs977337782 Chr.8: 95245358-95245358 on Build GRCh38
rs992564141 Chr.8: 95245200-95245200 on Build GRCh38
rs1034533183 Chr.8: 95245185-95245185 on Build GRCh38
rs1018932637 Chr.8: 95245109-95245109 on Build GRCh38
rs74318654 Chr.8: 95245092-95245092 on Build GRCh38
rs896016322 Chr.8: 95245452-95245452 on Build GRCh38
rs746473742 Chr.8: 95245181-95245181 on Build GRCh38
rs933678237 Chr.8: 95245249-95245249 on Build GRCh38
rs937477028 Chr.8: 95245443-95245443 on Build GRCh38
rs150859301 Chr.8: 95245287-95245287 on Build GRCh38
rs945690647 Chr.8: 95245351-95245351 on Build GRCh38
rs759363792 Chr.8: 95245111-95245111 on Build GRCh38
rs149998549 Chr.8: 95245451-95245451 on Build GRCh38
rs74748655 Chr.8: 95245198-95245198 on Build GRCh38
rs374820930 Chr.8: 95245152-95245152 on Build GRCh38
rs1050244177 Chr.8: 95245400-95245400 on Build GRCh38
rs192180459 Chr.8: 95245371-95245371 on Build GRCh38
rs922484784 Chr.8: 95245205-95245205 on Build GRCh38
rs183510816 Chr.8: 95245101-95245101 on Build GRCh38
rs745781812 Chr.8: 95245068-95245068 on Build GRCh38
rs554334722 Chr.8: 95245119-95245119 on Build GRCh38
rs565495269 Chr.8: 95245062-95245062 on Build GRCh38
rs75940981 Chr.8: 95245454-95245454 on Build GRCh38
rs61209192 Chr.8: 95245423-95245423 on Build GRCh38
rs1027824734 Chr.8: 95245131-95245131 on Build GRCh38
rs935844627 Chr.8: 95245374-95245374 on Build GRCh38
rs952321050 Chr.8: 95245139-95245139 on Build GRCh38
rs529486839 Chr.8: 95245125-95245125 on Build GRCh38
rs544512199 Chr.8: 95245442-95245443 on Build GRCh38

View Full Product Details