Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 95263599-95264114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf37
Entrez Gene ID
157657
Gene Name
chromosome 8 open reading frame 37
Gene Aliases
BBS21, CORD16, RP64, smalltalk
UniGene
Hs.548157
Gene Chromosome Location
Chr.8: 95244913-95270602 on Build GRCh38
Transcripts
NM_177965.3 XM_005250799.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5552437 c.191A>G p.D64G Chr.8: 95263739-95263739 on Build GRCh38
1102741 c.158C>T p.S53L Chr.8: 95263772-95263772 on Build GRCh38
5015807 c.211C>G p.L71V Chr.8: 95263719-95263719 on Build GRCh38
7138563 c.205G>A p.E69K Chr.8: 95263725-95263725 on Build GRCh38
1283849 c.156-1G>T p.? Chr.8: 95263775-95263775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372060493 Chr.8: 95263684-95263684 on Build GRCh38
rs942984443 Chr.8: 95263885-95263885 on Build GRCh38
rs7835921 Chr.8: 95264064-95264064 on Build GRCh38
rs758882277 Chr.8: 95263723-95263723 on Build GRCh38
rs765908663 Chr.8: 95263712-95263712 on Build GRCh38
rs1002257896 Chr.8: 95263781-95263781 on Build GRCh38
rs980446408 Chr.8: 95263758-95263758 on Build GRCh38
rs758931781 Chr.8: 95263694-95263694 on Build GRCh38
rs1040122106 Chr.8: 95263641-95263641 on Build GRCh38
rs565660329 Chr.8: 95263886-95263886 on Build GRCh38
rs747285368 Chr.8: 95263747-95263747 on Build GRCh38
rs1008501508 Chr.8: 95263953-95263953 on Build GRCh38
rs749037313 Chr.8: 95263803-95263803 on Build GRCh38
rs770746137 Chr.8: 95263805-95263805 on Build GRCh38
rs781320258 Chr.8: 95263784-95263784 on Build GRCh38
rs905230931 Chr.8: 95264015-95264015 on Build GRCh38
rs1049763461 Chr.8: 95263942-95263942 on Build GRCh38
rs757821731 Chr.8: 95263711-95263711 on Build GRCh38
rs765997213 Chr.8: 95263687-95263687 on Build GRCh38
rs1016283049 Chr.8: 95263980-95263980 on Build GRCh38
rs754238230 Chr.8: 95263698-95263698 on Build GRCh38
rs368806728 Chr.8: 95263797-95263797 on Build GRCh38
rs187988455 Chr.8: 95263870-95263870 on Build GRCh38
rs1008447795 Chr.8: 95263628-95263628 on Build GRCh38
rs1002143309 Chr.8: 95264030-95264030 on Build GRCh38
rs780373034 Chr.8: 95263746-95263746 on Build GRCh38
rs865984138 Chr.8: 95263729-95263729 on Build GRCh38
rs530174452 Chr.8: 95263811-95263811 on Build GRCh38
rs571233450 Chr.8: 95264003-95264003 on Build GRCh38
rs769781357 Chr.8: 95263675-95263675 on Build GRCh38
rs879607806 Chr.8: 95263976-95263976 on Build GRCh38
rs1015775150 Chr.8: 95263664-95263664 on Build GRCh38
rs969339085 Chr.8: 95263736-95263736 on Build GRCh38
rs774078559 Chr.8: 95263688-95263688 on Build GRCh38
rs527703861 Chr.8: 95263645-95263645 on Build GRCh38
rs142541651 Chr.8: 95263823-95263823 on Build GRCh38
rs74652296 Chr.8: 95263785-95263785 on Build GRCh38
rs145542989 Chr.8: 95263719-95263719 on Build GRCh38
rs551400585 Chr.8: 95263936-95263936 on Build GRCh38
rs372022413 Chr.8: 95263637-95263637 on Build GRCh38
rs996894867 Chr.8: 95263912-95263912 on Build GRCh38
rs10107484 Chr.8: 95263667-95263667 on Build GRCh38
rs754956516 Chr.8: 95263756-95263756 on Build GRCh38
rs772986421 Chr.8: 95263676-95263676 on Build GRCh38
rs116874509 Chr.8: 95264037-95264037 on Build GRCh38
rs1043557892 Chr.8: 95263874-95263874 on Build GRCh38
rs779256912 Chr.8: 95263693-95263693 on Build GRCh38
rs1035107515 Chr.8: 95264031-95264031 on Build GRCh38
rs769689540 Chr.8: 95263787-95263787 on Build GRCh38
rs891101029 Chr.8: 95263950-95263950 on Build GRCh38
rs752053068 Chr.8: 95264088-95264088 on Build GRCh38
rs375086899 Chr.8: 95263650-95263650 on Build GRCh38
rs533744581 Chr.8: 95264008-95264008 on Build GRCh38
rs767055943 Chr.8: 95263697-95263697 on Build GRCh38
rs913755039 Chr.8: 95263845-95263845 on Build GRCh38
rs897393589 Chr.8: 95263993-95263993 on Build GRCh38
rs750838114 Chr.8: 95263958-95263958 on Build GRCh38
rs755169866 Chr.8: 95263665-95263670 on Build GRCh38
rs941627683 Chr.8: 95264007-95264007 on Build GRCh38
rs767687820 Chr.8: 95263748-95263750 on Build GRCh38
rs571110891 Chr.8: 95263774-95263774 on Build GRCh38
rs568960419 Chr.8: 95263866-95263866 on Build GRCh38
rs939281036 Chr.8: 95263788-95263788 on Build GRCh38
rs534743353 Chr.8: 95263897-95263897 on Build GRCh38

View Full Product Details