Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96143450-96143956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF6
Entrez Gene ID
392255
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1
UniGene
Hs.492277
Gene Chromosome Location
Chr.8: 96142330-96160792 on Build GRCh38
Transcripts
NM_001001557.2

SNP Detail

SNP ID Chromosome Location
rs932354544 Chr.8: 96143647-96143647 on Build GRCh38
rs984235767 Chr.8: 96143740-96143740 on Build GRCh38
rs1008523077 Chr.8: 96143736-96143736 on Build GRCh38
rs945201046 Chr.8: 96143615-96143615 on Build GRCh38
rs539996889 Chr.8: 96143476-96143476 on Build GRCh38
rs928591416 Chr.8: 96143560-96143560 on Build GRCh38
rs531321032 Chr.8: 96143572-96143572 on Build GRCh38
rs192657480 Chr.8: 96143721-96143721 on Build GRCh38
rs528002823 Chr.8: 96143782-96143782 on Build GRCh38
rs567787852 Chr.8: 96143812-96143812 on Build GRCh38
rs549993103 Chr.8: 96143854-96143854 on Build GRCh38
rs1027526616 Chr.8: 96143838-96143838 on Build GRCh38
rs77181285 Chr.8: 96143479-96143479 on Build GRCh38
rs995691388 Chr.8: 96143835-96143835 on Build GRCh38
rs960102687 Chr.8: 96143718-96143718 on Build GRCh38
rs776489762 Chr.8: 96143637-96143637 on Build GRCh38
rs974182534 Chr.8: 96143508-96143508 on Build GRCh38
rs1035587712 Chr.8: 96143709-96143709 on Build GRCh38
rs940024191 Chr.8: 96143768-96143768 on Build GRCh38
rs980127007 Chr.8: 96143542-96143542 on Build GRCh38
rs886063196 Chr.8: 96143910-96143910 on Build GRCh38
rs765006954 Chr.8: 96143770-96143770 on Build GRCh38
rs1035980687 Chr.8: 96143916-96143916 on Build GRCh38
rs977279008 Chr.8: 96143694-96143694 on Build GRCh38
rs915761399 Chr.8: 96143723-96143723 on Build GRCh38
rs1042799995 Chr.8: 96143890-96143890 on Build GRCh38
rs569856936 Chr.8: 96143888-96143888 on Build GRCh38
rs569599062 Chr.8: 96143796-96143796 on Build GRCh38
rs562634886 Chr.8: 96143569-96143569 on Build GRCh38
rs1027083542 Chr.8: 96143509-96143509 on Build GRCh38
rs1043514292 Chr.8: 96143624-96143624 on Build GRCh38
rs114201878 Chr.8: 96143565-96143565 on Build GRCh38
rs73698505 Chr.8: 96143852-96143852 on Build GRCh38
rs185707933 Chr.8: 96143913-96143913 on Build GRCh38
rs992067681 Chr.8: 96143928-96143928 on Build GRCh38
rs902832718 Chr.8: 96143905-96143905 on Build GRCh38
rs149860626 Chr.8: 96143673-96143673 on Build GRCh38
rs908515537 Chr.8: 96143746-96143746 on Build GRCh38
rs80085212 Chr.8: 96143473-96143473 on Build GRCh38
rs913763920 Chr.8: 96143578-96143578 on Build GRCh38
rs376103537 Chr.8: 96143672-96143672 on Build GRCh38
rs1015841367 Chr.8: 96143742-96143742 on Build GRCh38
rs528954838 Chr.8: 96143529-96143529 on Build GRCh38
rs1035730550 Chr.8: 96143880-96143880 on Build GRCh38
rs1049413967 Chr.8: 96143658-96143658 on Build GRCh38
rs922887918 Chr.8: 96143828-96143828 on Build GRCh38
rs879001208 Chr.8: 96143652-96143652 on Build GRCh38

View Full Product Details