Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98093506-98094051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH5
Entrez Gene ID
203111
Gene Name
glutamate rich 5
Gene Aliases
C8orf47
UniGene
Hs.171455
Gene Chromosome Location
Chr.8: 98064522-98093610 on Build GRCh38
Transcripts
NM_001170806.1 NM_173549.2

SNP Detail

SNP ID Chromosome Location
rs190079492 Chr.8: 98093790-98093790 on Build GRCh38
rs928653636 Chr.8: 98093936-98093936 on Build GRCh38
rs893229122 Chr.8: 98093573-98093573 on Build GRCh38
rs988963851 Chr.8: 98093903-98093903 on Build GRCh38
rs73701241 Chr.8: 98093530-98093530 on Build GRCh38
rs957903240 Chr.8: 98093845-98093845 on Build GRCh38
rs551706928 Chr.8: 98093533-98093533 on Build GRCh38
rs1029456517 Chr.8: 98093550-98093550 on Build GRCh38
rs1042144294 Chr.8: 98093593-98093593 on Build GRCh38
rs913727853 Chr.8: 98093864-98093864 on Build GRCh38
rs982551170 Chr.8: 98093751-98093751 on Build GRCh38
rs894731065 Chr.8: 98094004-98094004 on Build GRCh38
rs554476445 Chr.8: 98093995-98093995 on Build GRCh38
rs932404203 Chr.8: 98093987-98093987 on Build GRCh38
rs995649059 Chr.8: 98093666-98093666 on Build GRCh38
rs1003999158 Chr.8: 98093646-98093646 on Build GRCh38
rs934855975 Chr.8: 98093899-98093899 on Build GRCh38
rs527956939 Chr.8: 98093640-98093640 on Build GRCh38
rs989321696 Chr.8: 98093859-98093859 on Build GRCh38
rs1056174697 Chr.8: 98093985-98093985 on Build GRCh38
rs946078880 Chr.8: 98093579-98093579 on Build GRCh38
rs780075088 Chr.8: 98093618-98093618 on Build GRCh38
rs370177354 Chr.8: 98093812-98093812 on Build GRCh38
rs186348769 Chr.8: 98093689-98093689 on Build GRCh38
rs748713069 Chr.8: 98093994-98093994 on Build GRCh38
rs76480147 Chr.8: 98093807-98093807 on Build GRCh38
rs981490883 Chr.8: 98093860-98093860 on Build GRCh38
rs536884161 Chr.8: 98093715-98093715 on Build GRCh38
rs535922619 Chr.8: 98094002-98094002 on Build GRCh38
rs75518287 Chr.8: 98093913-98093913 on Build GRCh38
rs1043797355 Chr.8: 98093969-98093969 on Build GRCh38
rs555980005 Chr.8: 98093717-98093717 on Build GRCh38
rs1034592365 Chr.8: 98093641-98093641 on Build GRCh38
rs1019724486 Chr.8: 98093805-98093805 on Build GRCh38
rs567299180 Chr.8: 98093624-98093624 on Build GRCh38
rs548133335 Chr.8: 98093683-98093683 on Build GRCh38
rs140223933 Chr.8: 98093907-98093907 on Build GRCh38
rs1015501480 Chr.8: 98093706-98093706 on Build GRCh38
rs755941656 Chr.8: 98093552-98093554 on Build GRCh38
rs894699865 Chr.8: 98093665-98093665 on Build GRCh38
rs777020728 Chr.8: 98093908-98093908 on Build GRCh38
rs970561355 Chr.8: 98093836-98093836 on Build GRCh38
rs569684085 Chr.8: 98093752-98093752 on Build GRCh38
rs571729184 Chr.8: 98093874-98093874 on Build GRCh38
rs899461159 Chr.8: 98093986-98093986 on Build GRCh38
rs995709157 Chr.8: 98094025-98094025 on Build GRCh38
rs1050839493 Chr.8: 98094003-98094003 on Build GRCh38
rs928791899 Chr.8: 98093865-98093865 on Build GRCh38
rs573403013 Chr.8: 98093815-98093815 on Build GRCh38

View Full Product Details