Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102838460-102838980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
UniGene
Hs.459106
Gene Chromosome Location
Chr.8: 102826302-102864200 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Gene Symbol
LOC107986873
Entrez Gene ID
107986873
Gene Name
forkhead box protein L2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 102810302-102863067 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346026 c.431G>T p.R144L Chr.8: 102838762-102838762 on Build GRCh38
6609180 c.431G>A p.R144H Chr.8: 102838762-102838762 on Build GRCh38
3833771 c.371C>T p.A124V Chr.8: 102838822-102838822 on Build GRCh38
5580317 c.313C>T p.P105S Chr.8: 102838880-102838880 on Build GRCh38
6609182 c.416T>C p.L139S Chr.8: 102838777-102838777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62522602 Chr.8: 102838668-102838668 on Build GRCh38
rs748852648 Chr.8: 102838859-102838859 on Build GRCh38
rs960783354 Chr.8: 102838728-102838728 on Build GRCh38
rs772776067 Chr.8: 102838735-102838735 on Build GRCh38
rs777195334 Chr.8: 102838852-102838852 on Build GRCh38
rs931628479 Chr.8: 102838895-102838895 on Build GRCh38
rs567170313 Chr.8: 102838657-102838657 on Build GRCh38
rs1023647916 Chr.8: 102838765-102838765 on Build GRCh38
rs777885961 Chr.8: 102838842-102838842 on Build GRCh38
rs776141664 Chr.8: 102838867-102838867 on Build GRCh38
rs771693363 Chr.8: 102838725-102838725 on Build GRCh38
rs747740829 Chr.8: 102838716-102838716 on Build GRCh38
rs910909375 Chr.8: 102838647-102838647 on Build GRCh38
rs904051367 Chr.8: 102838703-102838703 on Build GRCh38
rs944421915 Chr.8: 102838652-102838652 on Build GRCh38
rs769248037 Chr.8: 102838729-102838729 on Build GRCh38
rs1052304560 Chr.8: 102838687-102838687 on Build GRCh38
rs904101805 Chr.8: 102838940-102838940 on Build GRCh38
rs543344671 Chr.8: 102838663-102838663 on Build GRCh38
rs952168261 Chr.8: 102838935-102838935 on Build GRCh38
rs757864650 Chr.8: 102838513-102838515 on Build GRCh38
rs750936028 Chr.8: 102838636-102838636 on Build GRCh38
rs151270350 Chr.8: 102838819-102838819 on Build GRCh38
rs759030712 Chr.8: 102838732-102838733 on Build GRCh38
rs1035247001 Chr.8: 102838503-102838503 on Build GRCh38
rs765276991 Chr.8: 102838775-102838775 on Build GRCh38
rs935225757 Chr.8: 102838674-102838674 on Build GRCh38
rs764483089 Chr.8: 102838762-102838762 on Build GRCh38
rs765521005 Chr.8: 102838927-102838927 on Build GRCh38
rs763659703 Chr.8: 102838635-102838635 on Build GRCh38
rs781216249 Chr.8: 102838754-102838754 on Build GRCh38
rs761151867 Chr.8: 102838755-102838755 on Build GRCh38
rs952470347 Chr.8: 102838572-102838572 on Build GRCh38
rs144818576 Chr.8: 102838843-102838843 on Build GRCh38
rs921640537 Chr.8: 102838877-102838877 on Build GRCh38
rs140315608 Chr.8: 102838872-102838872 on Build GRCh38
rs750575685 Chr.8: 102838936-102838936 on Build GRCh38
rs750497005 Chr.8: 102838782-102838782 on Build GRCh38
rs146727615 Chr.8: 102838862-102838862 on Build GRCh38
rs962603067 Chr.8: 102838510-102838510 on Build GRCh38
rs898103027 Chr.8: 102838827-102838827 on Build GRCh38
rs920744391 Chr.8: 102838533-102838533 on Build GRCh38
rs761993454 Chr.8: 102838926-102838926 on Build GRCh38
rs769197838 Chr.8: 102838873-102838873 on Build GRCh38
rs369562933 Chr.8: 102838709-102838709 on Build GRCh38
rs763102393 Chr.8: 102838950-102838950 on Build GRCh38
rs371017657 Chr.8: 102838844-102838844 on Build GRCh38
rs1027836651 Chr.8: 102838890-102838890 on Build GRCh38
rs983844556 Chr.8: 102838577-102838577 on Build GRCh38
rs758885410 Chr.8: 102838658-102838658 on Build GRCh38
rs930458382 Chr.8: 102838619-102838619 on Build GRCh38
rs142294459 Chr.8: 102838763-102838763 on Build GRCh38
rs951921927 Chr.8: 102838839-102838839 on Build GRCh38
rs145369291 Chr.8: 102838876-102838876 on Build GRCh38
rs758365770 Chr.8: 102838789-102838789 on Build GRCh38
rs184518812 Chr.8: 102838700-102838700 on Build GRCh38
rs942304562 Chr.8: 102838654-102838654 on Build GRCh38
rs920496845 Chr.8: 102838838-102838838 on Build GRCh38
rs972298717 Chr.8: 102838533-102838535 on Build GRCh38
rs377694303 Chr.8: 102838712-102838712 on Build GRCh38
rs759999261 Chr.8: 102838741-102838741 on Build GRCh38
rs1062048 Chr.8: 102838824-102838824 on Build GRCh38
rs768043887 Chr.8: 102838753-102838753 on Build GRCh38
rs373433614 Chr.8: 102838705-102838705 on Build GRCh38

View Full Product Details