Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109336043-109336532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENY2
Entrez Gene ID
56943
Gene Name
ENY2, transcription and export complex 2 subunit
Gene Aliases
DC6, Sus1, e(y)2
UniGene
Hs.492555
Gene Chromosome Location
Chr.8: 109334323-109345960 on Build GRCh38
Transcripts
NM_001193557.1 NM_020189.5 NR_036471.1 NR_036472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4951160 c.57G>T p.K19N Chr.8: 109336178-109336178 on Build GRCh38
1454050 c.68C>T p.T23I Chr.8: 109336189-109336189 on Build GRCh38
3271670 c.11G>A p.S4N Chr.8: 109336132-109336132 on Build GRCh38
3884682 c.83G>A p.R28H Chr.8: 109336204-109336204 on Build GRCh38
3271671 c.29C>T p.A10V Chr.8: 109336150-109336150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759511430 Chr.8: 109336230-109336230 on Build GRCh38
rs529144878 Chr.8: 109336151-109336151 on Build GRCh38
rs376011554 Chr.8: 109336086-109336086 on Build GRCh38
rs888505432 Chr.8: 109336462-109336462 on Build GRCh38
rs989130683 Chr.8: 109336378-109336378 on Build GRCh38
rs368357396 Chr.8: 109336232-109336232 on Build GRCh38
rs750806375 Chr.8: 109336150-109336150 on Build GRCh38
rs562639905 Chr.8: 109336166-109336166 on Build GRCh38
rs945280422 Chr.8: 109336394-109336394 on Build GRCh38
rs1030615394 Chr.8: 109336203-109336203 on Build GRCh38
rs567863210 Chr.8: 109336332-109336332 on Build GRCh38
rs754316009 Chr.8: 109336165-109336165 on Build GRCh38
rs779980893 Chr.8: 109336226-109336226 on Build GRCh38
rs374512551 Chr.8: 109336179-109336179 on Build GRCh38
rs998325087 Chr.8: 109336416-109336416 on Build GRCh38
rs766926945 Chr.8: 109336164-109336164 on Build GRCh38
rs72682697 Chr.8: 109336420-109336420 on Build GRCh38
rs191754604 Chr.8: 109336507-109336507 on Build GRCh38
rs969227551 Chr.8: 109336280-109336280 on Build GRCh38
rs957700597 Chr.8: 109336481-109336481 on Build GRCh38
rs557558939 Chr.8: 109336331-109336331 on Build GRCh38
rs776298915 Chr.8: 109336086-109336086 on Build GRCh38
rs1036980504 Chr.8: 109336480-109336480 on Build GRCh38
rs897439159 Chr.8: 109336483-109336483 on Build GRCh38
rs184813509 Chr.8: 109336262-109336262 on Build GRCh38
rs1005531966 Chr.8: 109336478-109336478 on Build GRCh38
rs192510487 Chr.8: 109336088-109336088 on Build GRCh38
rs900016140 Chr.8: 109336412-109336412 on Build GRCh38
rs187155945 Chr.8: 109336348-109336348 on Build GRCh38
rs909867082 Chr.8: 109336357-109336357 on Build GRCh38
rs763020916 Chr.8: 109336104-109336104 on Build GRCh38
rs768738607 Chr.8: 109336238-109336238 on Build GRCh38
rs766143769 Chr.8: 109336079-109336079 on Build GRCh38
rs767904373 Chr.8: 109336133-109336133 on Build GRCh38
rs970213724 Chr.8: 109336286-109336287 on Build GRCh38
rs537144936 Chr.8: 109336318-109336318 on Build GRCh38
rs932466126 Chr.8: 109336437-109336437 on Build GRCh38
rs1030646424 Chr.8: 109336467-109336467 on Build GRCh38
rs913850605 Chr.8: 109336391-109336391 on Build GRCh38
rs930037506 Chr.8: 109336327-109336327 on Build GRCh38
rs760727131 Chr.8: 109336414-109336414 on Build GRCh38
rs761913745 Chr.8: 109336130-109336130 on Build GRCh38
rs772422344 Chr.8: 109336244-109336244 on Build GRCh38
rs957985131 Chr.8: 109336222-109336222 on Build GRCh38
rs980938006 Chr.8: 109336294-109336294 on Build GRCh38
rs138319872 Chr.8: 109336100-109336100 on Build GRCh38
rs982608997 Chr.8: 109336297-109336297 on Build GRCh38
rs571320053 Chr.8: 109336298-109336298 on Build GRCh38
rs941635849 Chr.8: 109336383-109336383 on Build GRCh38
rs751120372 Chr.8: 109336175-109336175 on Build GRCh38
rs890236712 Chr.8: 109336440-109336440 on Build GRCh38
rs901294332 Chr.8: 109336078-109336078 on Build GRCh38
rs1023514065 Chr.8: 109336509-109336509 on Build GRCh38
rs780932929 Chr.8: 109336092-109336092 on Build GRCh38
rs751164843 Chr.8: 109336450-109336450 on Build GRCh38
rs745656878 Chr.8: 109336213-109336213 on Build GRCh38
rs746664390 Chr.8: 109336087-109336087 on Build GRCh38
rs372434155 Chr.8: 109336241-109336241 on Build GRCh38
rs551489685 Chr.8: 109336293-109336293 on Build GRCh38
rs528132325 Chr.8: 109336285-109336285 on Build GRCh38
rs372926065 Chr.8: 109336123-109336123 on Build GRCh38
rs775471654 Chr.8: 109336103-109336103 on Build GRCh38
rs922429229 Chr.8: 109336425-109336425 on Build GRCh38
rs780879339 Chr.8: 109336204-109336204 on Build GRCh38
rs369182216 Chr.8: 109336242-109336242 on Build GRCh38
rs565079693 Chr.8: 109336072-109336072 on Build GRCh38
rs775334562 Chr.8: 109336095-109336095 on Build GRCh38
rs553676148 Chr.8: 109336404-109336404 on Build GRCh38
rs199896797 Chr.8: 109336114-109336114 on Build GRCh38
rs755904474 Chr.8: 109336225-109336225 on Build GRCh38

View Full Product Details