Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 112223565-112224109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
UniGene
Hs.91381
Gene Chromosome Location
Chr.8: 112222928-113437499 on Build GRCh38
Transcripts
NM_052900.2 NM_198123.1 NM_198124.1 XM_011516815.2 XM_011516816.2 XM_017013008.1 XM_017013009.1 XM_017013010.1 XM_017013011.1 XM_017013012.1

SNP Detail

SNP ID Chromosome Location
rs898162039 Chr.8: 112224066-112224066 on Build GRCh38
rs187117037 Chr.8: 112224046-112224046 on Build GRCh38
rs573140491 Chr.8: 112223853-112223853 on Build GRCh38
rs755796727 Chr.8: 112223968-112223968 on Build GRCh38
rs779929802 Chr.8: 112223990-112223990 on Build GRCh38
rs772706408 Chr.8: 112223683-112223683 on Build GRCh38
rs1038729698 Chr.8: 112223840-112223840 on Build GRCh38
rs986603966 Chr.8: 112223867-112223867 on Build GRCh38
rs1049191077 Chr.8: 112223903-112223903 on Build GRCh38
rs374942831 Chr.8: 112223786-112223786 on Build GRCh38
rs145554429 Chr.8: 112223656-112223656 on Build GRCh38
rs530601319 Chr.8: 112224013-112224013 on Build GRCh38
rs182827902 Chr.8: 112224014-112224014 on Build GRCh38
rs550288174 Chr.8: 112224018-112224018 on Build GRCh38
rs990861969 Chr.8: 112223666-112223666 on Build GRCh38
rs571649961 Chr.8: 112223744-112223744 on Build GRCh38
rs183697887 Chr.8: 112223620-112223620 on Build GRCh38
rs942875776 Chr.8: 112223826-112223826 on Build GRCh38
rs375197169 Chr.8: 112223699-112223699 on Build GRCh38
rs938868879 Chr.8: 112224015-112224015 on Build GRCh38
rs537184994 Chr.8: 112223677-112223677 on Build GRCh38
rs994474491 Chr.8: 112223602-112223602 on Build GRCh38
rs186219990 Chr.8: 112223976-112223976 on Build GRCh38
rs916676671 Chr.8: 112223721-112223721 on Build GRCh38
rs956770692 Chr.8: 112224012-112224012 on Build GRCh38
rs1034944801 Chr.8: 112223945-112223945 on Build GRCh38
rs1012101367 Chr.8: 112223959-112223959 on Build GRCh38
rs968109188 Chr.8: 112223970-112223970 on Build GRCh38
rs542596257 Chr.8: 112223817-112223817 on Build GRCh38
rs1051385187 Chr.8: 112223801-112223801 on Build GRCh38
rs563397723 Chr.8: 112224021-112224021 on Build GRCh38
rs547822019 Chr.8: 112223894-112223894 on Build GRCh38
rs988714473 Chr.8: 112223973-112223973 on Build GRCh38
rs534240246 Chr.8: 112223818-112223818 on Build GRCh38
rs914614054 Chr.8: 112223979-112223979 on Build GRCh38
rs1015239913 Chr.8: 112223766-112223766 on Build GRCh38
rs1027549237 Chr.8: 112223610-112223610 on Build GRCh38
rs774866879 Chr.8: 112223642-112223642 on Build GRCh38
rs565993920 Chr.8: 112223593-112223593 on Build GRCh38
rs867467501 Chr.8: 112224032-112224032 on Build GRCh38
rs979572078 Chr.8: 112223984-112223984 on Build GRCh38
rs572787409 Chr.8: 112224023-112224023 on Build GRCh38
rs190692163 Chr.8: 112224005-112224005 on Build GRCh38
rs556771838 Chr.8: 112223731-112223731 on Build GRCh38
rs946672827 Chr.8: 112223724-112223724 on Build GRCh38
rs930378567 Chr.8: 112224055-112224055 on Build GRCh38
rs1003415986 Chr.8: 112223613-112223613 on Build GRCh38
rs886138122 Chr.8: 112223925-112223925 on Build GRCh38
rs940979381 Chr.8: 112224025-112224025 on Build GRCh38
rs1005202669 Chr.8: 112223936-112223936 on Build GRCh38
rs995305511 Chr.8: 112223842-112223842 on Build GRCh38
rs1019561270 Chr.8: 112223955-112223955 on Build GRCh38
rs750385708 Chr.8: 112223843-112223843 on Build GRCh38
rs960567714 Chr.8: 112223657-112223657 on Build GRCh38
rs10505184 Chr.8: 112223644-112223644 on Build GRCh38

View Full Product Details