Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119928317-119928747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPTOR
Entrez Gene ID
64798
Gene Name
DEP domain containing MTOR-interacting protein
Gene Aliases
DEP.6, DEPDC6
UniGene
Hs.112981
Gene Chromosome Location
Chr.8: 119873655-120056202 on Build GRCh38
Transcripts
NM_001283012.1 NM_022783.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3896444 c.147T>C p.V49V Chr.8: 119928424-119928424 on Build GRCh38
6851951 c.284G>C p.R95P Chr.8: 119928561-119928561 on Build GRCh38
1095592 c.219G>T p.L73L Chr.8: 119928496-119928496 on Build GRCh38
3896445 c.283C>A p.R95R Chr.8: 119928560-119928560 on Build GRCh38
5942194 c.136G>A p.E46K Chr.8: 119928413-119928413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017077563 Chr.8: 119928432-119928432 on Build GRCh38
rs764932775 Chr.8: 119928440-119928440 on Build GRCh38
rs772835073 Chr.8: 119928442-119928446 on Build GRCh38
rs769104381 Chr.8: 119928583-119928583 on Build GRCh38
rs16893299 Chr.8: 119928412-119928412 on Build GRCh38
rs374411237 Chr.8: 119928362-119928362 on Build GRCh38
rs764904230 Chr.8: 119928347-119928347 on Build GRCh38
rs374195929 Chr.8: 119928557-119928557 on Build GRCh38
rs780378516 Chr.8: 119928573-119928573 on Build GRCh38
rs948590041 Chr.8: 119928550-119928550 on Build GRCh38
rs775505421 Chr.8: 119928498-119928498 on Build GRCh38
rs749445568 Chr.8: 119928576-119928576 on Build GRCh38
rs749726006 Chr.8: 119928499-119928499 on Build GRCh38
rs776303409 Chr.8: 119928521-119928521 on Build GRCh38
rs747175884 Chr.8: 119928437-119928437 on Build GRCh38
rs192306007 Chr.8: 119928690-119928690 on Build GRCh38
rs752278898 Chr.8: 119928532-119928532 on Build GRCh38
rs751318250 Chr.8: 119928560-119928560 on Build GRCh38
rs371284282 Chr.8: 119928606-119928606 on Build GRCh38
rs958389565 Chr.8: 119928646-119928646 on Build GRCh38
rs772718586 Chr.8: 119928592-119928592 on Build GRCh38
rs764663762 Chr.8: 119928402-119928402 on Build GRCh38
rs577576654 Chr.8: 119928636-119928636 on Build GRCh38
rs149224142 Chr.8: 119928473-119928473 on Build GRCh38
rs745902075 Chr.8: 119928494-119928494 on Build GRCh38
rs765007914 Chr.8: 119928529-119928529 on Build GRCh38
rs200534365 Chr.8: 119928506-119928506 on Build GRCh38
rs144874352 Chr.8: 119928438-119928438 on Build GRCh38
rs763167104 Chr.8: 119928381-119928381 on Build GRCh38
rs201319418 Chr.8: 119928492-119928492 on Build GRCh38
rs757210251 Chr.8: 119928561-119928561 on Build GRCh38
rs937108429 Chr.8: 119928374-119928374 on Build GRCh38
rs189004640 Chr.8: 119928472-119928472 on Build GRCh38
rs758124947 Chr.8: 119928455-119928455 on Build GRCh38
rs143993565 Chr.8: 119928427-119928427 on Build GRCh38
rs1018353484 Chr.8: 119928564-119928564 on Build GRCh38
rs374784228 Chr.8: 119928649-119928649 on Build GRCh38
rs1005767855 Chr.8: 119928647-119928647 on Build GRCh38
rs762378087 Chr.8: 119928511-119928511 on Build GRCh38
rs775250669 Chr.8: 119928600-119928600 on Build GRCh38
rs200438859 Chr.8: 119928373-119928373 on Build GRCh38
rs768884561 Chr.8: 119928503-119928503 on Build GRCh38
rs143382933 Chr.8: 119928493-119928493 on Build GRCh38
rs368372088 Chr.8: 119928587-119928587 on Build GRCh38
rs62528677 Chr.8: 119928634-119928634 on Build GRCh38
rs774968647 Chr.8: 119928505-119928505 on Build GRCh38
rs370834328 Chr.8: 119928593-119928593 on Build GRCh38
rs571325842 Chr.8: 119928474-119928474 on Build GRCh38
rs770019857 Chr.8: 119928497-119928497 on Build GRCh38
rs991249309 Chr.8: 119928398-119928398 on Build GRCh38
rs768096221 Chr.8: 119928517-119928517 on Build GRCh38
rs961456412 Chr.8: 119928466-119928466 on Build GRCh38
rs769166563 Chr.8: 119928599-119928599 on Build GRCh38
rs377750134 Chr.8: 119928369-119928369 on Build GRCh38
rs201759684 Chr.8: 119928586-119928586 on Build GRCh38
rs551593761 Chr.8: 119928462-119928462 on Build GRCh38
rs370751115 Chr.8: 119928528-119928528 on Build GRCh38
rs202200535 Chr.8: 119928504-119928504 on Build GRCh38
rs745949118 Chr.8: 119928566-119928566 on Build GRCh38
rs760207105 Chr.8: 119928560-119928564 on Build GRCh38
rs994291619 Chr.8: 119928701-119928701 on Build GRCh38
rs1005713882 Chr.8: 119928431-119928431 on Build GRCh38
rs1041618560 Chr.8: 119928463-119928463 on Build GRCh38
rs370601820 Chr.8: 119928375-119928375 on Build GRCh38
rs28461628 Chr.8: 119928627-119928627 on Build GRCh38
rs752543031 Chr.8: 119928448-119928448 on Build GRCh38
rs758264282 Chr.8: 119928552-119928552 on Build GRCh38
rs925752212 Chr.8: 119928574-119928574 on Build GRCh38
rs1035497414 Chr.8: 119928702-119928702 on Build GRCh38
rs140263695 Chr.8: 119928417-119928417 on Build GRCh38

View Full Product Details