Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120008892-120009303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPTOR
Entrez Gene ID
64798
Gene Name
DEP domain containing MTOR-interacting protein
Gene Aliases
DEP.6, DEPDC6
UniGene
Hs.112981
Gene Chromosome Location
Chr.8: 119873655-120056202 on Build GRCh38
Transcripts
NM_001283012.1 NM_022783.3 XM_017013735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367180 c.1005T>C p.G335G Chr.8: 120009037-120009037 on Build GRCh38
6179623 c.1011_1012GG>TT p.V338F Chr.8: 120009043-120009044 on Build GRCh38
6367860 c.1063G>A p.V355I Chr.8: 120009095-120009095 on Build GRCh38
6179622 c.1079C>T p.P360L Chr.8: 120009111-120009111 on Build GRCh38
1095594 c.1010C>T p.A337V Chr.8: 120009042-120009042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041518391 Chr.8: 120008923-120008923 on Build GRCh38
rs768075265 Chr.8: 120009134-120009134 on Build GRCh38
rs1009674696 Chr.8: 120009206-120009206 on Build GRCh38
rs962444119 Chr.8: 120009231-120009231 on Build GRCh38
rs546279612 Chr.8: 120009119-120009119 on Build GRCh38
rs867975860 Chr.8: 120008936-120008936 on Build GRCh38
rs759683552 Chr.8: 120008997-120008997 on Build GRCh38
rs903000597 Chr.8: 120008930-120008930 on Build GRCh38
rs758084166 Chr.8: 120009171-120009171 on Build GRCh38
rs1019519752 Chr.8: 120008966-120008966 on Build GRCh38
rs751954770 Chr.8: 120009124-120009124 on Build GRCh38
rs16893420 Chr.8: 120009223-120009223 on Build GRCh38
rs573865495 Chr.8: 120008989-120008989 on Build GRCh38
rs372070231 Chr.8: 120008987-120008987 on Build GRCh38
rs560129457 Chr.8: 120008921-120008922 on Build GRCh38
rs752101211 Chr.8: 120009164-120009164 on Build GRCh38
rs1001436992 Chr.8: 120009238-120009238 on Build GRCh38
rs141807460 Chr.8: 120009245-120009245 on Build GRCh38
rs201220982 Chr.8: 120009060-120009060 on Build GRCh38
rs763265051 Chr.8: 120009015-120009015 on Build GRCh38
rs777647166 Chr.8: 120009064-120009064 on Build GRCh38
rs760699568 Chr.8: 120008991-120008991 on Build GRCh38
rs182345570 Chr.8: 120009000-120009000 on Build GRCh38
rs747255387 Chr.8: 120009042-120009042 on Build GRCh38
rs751818546 Chr.8: 120009020-120009020 on Build GRCh38
rs778427689 Chr.8: 120009153-120009153 on Build GRCh38
rs539466463 Chr.8: 120009275-120009275 on Build GRCh38
rs371180983 Chr.8: 120009141-120009141 on Build GRCh38
rs148059116 Chr.8: 120008992-120008992 on Build GRCh38
rs752944549 Chr.8: 120009002-120009002 on Build GRCh38
rs781608833 Chr.8: 120009038-120009038 on Build GRCh38
rs750530440 Chr.8: 120009126-120009126 on Build GRCh38
rs770827189 Chr.8: 120009183-120009183 on Build GRCh38
rs887758973 Chr.8: 120009179-120009179 on Build GRCh38
rs1008517628 Chr.8: 120008937-120008937 on Build GRCh38
rs569495849 Chr.8: 120009265-120009265 on Build GRCh38
rs899723976 Chr.8: 120009165-120009165 on Build GRCh38
rs769961499 Chr.8: 120009100-120009100 on Build GRCh38
rs527411947 Chr.8: 120009082-120009082 on Build GRCh38
rs991722982 Chr.8: 120009276-120009276 on Build GRCh38
rs370014084 Chr.8: 120009118-120009118 on Build GRCh38
rs958932679 Chr.8: 120009268-120009268 on Build GRCh38
rs374650683 Chr.8: 120008996-120008996 on Build GRCh38
rs759737444 Chr.8: 120009094-120009094 on Build GRCh38
rs1036283786 Chr.8: 120009249-120009249 on Build GRCh38
rs1006215709 Chr.8: 120009207-120009207 on Build GRCh38
rs147712866 Chr.8: 120009066-120009066 on Build GRCh38
rs200067665 Chr.8: 120009040-120009040 on Build GRCh38
rs544954637 Chr.8: 120009043-120009043 on Build GRCh38
rs6998857 Chr.8: 120009177-120009177 on Build GRCh38
rs527750843 Chr.8: 120008956-120008956 on Build GRCh38
rs202059022 Chr.8: 120009022-120009022 on Build GRCh38
rs201511583 Chr.8: 120008986-120008986 on Build GRCh38
rs754525841 Chr.8: 120009142-120009142 on Build GRCh38
rs764548838 Chr.8: 120009019-120009019 on Build GRCh38
rs142466508 Chr.8: 120009076-120009076 on Build GRCh38
rs947134442 Chr.8: 120008913-120008913 on Build GRCh38
rs189470920 Chr.8: 120009176-120009176 on Build GRCh38
rs754476014 Chr.8: 120009041-120009041 on Build GRCh38

View Full Product Details