Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123013759-123014268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL1
Entrez Gene ID
79139
Gene Name
derlin 1
Gene Aliases
DER-1, DER1
UniGene
Hs.241576
Gene Chromosome Location
Chr.8: 123013164-123042423 on Build GRCh38
Transcripts
NM_001134671.2 NM_024295.5 XM_006716657.1 XM_011517299.1

SNP Detail

SNP ID Chromosome Location
rs1021241128 Chr.8: 123014005-123014005 on Build GRCh38
rs1045927885 Chr.8: 123014207-123014207 on Build GRCh38
rs915979663 Chr.8: 123014086-123014086 on Build GRCh38
rs991779036 Chr.8: 123014116-123014116 on Build GRCh38
rs919457628 Chr.8: 123014126-123014126 on Build GRCh38
rs1010726088 Chr.8: 123013985-123013985 on Build GRCh38
rs1020706489 Chr.8: 123014222-123014222 on Build GRCh38
rs972148385 Chr.8: 123014061-123014061 on Build GRCh38
rs1032972104 Chr.8: 123013852-123013852 on Build GRCh38
rs544088225 Chr.8: 123013877-123013877 on Build GRCh38
rs143284667 Chr.8: 123014153-123014153 on Build GRCh38
rs780340003 Chr.8: 123013913-123013913 on Build GRCh38
rs953243633 Chr.8: 123013952-123013952 on Build GRCh38
rs903260564 Chr.8: 123013826-123013826 on Build GRCh38
rs371745888 Chr.8: 123014073-123014074 on Build GRCh38
rs764605204 Chr.8: 123014234-123014234 on Build GRCh38
rs916010843 Chr.8: 123013859-123013859 on Build GRCh38
rs938699678 Chr.8: 123013832-123013832 on Build GRCh38
rs575461623 Chr.8: 123014014-123014014 on Build GRCh38
rs960003269 Chr.8: 123014060-123014060 on Build GRCh38
rs569328800 Chr.8: 123014173-123014173 on Build GRCh38
rs920245701 Chr.8: 123014077-123014077 on Build GRCh38
rs183837228 Chr.8: 123014041-123014041 on Build GRCh38
rs192912283 Chr.8: 123014040-123014040 on Build GRCh38
rs961731358 Chr.8: 123014043-123014043 on Build GRCh38
rs149167014 Chr.8: 123014100-123014100 on Build GRCh38
rs531976781 Chr.8: 123014217-123014217 on Build GRCh38
rs966650739 Chr.8: 123014025-123014025 on Build GRCh38
rs1045877943 Chr.8: 123013803-123013803 on Build GRCh38
rs1037860055 Chr.8: 123013882-123013882 on Build GRCh38
rs78558664 Chr.8: 123013904-123013904 on Build GRCh38
rs1007208105 Chr.8: 123014203-123014203 on Build GRCh38
rs1017336379 Chr.8: 123013963-123013963 on Build GRCh38
rs746086878 Chr.8: 123013838-123013838 on Build GRCh38
rs893721034 Chr.8: 123013866-123013866 on Build GRCh38
rs533082002 Chr.8: 123013925-123013925 on Build GRCh38
rs977441778 Chr.8: 123013786-123013786 on Build GRCh38
rs924690778 Chr.8: 123014219-123014219 on Build GRCh38
rs375712170 Chr.8: 123013838-123013839 on Build GRCh38
rs1051433563 Chr.8: 123013941-123013941 on Build GRCh38
rs890725626 Chr.8: 123014178-123014178 on Build GRCh38
rs957791118 Chr.8: 123013969-123013969 on Build GRCh38
rs893552348 Chr.8: 123014209-123014209 on Build GRCh38

View Full Product Details