Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123182420-123182922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83A
Entrez Gene ID
84985
Gene Name
family with sequence similarity 83 member A
Gene Aliases
BJ-TSA-9
UniGene
Hs.379821
Gene Chromosome Location
Chr.8: 123179047-123210079 on Build GRCh38
Transcripts
NM_001288587.2 NM_001321630.1 NM_032899.6 NM_207006.2 XM_005251087.2 XM_011517338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649162 c.57C>T p.S19S Chr.8: 123182913-123182913 on Build GRCh38
4649161 c.57C>T p.S19S Chr.8: 123182913-123182913 on Build GRCh38
4896961 c.66C>T p.V22V Chr.8: 123182922-123182922 on Build GRCh38
4649160 c.57C>T p.S19S Chr.8: 123182913-123182913 on Build GRCh38
4896959 c.66C>T p.V22V Chr.8: 123182922-123182922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057080617 Chr.8: 123182537-123182537 on Build GRCh38
rs766681376 Chr.8: 123182587-123182587 on Build GRCh38
rs183639465 Chr.8: 123182649-123182649 on Build GRCh38
rs764320016 Chr.8: 123182880-123182880 on Build GRCh38
rs3739291 Chr.8: 123182728-123182728 on Build GRCh38
rs923978850 Chr.8: 123182778-123182778 on Build GRCh38
rs763715707 Chr.8: 123182863-123182863 on Build GRCh38
rs150560576 Chr.8: 123182484-123182484 on Build GRCh38
rs771728659 Chr.8: 123182718-123182718 on Build GRCh38
rs753670829 Chr.8: 123182888-123182888 on Build GRCh38
rs747605371 Chr.8: 123182571-123182571 on Build GRCh38
rs540894291 Chr.8: 123182765-123182765 on Build GRCh38
rs927161513 Chr.8: 123182539-123182539 on Build GRCh38
rs934519092 Chr.8: 123182749-123182750 on Build GRCh38
rs996348022 Chr.8: 123182669-123182669 on Build GRCh38
rs756882891 Chr.8: 123182630-123182630 on Build GRCh38
rs557108903 Chr.8: 123182520-123182520 on Build GRCh38
rs766405780 Chr.8: 123182887-123182887 on Build GRCh38
rs779730009 Chr.8: 123182763-123182763 on Build GRCh38
rs967312563 Chr.8: 123182769-123182769 on Build GRCh38
rs1005103073 Chr.8: 123182842-123182842 on Build GRCh38
rs111237881 Chr.8: 123182850-123182850 on Build GRCh38
rs755245395 Chr.8: 123182822-123182822 on Build GRCh38
rs750359761 Chr.8: 123182726-123182726 on Build GRCh38
rs934444569 Chr.8: 123182531-123182531 on Build GRCh38
rs1042453051 Chr.8: 123182615-123182615 on Build GRCh38
rs1034557505 Chr.8: 123182745-123182745 on Build GRCh38
rs779242915 Chr.8: 123182894-123182894 on Build GRCh38
rs936799716 Chr.8: 123182631-123182631 on Build GRCh38
rs375653911 Chr.8: 123182485-123182485 on Build GRCh38
rs1053880840 Chr.8: 123182811-123182811 on Build GRCh38
rs754866325 Chr.8: 123182893-123182893 on Build GRCh38
rs548962438 Chr.8: 123182782-123182782 on Build GRCh38
rs771164210 Chr.8: 123182875-123182875 on Build GRCh38
rs113019555 Chr.8: 123182538-123182538 on Build GRCh38
rs973192647 Chr.8: 123182737-123182737 on Build GRCh38
rs947108460 Chr.8: 123182507-123182507 on Build GRCh38
rs570595703 Chr.8: 123182838-123182838 on Build GRCh38
rs371708485 Chr.8: 123182818-123182818 on Build GRCh38
rs767790522 Chr.8: 123182464-123182464 on Build GRCh38
rs975482757 Chr.8: 123182475-123182475 on Build GRCh38
rs750316321 Chr.8: 123182874-123182874 on Build GRCh38
rs917202072 Chr.8: 123182757-123182757 on Build GRCh38
rs920946466 Chr.8: 123182670-123182670 on Build GRCh38
rs552712981 Chr.8: 123182618-123182618 on Build GRCh38
rs141153009 Chr.8: 123182900-123182900 on Build GRCh38
rs1054334894 Chr.8: 123182643-123182643 on Build GRCh38
rs887190681 Chr.8: 123182802-123182802 on Build GRCh38
rs774575483 Chr.8: 123182530-123182530 on Build GRCh38
rs1019905077 Chr.8: 123182649-123182649 on Build GRCh38
rs376459350 Chr.8: 123182819-123182819 on Build GRCh38
rs536891385 Chr.8: 123182519-123182519 on Build GRCh38
rs773725908 Chr.8: 123182846-123182846 on Build GRCh38
rs539364167 Chr.8: 123182864-123182864 on Build GRCh38
rs190150841 Chr.8: 123182761-123182761 on Build GRCh38
rs748214447 Chr.8: 123182827-123182827 on Build GRCh38
rs139320109 Chr.8: 123182767-123182767 on Build GRCh38
rs975137312 Chr.8: 123182652-123182652 on Build GRCh38
rs776858049 Chr.8: 123182858-123182858 on Build GRCh38
rs539696046 Chr.8: 123182613-123182613 on Build GRCh38
rs968352784 Chr.8: 123182721-123182721 on Build GRCh38
rs763588808 Chr.8: 123182591-123182591 on Build GRCh38
rs992745137 Chr.8: 123182608-123182608 on Build GRCh38
rs16898097 Chr.8: 123182794-123182794 on Build GRCh38
rs758554435 Chr.8: 123182835-123182835 on Build GRCh38
rs749417122 Chr.8: 123182865-123182865 on Build GRCh38
rs761090130 Chr.8: 123182542-123182542 on Build GRCh38
rs995567440 Chr.8: 123182570-123182570 on Build GRCh38
rs544862703 Chr.8: 123182744-123182744 on Build GRCh38
rs1036412818 Chr.8: 123182854-123182854 on Build GRCh38
rs902578660 Chr.8: 123182617-123182617 on Build GRCh38
rs28694222 Chr.8: 123182777-123182777 on Build GRCh38
rs747488253 Chr.8: 123182849-123182849 on Build GRCh38
rs541958395 Chr.8: 123182651-123182651 on Build GRCh38
rs779334649 Chr.8: 123182823-123182823 on Build GRCh38
rs186987067 Chr.8: 123182753-123182753 on Build GRCh38
rs11988157 Chr.8: 123182500-123182500 on Build GRCh38
rs762049179 Chr.8: 123182536-123182536 on Build GRCh38
rs954381552 Chr.8: 123182479-123182479 on Build GRCh38
rs1004485948 Chr.8: 123182696-123182696 on Build GRCh38
rs905173451 Chr.8: 123182563-123182563 on Build GRCh38
rs775154905 Chr.8: 123182607-123182607 on Build GRCh38
rs758450873 Chr.8: 123182895-123182895 on Build GRCh38
rs778579025 Chr.8: 123182695-123182695 on Build GRCh38
rs559388952 Chr.8: 123182692-123182692 on Build GRCh38
rs1030550025 Chr.8: 123182689-123182689 on Build GRCh38
rs965107989 Chr.8: 123182466-123182466 on Build GRCh38
rs766721985 Chr.8: 123182573-123182573 on Build GRCh38
rs779569027 Chr.8: 123182480-123182480 on Build GRCh38
rs900689722 Chr.8: 123182667-123182667 on Build GRCh38
rs555457531 Chr.8: 123182666-123182666 on Build GRCh38
rs759954523 Chr.8: 123182859-123182859 on Build GRCh38
rs767460616 Chr.8: 123182704-123182704 on Build GRCh38
rs924346632 Chr.8: 123182746-123182746 on Build GRCh38
rs368720159 Chr.8: 123182760-123182760 on Build GRCh38

View Full Product Details