Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123191724-123192228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83A
Entrez Gene ID
84985
Gene Name
family with sequence similarity 83 member A
Gene Aliases
BJ-TSA-9
UniGene
Hs.379821
Gene Chromosome Location
Chr.8: 123179047-123210079 on Build GRCh38
Transcripts
NM_001288587.2 NM_001321630.1 NM_032899.6 NM_207006.2 XM_005251087.2 XM_011517338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5531005 c.558G>A p.G186G Chr.8: 123191880-123191880 on Build GRCh38
3896535 c.481-2A>G p.? Chr.8: 123191801-123191801 on Build GRCh38
6179575 c.585G>T p.Q195H Chr.8: 123191907-123191907 on Build GRCh38
5414776 c.506T>C p.F169S Chr.8: 123191828-123191828 on Build GRCh38
3896534 c.481-2A>G p.? Chr.8: 123191801-123191801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535133469 Chr.8: 123191826-123191826 on Build GRCh38
rs757035932 Chr.8: 123191933-123191933 on Build GRCh38
rs2384984 Chr.8: 123192190-123192190 on Build GRCh38
rs574866560 Chr.8: 123191831-123191831 on Build GRCh38
rs751961401 Chr.8: 123191860-123191860 on Build GRCh38
rs754460595 Chr.8: 123191875-123191875 on Build GRCh38
rs989384253 Chr.8: 123192074-123192074 on Build GRCh38
rs543686126 Chr.8: 123192165-123192165 on Build GRCh38
rs1040748674 Chr.8: 123191902-123191902 on Build GRCh38
rs929731484 Chr.8: 123191920-123191920 on Build GRCh38
rs768716147 Chr.8: 123192043-123192043 on Build GRCh38
rs76206404 Chr.8: 123192064-123192064 on Build GRCh38
rs748105122 Chr.8: 123191777-123191777 on Build GRCh38
rs368256447 Chr.8: 123191917-123191917 on Build GRCh38
rs111653212 Chr.8: 123192022-123192022 on Build GRCh38
rs571659196 Chr.8: 123192054-123192054 on Build GRCh38
rs560862337 Chr.8: 123192081-123192081 on Build GRCh38
rs746096118 Chr.8: 123191938-123191938 on Build GRCh38
rs200388269 Chr.8: 123191936-123191936 on Build GRCh38
rs529782519 Chr.8: 123192082-123192082 on Build GRCh38
rs764932250 Chr.8: 123192016-123192016 on Build GRCh38
rs1017455900 Chr.8: 123192047-123192047 on Build GRCh38
rs1010289666 Chr.8: 123192118-123192118 on Build GRCh38
rs374457659 Chr.8: 123191983-123191983 on Build GRCh38
rs773684079 Chr.8: 123192171-123192171 on Build GRCh38
rs769790828 Chr.8: 123191832-123191832 on Build GRCh38
rs897517375 Chr.8: 123191972-123191972 on Build GRCh38
rs749439563 Chr.8: 123191979-123191979 on Build GRCh38
rs1035730628 Chr.8: 123191772-123191772 on Build GRCh38
rs766021438 Chr.8: 123191872-123191872 on Build GRCh38
rs201430634 Chr.8: 123191763-123191763 on Build GRCh38
rs867668517 Chr.8: 123191914-123191914 on Build GRCh38
rs753464344 Chr.8: 123191874-123191874 on Build GRCh38
rs759111520 Chr.8: 123192015-123192015 on Build GRCh38
rs979408962 Chr.8: 123191864-123191864 on Build GRCh38
rs1037947424 Chr.8: 123191940-123191940 on Build GRCh38
rs969271112 Chr.8: 123192161-123192161 on Build GRCh38
rs564546949 Chr.8: 123192036-123192036 on Build GRCh38
rs769845744 Chr.8: 123191955-123191955 on Build GRCh38
rs767794505 Chr.8: 123192010-123192010 on Build GRCh38
rs747452014 Chr.8: 123192037-123192037 on Build GRCh38
rs770191413 Chr.8: 123192133-123192133 on Build GRCh38
rs771792615 Chr.8: 123191781-123191781 on Build GRCh38
rs200869919 Chr.8: 123191842-123191842 on Build GRCh38
rs905143693 Chr.8: 123192020-123192020 on Build GRCh38
rs754485228 Chr.8: 123191762-123191762 on Build GRCh38
rs764767116 Chr.8: 123191854-123191854 on Build GRCh38
rs749119987 Chr.8: 123191852-123191852 on Build GRCh38
rs188113331 Chr.8: 123191997-123191997 on Build GRCh38
rs147466292 Chr.8: 123192011-123192011 on Build GRCh38
rs750592210 Chr.8: 123191933-123191933 on Build GRCh38
rs372509349 Chr.8: 123191982-123191982 on Build GRCh38
rs555146602 Chr.8: 123191830-123191830 on Build GRCh38
rs775708098 Chr.8: 123191974-123191974 on Build GRCh38
rs183187828 Chr.8: 123191794-123191794 on Build GRCh38
rs752559993 Chr.8: 123191906-123191906 on Build GRCh38
rs768933062 Chr.8: 123192004-123192005 on Build GRCh38
rs566046253 Chr.8: 123191821-123191821 on Build GRCh38
rs778297294 Chr.8: 123191876-123191876 on Build GRCh38
rs556300862 Chr.8: 123191924-123191924 on Build GRCh38
rs139389694 Chr.8: 123191865-123191865 on Build GRCh38
rs1006489166 Chr.8: 123191881-123191881 on Build GRCh38
rs369077900 Chr.8: 123191887-123191887 on Build GRCh38
rs1020353454 Chr.8: 123192126-123192126 on Build GRCh38
rs928715498 Chr.8: 123191785-123191785 on Build GRCh38
rs767627161 Chr.8: 123191906-123191907 on Build GRCh38
rs758325895 Chr.8: 123191912-123191912 on Build GRCh38
rs77950215 Chr.8: 123192179-123192179 on Build GRCh38
rs746778551 Chr.8: 123191922-123191922 on Build GRCh38
rs776084131 Chr.8: 123191839-123191839 on Build GRCh38
rs913420690 Chr.8: 123191843-123191843 on Build GRCh38

View Full Product Details