Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123206891-123207381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83A
Entrez Gene ID
84985
Gene Name
family with sequence similarity 83 member A
Gene Aliases
BJ-TSA-9
UniGene
Hs.379821
Gene Chromosome Location
Chr.8: 123179047-123210079 on Build GRCh38
Transcripts
NM_001288587.2 NM_032899.6 NM_207006.2 XM_005251087.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4162650 c.634C>A p.R212R Chr.8: 123207185-123207185 on Build GRCh38
3166178 c.759G>A p.P253P Chr.8: 123207310-123207310 on Build GRCh38
5408772 c.820T>C p.F274L Chr.8: 123207203-123207203 on Build GRCh38
4150508 c.705C>A p.A235A Chr.8: 123207256-123207256 on Build GRCh38
5408771 c.820T>C p.F274L Chr.8: 123207203-123207203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370437311 Chr.8: 123207233-123207233 on Build GRCh38
rs773721568 Chr.8: 123207120-123207120 on Build GRCh38
rs754046352 Chr.8: 123207172-123207172 on Build GRCh38
rs746571188 Chr.8: 123207314-123207314 on Build GRCh38
rs774094884 Chr.8: 123207217-123207217 on Build GRCh38
rs970541071 Chr.8: 123207020-123207020 on Build GRCh38
rs963262466 Chr.8: 123206983-123206983 on Build GRCh38
rs772844896 Chr.8: 123207313-123207313 on Build GRCh38
rs767328146 Chr.8: 123207153-123207153 on Build GRCh38
rs566874108 Chr.8: 123206940-123206940 on Build GRCh38
rs987851470 Chr.8: 123206964-123206966 on Build GRCh38
rs1017115935 Chr.8: 123206953-123206953 on Build GRCh38
rs781700373 Chr.8: 123207276-123207276 on Build GRCh38
rs752992647 Chr.8: 123207253-123207253 on Build GRCh38
rs761190376 Chr.8: 123207223-123207223 on Build GRCh38
rs980364501 Chr.8: 123207311-123207311 on Build GRCh38
rs34959620 Chr.8: 123207178-123207179 on Build GRCh38
rs771416766 Chr.8: 123207307-123207307 on Build GRCh38
rs760495296 Chr.8: 123207162-123207162 on Build GRCh38
rs750404160 Chr.8: 123207155-123207155 on Build GRCh38
rs559467537 Chr.8: 123206962-123206962 on Build GRCh38
rs781537157 Chr.8: 123207196-123207196 on Build GRCh38
rs771632137 Chr.8: 123207224-123207224 on Build GRCh38
rs571450011 Chr.8: 123206917-123206919 on Build GRCh38
rs757751864 Chr.8: 123207275-123207275 on Build GRCh38
rs759362452 Chr.8: 123207359-123207359 on Build GRCh38
rs928947272 Chr.8: 123207088-123207088 on Build GRCh38
rs764242212 Chr.8: 123207258-123207258 on Build GRCh38
rs746290015 Chr.8: 123207198-123207198 on Build GRCh38
rs1052591221 Chr.8: 123206919-123206919 on Build GRCh38
rs369984200 Chr.8: 123207106-123207106 on Build GRCh38
rs747848623 Chr.8: 123207304-123207304 on Build GRCh38
rs893572508 Chr.8: 123207152-123207152 on Build GRCh38
rs972959219 Chr.8: 123207027-123207027 on Build GRCh38
rs980479111 Chr.8: 123207038-123207038 on Build GRCh38
rs375115765 Chr.8: 123207178-123207178 on Build GRCh38
rs992498626 Chr.8: 123207097-123207097 on Build GRCh38
rs552436459 Chr.8: 123207021-123207021 on Build GRCh38
rs972769111 Chr.8: 123207246-123207246 on Build GRCh38
rs933456114 Chr.8: 123207216-123207216 on Build GRCh38
rs753286152 Chr.8: 123207299-123207300 on Build GRCh38
rs201332673 Chr.8: 123206911-123206919 on Build GRCh38
rs758353874 Chr.8: 123207254-123207254 on Build GRCh38
rs1011103610 Chr.8: 123207154-123207154 on Build GRCh38
rs1032485513 Chr.8: 123207261-123207261 on Build GRCh38
rs766289244 Chr.8: 123207165-123207165 on Build GRCh38
rs747530847 Chr.8: 123207186-123207186 on Build GRCh38
rs768367894 Chr.8: 123207115-123207115 on Build GRCh38
rs756535027 Chr.8: 123207291-123207291 on Build GRCh38
rs770966276 Chr.8: 123207325-123207325 on Build GRCh38
rs926339884 Chr.8: 123207089-123207089 on Build GRCh38
rs186087312 Chr.8: 123207265-123207265 on Build GRCh38
rs1028766741 Chr.8: 123207171-123207171 on Build GRCh38
rs776747428 Chr.8: 123207357-123207357 on Build GRCh38
rs746201861 Chr.8: 123207277-123207277 on Build GRCh38
rs199931629 Chr.8: 123207252-123207252 on Build GRCh38
rs766648885 Chr.8: 123207124-123207125 on Build GRCh38
rs766490491 Chr.8: 123207236-123207236 on Build GRCh38
rs761437034 Chr.8: 123207144-123207144 on Build GRCh38
rs142743626 Chr.8: 123207185-123207185 on Build GRCh38
rs779515522 Chr.8: 123207190-123207211 on Build GRCh38
rs917554175 Chr.8: 123206992-123206992 on Build GRCh38
rs867697826 Chr.8: 123207326-123207326 on Build GRCh38
rs373188549 Chr.8: 123207173-123207173 on Build GRCh38
rs1024675782 Chr.8: 123207286-123207286 on Build GRCh38
rs970806160 Chr.8: 123207309-123207309 on Build GRCh38
rs760692553 Chr.8: 123207235-123207235 on Build GRCh38
rs965733270 Chr.8: 123207232-123207232 on Build GRCh38
rs573187518 Chr.8: 123206998-123206998 on Build GRCh38
rs778379212 Chr.8: 123207295-123207295 on Build GRCh38
rs754164218 Chr.8: 123207134-123207137 on Build GRCh38
rs918870011 Chr.8: 123207082-123207082 on Build GRCh38
rs776549902 Chr.8: 123207248-123207248 on Build GRCh38
rs755408870 Chr.8: 123207162-123207163 on Build GRCh38
rs56221642 Chr.8: 123207229-123207229 on Build GRCh38
rs948211098 Chr.8: 123207324-123207324 on Build GRCh38
rs957155869 Chr.8: 123206911-123206916 on Build GRCh38
rs1032599880 Chr.8: 123206911-123206913 on Build GRCh38
rs892739913 Chr.8: 123207273-123207273 on Build GRCh38
rs768275935 Chr.8: 123207205-123207205 on Build GRCh38

View Full Product Details