Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123208589-123209099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83A
Entrez Gene ID
84985
Gene Name
family with sequence similarity 83 member A
Gene Aliases
BJ-TSA-9
UniGene
Hs.379821
Gene Chromosome Location
Chr.8: 123179047-123210079 on Build GRCh38
Transcripts
NM_001288587.2 NM_032899.6 NM_207006.2 XM_005251087.2

SNP Detail

SNP ID Chromosome Location
rs370992907 Chr.8: 123209061-123209061 on Build GRCh38
rs938639588 Chr.8: 123208911-123208911 on Build GRCh38
rs935862674 Chr.8: 123208965-123208965 on Build GRCh38
rs796374268 Chr.8: 123208966-123208966 on Build GRCh38
rs965660268 Chr.8: 123208689-123208689 on Build GRCh38
rs1010146960 Chr.8: 123208966-123208968 on Build GRCh38
rs752871147 Chr.8: 123208863-123208863 on Build GRCh38
rs950502120 Chr.8: 123208748-123208748 on Build GRCh38
rs542514312 Chr.8: 123208721-123208721 on Build GRCh38
rs796992368 Chr.8: 123208975-123208976 on Build GRCh38
rs896819078 Chr.8: 123208730-123208731 on Build GRCh38
rs575452575 Chr.8: 123208637-123208637 on Build GRCh38
rs979978797 Chr.8: 123208836-123208836 on Build GRCh38
rs371147342 Chr.8: 123208965-123208966 on Build GRCh38
rs868426445 Chr.8: 123208872-123208872 on Build GRCh38
rs889512814 Chr.8: 123208824-123208824 on Build GRCh38
rs781387044 Chr.8: 123208771-123208771 on Build GRCh38
rs113597854 Chr.8: 123208620-123208620 on Build GRCh38
rs945444153 Chr.8: 123209009-123209009 on Build GRCh38
rs982486406 Chr.8: 123209003-123209003 on Build GRCh38
rs1021412585 Chr.8: 123208981-123208981 on Build GRCh38
rs941601111 Chr.8: 123208720-123208720 on Build GRCh38
rs893088346 Chr.8: 123208937-123208937 on Build GRCh38
rs910152054 Chr.8: 123208690-123208690 on Build GRCh38
rs1056847043 Chr.8: 123208925-123208925 on Build GRCh38
rs74776346 Chr.8: 123208979-123208979 on Build GRCh38
rs891754544 Chr.8: 123209023-123209023 on Build GRCh38
rs1043797147 Chr.8: 123209052-123209052 on Build GRCh38
rs575427206 Chr.8: 123208964-123208965 on Build GRCh38
rs559849372 Chr.8: 123209008-123209008 on Build GRCh38
rs892505749 Chr.8: 123208930-123208930 on Build GRCh38
rs549343931 Chr.8: 123208980-123208980 on Build GRCh38
rs75927713 Chr.8: 123208967-123208967 on Build GRCh38
rs544454233 Chr.8: 123208712-123208712 on Build GRCh38
rs966738502 Chr.8: 123208988-123208988 on Build GRCh38
rs948543977 Chr.8: 123208805-123208805 on Build GRCh38
rs959423732 Chr.8: 123209016-123209016 on Build GRCh38
rs556002192 Chr.8: 123208980-123208981 on Build GRCh38
rs1026008595 Chr.8: 123208722-123208722 on Build GRCh38
rs536327689 Chr.8: 123208976-123208977 on Build GRCh38
rs183181995 Chr.8: 123208959-123208959 on Build GRCh38
rs909041198 Chr.8: 123208804-123208804 on Build GRCh38
rs1052958831 Chr.8: 123209019-123209019 on Build GRCh38

View Full Product Details