Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123320175-123320685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
UniGene
Hs.370834
Gene Chromosome Location
Chr.8: 123319850-123396992 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516996.2

SNP Detail

SNP ID Chromosome Location
rs117698329 Chr.8: 123320316-123320316 on Build GRCh38
rs1045508733 Chr.8: 123320356-123320357 on Build GRCh38
rs998666329 Chr.8: 123320530-123320530 on Build GRCh38
rs929303589 Chr.8: 123320204-123320204 on Build GRCh38
rs1006608757 Chr.8: 123320576-123320576 on Build GRCh38
rs1053702287 Chr.8: 123320306-123320306 on Build GRCh38
rs571480567 Chr.8: 123320635-123320635 on Build GRCh38
rs1015190081 Chr.8: 123320491-123320491 on Build GRCh38
rs112189199 Chr.8: 123320502-123320502 on Build GRCh38
rs936130369 Chr.8: 123320269-123320269 on Build GRCh38
rs1029737710 Chr.8: 123320615-123320615 on Build GRCh38
rs1044470525 Chr.8: 123320263-123320263 on Build GRCh38
rs112308623 Chr.8: 123320235-123320235 on Build GRCh38
rs1049004142 Chr.8: 123320385-123320385 on Build GRCh38
rs113170326 Chr.8: 123320492-123320492 on Build GRCh38
rs746062258 Chr.8: 123320620-123320620 on Build GRCh38
rs1001234820 Chr.8: 123320367-123320367 on Build GRCh38
rs1025553810 Chr.8: 123320638-123320638 on Build GRCh38
rs1011238418 Chr.8: 123320350-123320350 on Build GRCh38
rs185528108 Chr.8: 123320211-123320211 on Build GRCh38
rs889456530 Chr.8: 123320566-123320566 on Build GRCh38
rs78146742 Chr.8: 123320470-123320470 on Build GRCh38
rs4006508 Chr.8: 123320463-123320463 on Build GRCh38
rs961774428 Chr.8: 123320208-123320208 on Build GRCh38
rs954076301 Chr.8: 123320233-123320233 on Build GRCh38
rs907341099 Chr.8: 123320298-123320298 on Build GRCh38
rs954045535 Chr.8: 123320621-123320621 on Build GRCh38
rs4006507 Chr.8: 123320493-123320493 on Build GRCh38
rs9021 Chr.8: 123320660-123320660 on Build GRCh38
rs926173108 Chr.8: 123320636-123320636 on Build GRCh38
rs530182708 Chr.8: 123320212-123320212 on Build GRCh38
rs1019386006 Chr.8: 123320577-123320577 on Build GRCh38
rs887702405 Chr.8: 123320394-123320394 on Build GRCh38
rs4006509 Chr.8: 123320461-123320461 on Build GRCh38
rs1004690997 Chr.8: 123320424-123320424 on Build GRCh38
rs905513108 Chr.8: 123320364-123320364 on Build GRCh38
rs532639359 Chr.8: 123320593-123320593 on Build GRCh38
rs892072030 Chr.8: 123320333-123320333 on Build GRCh38
rs565474215 Chr.8: 123320324-123320324 on Build GRCh38

View Full Product Details