Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123500574-123501084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO32
Entrez Gene ID
114907
Gene Name
F-box protein 32
Gene Aliases
Fbx32, MAFbx
UniGene
Hs.403933
Gene Chromosome Location
Chr.8: 123497887-123541253 on Build GRCh38
Transcripts
NM_001242463.1 NM_058229.3 NM_148177.2

SNP Detail

SNP ID Chromosome Location
rs946874158 Chr.8: 123500678-123500678 on Build GRCh38
rs533989811 Chr.8: 123500957-123500957 on Build GRCh38
rs958338411 Chr.8: 123500876-123500876 on Build GRCh38
rs35157801 Chr.8: 123500672-123500672 on Build GRCh38
rs140824076 Chr.8: 123500841-123500842 on Build GRCh38
rs1010427837 Chr.8: 123500791-123500791 on Build GRCh38
rs567487259 Chr.8: 123500679-123500679 on Build GRCh38
rs934053396 Chr.8: 123500655-123500655 on Build GRCh38
rs911184563 Chr.8: 123500695-123500695 on Build GRCh38
rs552752786 Chr.8: 123501013-123501013 on Build GRCh38
rs1054684940 Chr.8: 123500725-123500725 on Build GRCh38
rs769120890 Chr.8: 123500842-123500842 on Build GRCh38
rs942962683 Chr.8: 123500702-123500702 on Build GRCh38
rs893854926 Chr.8: 123500831-123500831 on Build GRCh38
rs955442409 Chr.8: 123501023-123501023 on Build GRCh38
rs113676977 Chr.8: 123500687-123500687 on Build GRCh38
rs776385329 Chr.8: 123500735-123500735 on Build GRCh38
rs142807337 Chr.8: 123500913-123500913 on Build GRCh38
rs116819300 Chr.8: 123500921-123500921 on Build GRCh38
rs1052541429 Chr.8: 123500775-123500775 on Build GRCh38
rs1042633869 Chr.8: 123500686-123500686 on Build GRCh38
rs557780553 Chr.8: 123501025-123501025 on Build GRCh38
rs201180378 Chr.8: 123500694-123500694 on Build GRCh38
rs148518253 Chr.8: 123500875-123500875 on Build GRCh38
rs893364554 Chr.8: 123500737-123500737 on Build GRCh38
rs112882868 Chr.8: 123500808-123500808 on Build GRCh38
rs151024324 Chr.8: 123500920-123500920 on Build GRCh38
rs1002539360 Chr.8: 123500838-123500838 on Build GRCh38
rs1016207584 Chr.8: 123500907-123500907 on Build GRCh38
rs961728633 Chr.8: 123500936-123500936 on Build GRCh38
rs560625832 Chr.8: 123500661-123500661 on Build GRCh38
rs889395164 Chr.8: 123500914-123500914 on Build GRCh38
rs1042270826 Chr.8: 123500811-123500811 on Build GRCh38
rs906514343 Chr.8: 123500677-123500677 on Build GRCh38
rs774692342 Chr.8: 123500867-123500867 on Build GRCh38
rs972105876 Chr.8: 123500984-123500984 on Build GRCh38
rs1029088988 Chr.8: 123500884-123500884 on Build GRCh38
rs529614437 Chr.8: 123500663-123500663 on Build GRCh38
rs1027408160 Chr.8: 123500987-123500987 on Build GRCh38
rs923063988 Chr.8: 123500981-123500981 on Build GRCh38

View Full Product Details