Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123513107-123513607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO32
Entrez Gene ID
114907
Gene Name
F-box protein 32
Gene Aliases
Fbx32, MAFbx
UniGene
Hs.403933
Gene Chromosome Location
Chr.8: 123497887-123541253 on Build GRCh38
Transcripts
NM_001242463.1 NM_058229.3 NM_148177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3982384 c.606G>A p.T202T Chr.8: 123513243-123513243 on Build GRCh38
1095783 c.467-1G>A p.? Chr.8: 123513383-123513383 on Build GRCh38
7178155 c.513G>A p.Q171Q Chr.8: 123513336-123513336 on Build GRCh38
7157568 c.553G>A p.G185S Chr.8: 123513296-123513296 on Build GRCh38
1095782 c.593A>G p.Y198C Chr.8: 123513256-123513256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777521319 Chr.8: 123513558-123513558 on Build GRCh38
rs985278456 Chr.8: 123513453-123513454 on Build GRCh38
rs111273552 Chr.8: 123513197-123513197 on Build GRCh38
rs757137859 Chr.8: 123513421-123513421 on Build GRCh38
rs938169881 Chr.8: 123513253-123513253 on Build GRCh38
rs977991597 Chr.8: 123513445-123513445 on Build GRCh38
rs755830908 Chr.8: 123513533-123513533 on Build GRCh38
rs373138610 Chr.8: 123513266-123513266 on Build GRCh38
rs896851191 Chr.8: 123513310-123513310 on Build GRCh38
rs1031246918 Chr.8: 123513482-123513482 on Build GRCh38
rs982076376 Chr.8: 123513580-123513580 on Build GRCh38
rs867627212 Chr.8: 123513362-123513362 on Build GRCh38
rs752025647 Chr.8: 123513189-123513189 on Build GRCh38
rs533921831 Chr.8: 123513474-123513474 on Build GRCh38
rs1029541303 Chr.8: 123513389-123513389 on Build GRCh38
rs745728555 Chr.8: 123513276-123513276 on Build GRCh38
rs776038524 Chr.8: 123513396-123513397 on Build GRCh38
rs183614038 Chr.8: 123513483-123513483 on Build GRCh38
rs35117009 Chr.8: 123513297-123513297 on Build GRCh38
rs142379196 Chr.8: 123513214-123513214 on Build GRCh38
rs778776607 Chr.8: 123513229-123513229 on Build GRCh38
rs970866738 Chr.8: 123513408-123513408 on Build GRCh38
rs1037602883 Chr.8: 123513134-123513134 on Build GRCh38
rs989995513 Chr.8: 123513510-123513510 on Build GRCh38
rs754667846 Chr.8: 123513219-123513219 on Build GRCh38
rs777160528 Chr.8: 123513406-123513406 on Build GRCh38
rs768131818 Chr.8: 123513349-123513349 on Build GRCh38
rs756587760 Chr.8: 123513366-123513366 on Build GRCh38
rs775539277 Chr.8: 123513153-123513153 on Build GRCh38
rs1033230212 Chr.8: 123513473-123513473 on Build GRCh38
rs3739287 Chr.8: 123513243-123513243 on Build GRCh38
rs528657541 Chr.8: 123513246-123513246 on Build GRCh38
rs3739288 Chr.8: 123513137-123513137 on Build GRCh38
rs373616023 Chr.8: 123513353-123513353 on Build GRCh38
rs1008425331 Chr.8: 123513133-123513133 on Build GRCh38
rs559932736 Chr.8: 123513244-123513244 on Build GRCh38
rs376347679 Chr.8: 123513273-123513273 on Build GRCh38
rs375536292 Chr.8: 123513171-123513171 on Build GRCh38
rs139769356 Chr.8: 123513549-123513549 on Build GRCh38
rs770699985 Chr.8: 123513264-123513264 on Build GRCh38
rs551015582 Chr.8: 123513419-123513419 on Build GRCh38
rs774858364 Chr.8: 123513329-123513329 on Build GRCh38
rs775733561 Chr.8: 123513291-123513291 on Build GRCh38
rs1024707224 Chr.8: 123513350-123513350 on Build GRCh38
rs562900644 Chr.8: 123513145-123513145 on Build GRCh38
rs3892087 Chr.8: 123513306-123513306 on Build GRCh38
rs762242955 Chr.8: 123513333-123513333 on Build GRCh38
rs146542406 Chr.8: 123513374-123513374 on Build GRCh38
rs566656802 Chr.8: 123513301-123513301 on Build GRCh38
rs757604245 Chr.8: 123513192-123513192 on Build GRCh38
rs753487889 Chr.8: 123513215-123513215 on Build GRCh38
rs570824678 Chr.8: 123513451-123513451 on Build GRCh38
rs763655642 Chr.8: 123513404-123513407 on Build GRCh38
rs763427515 Chr.8: 123513397-123513397 on Build GRCh38
rs764963323 Chr.8: 123513370-123513370 on Build GRCh38
rs1011837108 Chr.8: 123513344-123513344 on Build GRCh38
rs553481249 Chr.8: 123513537-123513537 on Build GRCh38
rs267601762 Chr.8: 123513372-123513372 on Build GRCh38
rs757855713 Chr.8: 123513388-123513388 on Build GRCh38
rs200573696 Chr.8: 123513278-123513278 on Build GRCh38

View Full Product Details