Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123712572-123713068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA13
Entrez Gene ID
312
Gene Name
annexin A13
Gene Aliases
ANX13, ISA
UniGene
Hs.181107
Gene Chromosome Location
Chr.8: 123680794-123737426 on Build GRCh38
Transcripts
NM_001003954.2 NM_004306.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4534549 c.209G>A p.G70E Chr.8: 123712683-123712683 on Build GRCh38
404106 c.176G>A p.R59Q Chr.8: 123712716-123712716 on Build GRCh38
355317 c.201C>T p.A67A Chr.8: 123712691-123712691 on Build GRCh38
355316 c.203G>T p.C68F Chr.8: 123712689-123712689 on Build GRCh38
7302941 c.182C>T p.A61V Chr.8: 123712710-123712710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527809597 Chr.8: 123713038-123713038 on Build GRCh38
rs759997274 Chr.8: 123712668-123712668 on Build GRCh38
rs944715582 Chr.8: 123712978-123712978 on Build GRCh38
rs771008500 Chr.8: 123712775-123712775 on Build GRCh38
rs577182990 Chr.8: 123712821-123712821 on Build GRCh38
rs767161062 Chr.8: 123712697-123712697 on Build GRCh38
rs906005848 Chr.8: 123712752-123712752 on Build GRCh38
rs911095667 Chr.8: 123712613-123712613 on Build GRCh38
rs1018218818 Chr.8: 123712855-123712855 on Build GRCh38
rs773503182 Chr.8: 123712746-123712746 on Build GRCh38
rs772388067 Chr.8: 123712745-123712745 on Build GRCh38
rs895578582 Chr.8: 123712800-123712800 on Build GRCh38
rs749245581 Chr.8: 123712715-123712715 on Build GRCh38
rs1048221390 Chr.8: 123712717-123712717 on Build GRCh38
rs200830719 Chr.8: 123712683-123712683 on Build GRCh38
rs960343384 Chr.8: 123712832-123712832 on Build GRCh38
rs978607442 Chr.8: 123712597-123712597 on Build GRCh38
rs145656611 Chr.8: 123713028-123713028 on Build GRCh38
rs142216753 Chr.8: 123712742-123712742 on Build GRCh38
rs574263849 Chr.8: 123712906-123712906 on Build GRCh38
rs779471598 Chr.8: 123712708-123712708 on Build GRCh38
rs545067515 Chr.8: 123712980-123712980 on Build GRCh38
rs942050291 Chr.8: 123713005-123713005 on Build GRCh38
rs886687738 Chr.8: 123712741-123712741 on Build GRCh38
rs980262686 Chr.8: 123712893-123712893 on Build GRCh38
rs375255283 Chr.8: 123712793-123712793 on Build GRCh38
rs1040446207 Chr.8: 123712673-123712673 on Build GRCh38
rs992271149 Chr.8: 123712834-123712834 on Build GRCh38
rs983687381 Chr.8: 123712949-123712949 on Build GRCh38
rs368241219 Chr.8: 123712688-123712688 on Build GRCh38
rs778797279 Chr.8: 123712722-123712722 on Build GRCh38
rs576586646 Chr.8: 123712979-123712979 on Build GRCh38
rs756844002 Chr.8: 123712692-123712692 on Build GRCh38
rs113828413 Chr.8: 123712850-123712850 on Build GRCh38
rs375342008 Chr.8: 123712644-123712644 on Build GRCh38
rs562634698 Chr.8: 123712992-123712992 on Build GRCh38
rs776640854 Chr.8: 123712777-123712777 on Build GRCh38
rs1040815406 Chr.8: 123713019-123713019 on Build GRCh38
rs755630498 Chr.8: 123712707-123712707 on Build GRCh38
rs557902033 Chr.8: 123712839-123712839 on Build GRCh38
rs368850792 Chr.8: 123712792-123712792 on Build GRCh38
rs148699158 Chr.8: 123712716-123712716 on Build GRCh38
rs144665201 Chr.8: 123712680-123712680 on Build GRCh38
rs542963468 Chr.8: 123712929-123712929 on Build GRCh38
rs374578614 Chr.8: 123712789-123712789 on Build GRCh38
rs192954521 Chr.8: 123712964-123712964 on Build GRCh38
rs370718427 Chr.8: 123712627-123712627 on Build GRCh38
rs976607499 Chr.8: 123712865-123712865 on Build GRCh38
rs760837060 Chr.8: 123712642-123712642 on Build GRCh38
rs568208622 Chr.8: 123712891-123712891 on Build GRCh38
rs765581612 Chr.8: 123712675-123712675 on Build GRCh38
rs963850112 Chr.8: 123712862-123712862 on Build GRCh38
rs765745163 Chr.8: 123712783-123712783 on Build GRCh38
rs933436029 Chr.8: 123712925-123712925 on Build GRCh38
rs553848349 Chr.8: 123712892-123712892 on Build GRCh38
rs764675976 Chr.8: 123713031-123713031 on Build GRCh38
rs920774735 Chr.8: 123712900-123712900 on Build GRCh38
rs371324945 Chr.8: 123712773-123712773 on Build GRCh38
rs930858712 Chr.8: 123712946-123712946 on Build GRCh38
rs558920512 Chr.8: 123712813-123712813 on Build GRCh38
rs760022262 Chr.8: 123712782-123712782 on Build GRCh38
rs549119255 Chr.8: 123712667-123712667 on Build GRCh38
rs112304259 Chr.8: 123712838-123712838 on Build GRCh38
rs921531552 Chr.8: 123712698-123712698 on Build GRCh38
rs370017071 Chr.8: 123712791-123712791 on Build GRCh38
rs773361407 Chr.8: 123712631-123712631 on Build GRCh38

View Full Product Details