Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123735659-123736195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA13
Entrez Gene ID
312
Gene Name
annexin A13
Gene Aliases
ANX13, ISA
UniGene
Hs.181107
Gene Chromosome Location
Chr.8: 123680794-123737426 on Build GRCh38
Transcripts
NM_001003954.2 NM_004306.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834003 c.132A>C p.P44P Chr.8: 123735761-123735761 on Build GRCh38
6601301 c.114T>C p.N38N Chr.8: 123735779-123735779 on Build GRCh38
6601302 c.74C>A p.P25H Chr.8: 123735819-123735819 on Build GRCh38
5967874 c.62G>T p.G21V Chr.8: 123735831-123735831 on Build GRCh38
7132283 c.62G>A p.G21E Chr.8: 123735831-123735831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780064945 Chr.8: 123735939-123735939 on Build GRCh38
rs779028210 Chr.8: 123735717-123735717 on Build GRCh38
rs74368673 Chr.8: 123735847-123735847 on Build GRCh38
rs548690719 Chr.8: 123736092-123736092 on Build GRCh38
rs769061885 Chr.8: 123735791-123735791 on Build GRCh38
rs781300550 Chr.8: 123735819-123735819 on Build GRCh38
rs771114559 Chr.8: 123735738-123735738 on Build GRCh38
rs577109572 Chr.8: 123735902-123735902 on Build GRCh38
rs770511781 Chr.8: 123735886-123735887 on Build GRCh38
rs113405898 Chr.8: 123736170-123736170 on Build GRCh38
rs201094354 Chr.8: 123735783-123735783 on Build GRCh38
rs565031245 Chr.8: 123735844-123735844 on Build GRCh38
rs752646367 Chr.8: 123735812-123735812 on Build GRCh38
rs1021824838 Chr.8: 123735899-123735899 on Build GRCh38
rs764832836 Chr.8: 123735870-123735870 on Build GRCh38
rs759287150 Chr.8: 123735897-123735897 on Build GRCh38
rs964308236 Chr.8: 123735770-123735770 on Build GRCh38
rs202199207 Chr.8: 123735777-123735777 on Build GRCh38
rs373688763 Chr.8: 123735869-123735869 on Build GRCh38
rs777466725 Chr.8: 123735723-123735723 on Build GRCh38
rs1000258636 Chr.8: 123735974-123735974 on Build GRCh38
rs750639679 Chr.8: 123735924-123735924 on Build GRCh38
rs968224298 Chr.8: 123735733-123735733 on Build GRCh38
rs144650739 Chr.8: 123735816-123735816 on Build GRCh38
rs758240911 Chr.8: 123735722-123735722 on Build GRCh38
rs904842896 Chr.8: 123735824-123735824 on Build GRCh38
rs939625211 Chr.8: 123735795-123735795 on Build GRCh38
rs751328905 Chr.8: 123735904-123735904 on Build GRCh38
rs746138751 Chr.8: 123735822-123735822 on Build GRCh38
rs763940128 Chr.8: 123735901-123735901 on Build GRCh38
rs780177238 Chr.8: 123735838-123735838 on Build GRCh38
rs930095579 Chr.8: 123735806-123735806 on Build GRCh38
rs528315381 Chr.8: 123735713-123735714 on Build GRCh38
rs879792881 Chr.8: 123736106-123736106 on Build GRCh38
rs765903624 Chr.8: 123735712-123735712 on Build GRCh38
rs530802582 Chr.8: 123735815-123735815 on Build GRCh38
rs374772372 Chr.8: 123735813-123735813 on Build GRCh38
rs532281882 Chr.8: 123735908-123735908 on Build GRCh38
rs754898813 Chr.8: 123735716-123735716 on Build GRCh38
rs964006148 Chr.8: 123736034-123736034 on Build GRCh38
rs565557819 Chr.8: 123736003-123736003 on Build GRCh38
rs773732962 Chr.8: 123735732-123735733 on Build GRCh38
rs992145059 Chr.8: 123735805-123735805 on Build GRCh38
rs770573000 Chr.8: 123735863-123735863 on Build GRCh38
rs1009850452 Chr.8: 123735897-123735897 on Build GRCh38
rs370410241 Chr.8: 123735840-123735840 on Build GRCh38
rs199508519 Chr.8: 123735774-123735774 on Build GRCh38
rs769834262 Chr.8: 123735826-123735826 on Build GRCh38
rs753352598 Chr.8: 123735809-123735809 on Build GRCh38
rs770245157 Chr.8: 123735766-123735766 on Build GRCh38
rs377247795 Chr.8: 123735912-123735912 on Build GRCh38
rs747899164 Chr.8: 123735721-123735721 on Build GRCh38
rs748125685 Chr.8: 123736085-123736085 on Build GRCh38
rs983383262 Chr.8: 123736115-123736115 on Build GRCh38
rs916584818 Chr.8: 123736102-123736102 on Build GRCh38
rs746833787 Chr.8: 123735779-123735779 on Build GRCh38
rs762120273 Chr.8: 123735851-123735851 on Build GRCh38
rs978787727 Chr.8: 123735689-123735689 on Build GRCh38
rs978209521 Chr.8: 123735943-123735943 on Build GRCh38
rs547972674 Chr.8: 123736073-123736073 on Build GRCh38
rs759142736 Chr.8: 123735810-123735810 on Build GRCh38
rs151208437 Chr.8: 123735807-123735807 on Build GRCh38
rs747246829 Chr.8: 123735736-123735736 on Build GRCh38
rs752310860 Chr.8: 123735894-123735894 on Build GRCh38
rs534534061 Chr.8: 123736072-123736072 on Build GRCh38
rs202001254 Chr.8: 123735811-123735811 on Build GRCh38
rs375350220 Chr.8: 123735843-123735843 on Build GRCh38
rs372116424 Chr.8: 123735738-123735739 on Build GRCh38
rs745950892 Chr.8: 123735742-123735742 on Build GRCh38
rs568198014 Chr.8: 123736082-123736082 on Build GRCh38
rs781273901 Chr.8: 123735739-123735739 on Build GRCh38
rs780248172 Chr.8: 123735765-123735765 on Build GRCh38
rs200664573 Chr.8: 123735808-123735808 on Build GRCh38
rs746389255 Chr.8: 123735715-123735715 on Build GRCh38
rs1022420573 Chr.8: 123735724-123735724 on Build GRCh38
rs755790377 Chr.8: 123735711-123735711 on Build GRCh38
rs201099803 Chr.8: 123735887-123735887 on Build GRCh38
rs762888474 Chr.8: 123735896-123735896 on Build GRCh38

View Full Product Details