Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123775063-123775571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3995848 c.189C>A p.Y63* Chr.8: 123775178-123775178 on Build GRCh38
1095813 c.215G>T p.R72L Chr.8: 123775204-123775204 on Build GRCh38
1095812 c.207G>A p.K69K Chr.8: 123775196-123775196 on Build GRCh38
1095811 c.182G>A p.R61H Chr.8: 123775171-123775171 on Build GRCh38
389680 c.309G>T p.E103D Chr.8: 123775298-123775298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943935293 Chr.8: 123775414-123775414 on Build GRCh38
rs929378261 Chr.8: 123775434-123775434 on Build GRCh38
rs201580142 Chr.8: 123775285-123775285 on Build GRCh38
rs185534486 Chr.8: 123775347-123775347 on Build GRCh38
rs764256315 Chr.8: 123775121-123775121 on Build GRCh38
rs769599264 Chr.8: 123775195-123775195 on Build GRCh38
rs570796426 Chr.8: 123775368-123775368 on Build GRCh38
rs777204751 Chr.8: 123775171-123775171 on Build GRCh38
rs914244188 Chr.8: 123775373-123775373 on Build GRCh38
rs759403918 Chr.8: 123775098-123775098 on Build GRCh38
rs1016834312 Chr.8: 123775271-123775271 on Build GRCh38
rs762662910 Chr.8: 123775092-123775092 on Build GRCh38
rs946956745 Chr.8: 123775544-123775544 on Build GRCh38
rs1003510438 Chr.8: 123775533-123775533 on Build GRCh38
rs749464157 Chr.8: 123775218-123775218 on Build GRCh38
rs191570000 Chr.8: 123775169-123775169 on Build GRCh38
rs932286451 Chr.8: 123775388-123775388 on Build GRCh38
rs369536417 Chr.8: 123775426-123775427 on Build GRCh38
rs1049140323 Chr.8: 123775438-123775438 on Build GRCh38
rs370669475 Chr.8: 123775097-123775097 on Build GRCh38
rs777376602 Chr.8: 123775459-123775459 on Build GRCh38
rs202233843 Chr.8: 123775309-123775309 on Build GRCh38
rs369452533 Chr.8: 123775320-123775320 on Build GRCh38
rs78916591 Chr.8: 123775539-123775539 on Build GRCh38
rs757465424 Chr.8: 123775395-123775395 on Build GRCh38
rs986987130 Chr.8: 123775336-123775336 on Build GRCh38
rs200807355 Chr.8: 123775335-123775335 on Build GRCh38
rs565827221 Chr.8: 123775455-123775455 on Build GRCh38
rs745841222 Chr.8: 123775193-123775193 on Build GRCh38
rs750707539 Chr.8: 123775136-123775136 on Build GRCh38
rs993551792 Chr.8: 123775173-123775173 on Build GRCh38
rs756481945 Chr.8: 123775143-123775143 on Build GRCh38
rs368811106 Chr.8: 123775253-123775253 on Build GRCh38
rs1036879243 Chr.8: 123775417-123775417 on Build GRCh38
rs774534378 Chr.8: 123775238-123775238 on Build GRCh38
rs780606580 Chr.8: 123775330-123775330 on Build GRCh38
rs1021389529 Chr.8: 123775207-123775207 on Build GRCh38
rs955253403 Chr.8: 123775482-123775482 on Build GRCh38
rs896966789 Chr.8: 123775425-123775425 on Build GRCh38
rs375307232 Chr.8: 123775101-123775101 on Build GRCh38
rs1042353948 Chr.8: 123775394-123775394 on Build GRCh38
rs373616871 Chr.8: 123775389-123775389 on Build GRCh38
rs781210595 Chr.8: 123775190-123775190 on Build GRCh38
rs894036738 Chr.8: 123775372-123775372 on Build GRCh38
rs767545105 Chr.8: 123775246-123775246 on Build GRCh38
rs371825325 Chr.8: 123775318-123775318 on Build GRCh38
rs912063822 Chr.8: 123775353-123775353 on Build GRCh38
rs766757469 Chr.8: 123775284-123775284 on Build GRCh38
rs745813257 Chr.8: 123775334-123775334 on Build GRCh38
rs751702756 Chr.8: 123775134-123775134 on Build GRCh38
rs756126898 Chr.8: 123775325-123775335 on Build GRCh38
rs775214727 Chr.8: 123775201-123775201 on Build GRCh38
rs770864530 Chr.8: 123775096-123775096 on Build GRCh38
rs1023367912 Chr.8: 123775479-123775479 on Build GRCh38
rs769005370 Chr.8: 123775237-123775237 on Build GRCh38
rs774139830 Chr.8: 123775264-123775264 on Build GRCh38
rs766697822 Chr.8: 123775157-123775157 on Build GRCh38
rs1003624312 Chr.8: 123775398-123775398 on Build GRCh38
rs907479668 Chr.8: 123775501-123775501 on Build GRCh38
rs200887433 Chr.8: 123775120-123775120 on Build GRCh38
rs1008096251 Chr.8: 123775125-123775125 on Build GRCh38
rs768102914 Chr.8: 123775135-123775135 on Build GRCh38
rs990677816 Chr.8: 123775269-123775269 on Build GRCh38
rs374789729 Chr.8: 123775180-123775180 on Build GRCh38
rs963803345 Chr.8: 123775519-123775519 on Build GRCh38
rs943543652 Chr.8: 123775357-123775357 on Build GRCh38
rs76957815 Chr.8: 123775450-123775450 on Build GRCh38
rs911005670 Chr.8: 123775502-123775502 on Build GRCh38
rs756019847 Chr.8: 123775346-123775346 on Build GRCh38
rs200016935 Chr.8: 123775281-123775281 on Build GRCh38
rs532433627 Chr.8: 123775441-123775441 on Build GRCh38
rs897937012 Chr.8: 123775170-123775170 on Build GRCh38
rs989836179 Chr.8: 123775365-123775365 on Build GRCh38
rs762008355 Chr.8: 123775241-123775241 on Build GRCh38
rs775758402 Chr.8: 123775104-123775104 on Build GRCh38
rs375719750 Chr.8: 123775302-123775302 on Build GRCh38
rs981656328 Chr.8: 123775298-123775298 on Build GRCh38
rs759264186 Chr.8: 123775410-123775410 on Build GRCh38
rs959233207 Chr.8: 123775267-123775267 on Build GRCh38

View Full Product Details