Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130060885-130061394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887477 c.2722C>T p.H908Y Chr.8: 130061049-130061049 on Build GRCh38
3896751 c.2840C>T p.A947V Chr.8: 130060931-130060931 on Build GRCh38
6605421 c.2814C>T p.D938D Chr.8: 130060957-130060957 on Build GRCh38
3778903 c.2766G>A p.Q922Q Chr.8: 130061005-130061005 on Build GRCh38
7213669 c.2704G>A p.A902T Chr.8: 130061067-130061067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768671699 Chr.8: 130061016-130061016 on Build GRCh38
rs755592394 Chr.8: 130061095-130061095 on Build GRCh38
rs762942975 Chr.8: 130061025-130061025 on Build GRCh38
rs930902844 Chr.8: 130061126-130061126 on Build GRCh38
rs750559289 Chr.8: 130061074-130061074 on Build GRCh38
rs1033588571 Chr.8: 130061271-130061271 on Build GRCh38
rs748604169 Chr.8: 130061249-130061249 on Build GRCh38
rs750552166 Chr.8: 130060961-130060961 on Build GRCh38
rs1014004569 Chr.8: 130061275-130061275 on Build GRCh38
rs1022136622 Chr.8: 130061237-130061237 on Build GRCh38
rs373062629 Chr.8: 130061043-130061043 on Build GRCh38
rs926280708 Chr.8: 130060931-130060931 on Build GRCh38
rs745406996 Chr.8: 130061021-130061021 on Build GRCh38
rs200682369 Chr.8: 130060909-130060909 on Build GRCh38
rs890457050 Chr.8: 130061065-130061065 on Build GRCh38
rs553616348 Chr.8: 130060979-130060979 on Build GRCh38
rs765251399 Chr.8: 130060917-130060917 on Build GRCh38
rs374383935 Chr.8: 130061227-130061227 on Build GRCh38
rs775241259 Chr.8: 130060921-130060921 on Build GRCh38
rs751349147 Chr.8: 130060925-130060925 on Build GRCh38
rs753956266 Chr.8: 130061094-130061094 on Build GRCh38
rs961166261 Chr.8: 130061333-130061333 on Build GRCh38
rs187787472 Chr.8: 130061229-130061229 on Build GRCh38
rs757530775 Chr.8: 130060932-130060932 on Build GRCh38
rs779481105 Chr.8: 130061017-130061017 on Build GRCh38
rs1000368670 Chr.8: 130061223-130061223 on Build GRCh38
rs779532329 Chr.8: 130061107-130061107 on Build GRCh38
rs377622334 Chr.8: 130061020-130061020 on Build GRCh38
rs980144574 Chr.8: 130061330-130061330 on Build GRCh38
rs755540573 Chr.8: 130060999-130060999 on Build GRCh38
rs967835200 Chr.8: 130061265-130061265 on Build GRCh38
rs140559309 Chr.8: 130061022-130061022 on Build GRCh38
rs578121499 Chr.8: 130060989-130060989 on Build GRCh38
rs752747266 Chr.8: 130061345-130061345 on Build GRCh38
rs767610218 Chr.8: 130060954-130060954 on Build GRCh38
rs183537140 Chr.8: 130061169-130061169 on Build GRCh38
rs960389219 Chr.8: 130061311-130061311 on Build GRCh38
rs749695045 Chr.8: 130060992-130060992 on Build GRCh38
rs920765188 Chr.8: 130061340-130061340 on Build GRCh38
rs767808130 Chr.8: 130061067-130061067 on Build GRCh38
rs776255255 Chr.8: 130060907-130060907 on Build GRCh38
rs920734153 Chr.8: 130061106-130061106 on Build GRCh38
rs547148982 Chr.8: 130061360-130061360 on Build GRCh38
rs147223205 Chr.8: 130061050-130061050 on Build GRCh38
rs904753736 Chr.8: 130061156-130061156 on Build GRCh38
rs1002160627 Chr.8: 130061268-130061268 on Build GRCh38
rs974193176 Chr.8: 130061339-130061339 on Build GRCh38
rs369455130 Chr.8: 130061023-130061023 on Build GRCh38
rs774177834 Chr.8: 130061048-130061048 on Build GRCh38
rs111808729 Chr.8: 130061079-130061079 on Build GRCh38
rs1050667817 Chr.8: 130061350-130061350 on Build GRCh38
rs1024416174 Chr.8: 130061322-130061322 on Build GRCh38
rs747729624 Chr.8: 130061098-130061098 on Build GRCh38
rs889151861 Chr.8: 130061158-130061158 on Build GRCh38
rs529153542 Chr.8: 130061353-130061353 on Build GRCh38
rs910787545 Chr.8: 130061359-130061359 on Build GRCh38
rs748502745 Chr.8: 130061018-130061018 on Build GRCh38
rs779430448 Chr.8: 130061127-130061127 on Build GRCh38
rs1008889761 Chr.8: 130061170-130061170 on Build GRCh38
rs766437902 Chr.8: 130061083-130061083 on Build GRCh38
rs991819847 Chr.8: 130061326-130061326 on Build GRCh38
rs969736508 Chr.8: 130061326-130061326 on Build GRCh38
rs773970471 Chr.8: 130061093-130061093 on Build GRCh38

View Full Product Details