Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138138906-138139407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135B
Entrez Gene ID
51059
Gene Name
family with sequence similarity 135 member B
Gene Aliases
C8ORFK32
UniGene
Hs.126024
Gene Chromosome Location
Chr.8: 138130023-138498390 on Build GRCh38
Transcripts
NM_015912.3 XM_011517056.2 XM_011517058.2 XM_011517059.1 XM_011517060.2 XM_011517061.2 XM_011517062.1 XM_011517063.2 XM_011517064.2 XM_011517065.1 XM_011517068.2 XM_011517069.2 XM_011517072.2 XM_011517073.2 XM_011517074.1 XM_017013470.1 XM_017013471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3645413 c.3859G>A p.D1287N Chr.8: 138139028-138139028 on Build GRCh38
7242571 c.3829C>G p.L1277V Chr.8: 138139058-138139058 on Build GRCh38
244362 c.3842C>A p.T1281N Chr.8: 138139045-138139045 on Build GRCh38
3645418 c.3821C>T p.S1274F Chr.8: 138139066-138139066 on Build GRCh38
244363 c.3842C>A p.T1281N Chr.8: 138139045-138139045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778515491 Chr.8: 138139031-138139031 on Build GRCh38
rs796480424 Chr.8: 138139237-138139237 on Build GRCh38
rs554180607 Chr.8: 138139186-138139186 on Build GRCh38
rs754660861 Chr.8: 138139233-138139233 on Build GRCh38
rs998088715 Chr.8: 138139121-138139121 on Build GRCh38
rs527332547 Chr.8: 138138942-138138942 on Build GRCh38
rs771321489 Chr.8: 138139298-138139298 on Build GRCh38
rs936648188 Chr.8: 138139312-138139312 on Build GRCh38
rs933511700 Chr.8: 138139369-138139369 on Build GRCh38
rs201183176 Chr.8: 138139002-138139002 on Build GRCh38
rs772473976 Chr.8: 138138935-138138935 on Build GRCh38
rs145788864 Chr.8: 138139056-138139056 on Build GRCh38
rs772889502 Chr.8: 138139123-138139123 on Build GRCh38
rs189582889 Chr.8: 138139270-138139270 on Build GRCh38
rs570743642 Chr.8: 138139230-138139230 on Build GRCh38
rs758892783 Chr.8: 138139007-138139007 on Build GRCh38
rs778261848 Chr.8: 138139106-138139107 on Build GRCh38
rs377712830 Chr.8: 138138949-138138949 on Build GRCh38
rs770038980 Chr.8: 138139089-138139089 on Build GRCh38
rs779550865 Chr.8: 138139005-138139005 on Build GRCh38
rs1052811522 Chr.8: 138139376-138139376 on Build GRCh38
rs397819719 Chr.8: 138139187-138139188 on Build GRCh38
rs34567983 Chr.8: 138139035-138139036 on Build GRCh38
rs535284124 Chr.8: 138139117-138139117 on Build GRCh38
rs752047818 Chr.8: 138139143-138139145 on Build GRCh38
rs888929960 Chr.8: 138139115-138139115 on Build GRCh38
rs4434667 Chr.8: 138139276-138139276 on Build GRCh38
rs371955242 Chr.8: 138139223-138139223 on Build GRCh38
rs1045050914 Chr.8: 138139008-138139008 on Build GRCh38
rs116855100 Chr.8: 138139053-138139053 on Build GRCh38
rs377399248 Chr.8: 138139026-138139026 on Build GRCh38
rs542442477 Chr.8: 138139280-138139280 on Build GRCh38
rs374282935 Chr.8: 138138943-138138943 on Build GRCh38
rs747836721 Chr.8: 138139044-138139044 on Build GRCh38
rs767454102 Chr.8: 138138977-138138977 on Build GRCh38
rs555991951 Chr.8: 138139374-138139374 on Build GRCh38
rs943194345 Chr.8: 138139325-138139325 on Build GRCh38
rs910390551 Chr.8: 138139318-138139318 on Build GRCh38
rs919126991 Chr.8: 138138948-138138948 on Build GRCh38
rs374711628 Chr.8: 138139064-138139064 on Build GRCh38
rs970240194 Chr.8: 138139239-138139239 on Build GRCh38
rs934391868 Chr.8: 138139358-138139358 on Build GRCh38
rs572539584 Chr.8: 138139373-138139373 on Build GRCh38
rs893427116 Chr.8: 138139197-138139197 on Build GRCh38
rs763294346 Chr.8: 138139108-138139108 on Build GRCh38
rs776583585 Chr.8: 138138946-138138946 on Build GRCh38
rs114482324 Chr.8: 138139142-138139142 on Build GRCh38
rs4634694 Chr.8: 138139132-138139132 on Build GRCh38
rs1035736879 Chr.8: 138139269-138139269 on Build GRCh38
rs746389235 Chr.8: 138139357-138139357 on Build GRCh38
rs1011834889 Chr.8: 138139218-138139218 on Build GRCh38
rs750328290 Chr.8: 138138991-138138991 on Build GRCh38
rs772393893 Chr.8: 138139323-138139323 on Build GRCh38
rs141432978 Chr.8: 138139275-138139275 on Build GRCh38
rs35147483 Chr.8: 138139185-138139186 on Build GRCh38
rs759732424 Chr.8: 138138953-138138953 on Build GRCh38
rs781343438 Chr.8: 138139327-138139327 on Build GRCh38
rs372371106 Chr.8: 138139077-138139077 on Build GRCh38
rs146649599 Chr.8: 138139116-138139116 on Build GRCh38
rs760584927 Chr.8: 138139131-138139131 on Build GRCh38
rs371299779 Chr.8: 138138970-138138970 on Build GRCh38
rs374737174 Chr.8: 138139025-138139025 on Build GRCh38
rs765546631 Chr.8: 138138959-138138959 on Build GRCh38
rs984659608 Chr.8: 138139314-138139314 on Build GRCh38
rs775816772 Chr.8: 138139107-138139107 on Build GRCh38
rs923012505 Chr.8: 138139353-138139353 on Build GRCh38
rs368204243 Chr.8: 138139065-138139065 on Build GRCh38
rs981949661 Chr.8: 138139268-138139268 on Build GRCh38
rs745917025 Chr.8: 138139085-138139085 on Build GRCh38
rs4634695 Chr.8: 138139178-138139178 on Build GRCh38

View Full Product Details