Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138588839-138589331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
UniGene
Hs.117169
Gene Chromosome Location
Chr.8: 138588235-138914087 on Build GRCh38
Transcripts
NM_152888.2 XM_011516883.2 XM_011516884.2 XM_011516885.2 XM_011516886.2 XM_011516887.2 XM_011516889.2 XM_017013150.1 XM_017013151.1 XM_017013152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715957 c.4860G>A p.P1620P Chr.8: 138589274-138589274 on Build GRCh38
3951340 c.4857G>T p.R1619R Chr.8: 138589277-138589277 on Build GRCh38
1700687 c.4853_4854CC>TT p.A1618V Chr.8: 138589280-138589281 on Build GRCh38
4365248 c.4876C>T p.P1626S Chr.8: 138589258-138589258 on Build GRCh38
6995956 c.4823C>T p.S1608F Chr.8: 138589311-138589311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772740860 Chr.8: 138588917-138588917 on Build GRCh38
rs747100169 Chr.8: 138589302-138589302 on Build GRCh38
rs760937152 Chr.8: 138589223-138589223 on Build GRCh38
rs190592589 Chr.8: 138589069-138589069 on Build GRCh38
rs917425356 Chr.8: 138588970-138588970 on Build GRCh38
rs755307992 Chr.8: 138589279-138589279 on Build GRCh38
rs755759145 Chr.8: 138589277-138589277 on Build GRCh38
rs777805657 Chr.8: 138589295-138589295 on Build GRCh38
rs1024343126 Chr.8: 138589268-138589268 on Build GRCh38
rs983408858 Chr.8: 138589169-138589169 on Build GRCh38
rs377250129 Chr.8: 138588956-138588956 on Build GRCh38
rs991495875 Chr.8: 138588965-138588965 on Build GRCh38
rs539492044 Chr.8: 138588905-138588905 on Build GRCh38
rs575147648 Chr.8: 138589028-138589028 on Build GRCh38
rs73442789 Chr.8: 138588907-138588907 on Build GRCh38
rs570355598 Chr.8: 138588899-138588899 on Build GRCh38
rs534229625 Chr.8: 138589068-138589068 on Build GRCh38
rs1027309679 Chr.8: 138589142-138589142 on Build GRCh38
rs1024604292 Chr.8: 138589171-138589171 on Build GRCh38
rs377129806 Chr.8: 138589307-138589307 on Build GRCh38
rs983479980 Chr.8: 138589064-138589064 on Build GRCh38
rs775326883 Chr.8: 138589239-138589239 on Build GRCh38
rs150588004 Chr.8: 138588906-138588906 on Build GRCh38
rs963012276 Chr.8: 138589300-138589300 on Build GRCh38
rs372087339 Chr.8: 138589126-138589126 on Build GRCh38
rs958666895 Chr.8: 138589257-138589257 on Build GRCh38
rs761344086 Chr.8: 138589261-138589261 on Build GRCh38
rs148594998 Chr.8: 138589275-138589275 on Build GRCh38
rs748648857 Chr.8: 138589293-138589293 on Build GRCh38
rs868014610 Chr.8: 138589276-138589276 on Build GRCh38
rs774444364 Chr.8: 138589259-138589259 on Build GRCh38
rs894077433 Chr.8: 138589065-138589065 on Build GRCh38
rs371827946 Chr.8: 138589152-138589153 on Build GRCh38
rs368407433 Chr.8: 138589075-138589075 on Build GRCh38
rs549521277 Chr.8: 138588946-138588946 on Build GRCh38
rs971211984 Chr.8: 138589195-138589195 on Build GRCh38
rs1017398169 Chr.8: 138589128-138589128 on Build GRCh38
rs556264650 Chr.8: 138589304-138589304 on Build GRCh38
rs772139906 Chr.8: 138589217-138589217 on Build GRCh38
rs932391665 Chr.8: 138589089-138589089 on Build GRCh38
rs774694947 Chr.8: 138589276-138589283 on Build GRCh38
rs773344239 Chr.8: 138589222-138589222 on Build GRCh38
rs1033537032 Chr.8: 138588941-138588941 on Build GRCh38
rs567715461 Chr.8: 138589130-138589130 on Build GRCh38
rs758961090 Chr.8: 138589294-138589294 on Build GRCh38
rs755750005 Chr.8: 138588876-138588876 on Build GRCh38
rs1021312609 Chr.8: 138589146-138589146 on Build GRCh38
rs950214663 Chr.8: 138589054-138589054 on Build GRCh38
rs994529952 Chr.8: 138589134-138589137 on Build GRCh38
rs903754180 Chr.8: 138589152-138589152 on Build GRCh38
rs779325657 Chr.8: 138589285-138589285 on Build GRCh38
rs779527742 Chr.8: 138588884-138588884 on Build GRCh38
rs938048602 Chr.8: 138589036-138589036 on Build GRCh38
rs1050797736 Chr.8: 138589090-138589090 on Build GRCh38
rs764074505 Chr.8: 138589254-138589254 on Build GRCh38
rs1043008843 Chr.8: 138589074-138589074 on Build GRCh38
rs749054891 Chr.8: 138589308-138589308 on Build GRCh38
rs747349533 Chr.8: 138589226-138589226 on Build GRCh38
rs372942802 Chr.8: 138589274-138589274 on Build GRCh38
rs534827614 Chr.8: 138589147-138589147 on Build GRCh38
rs182324250 Chr.8: 138589154-138589154 on Build GRCh38
rs762688241 Chr.8: 138589252-138589252 on Build GRCh38
rs765975398 Chr.8: 138589278-138589278 on Build GRCh38
rs1009399507 Chr.8: 138589131-138589131 on Build GRCh38
rs547327197 Chr.8: 138588869-138588869 on Build GRCh38
rs899476901 Chr.8: 138589083-138589083 on Build GRCh38
rs538488990 Chr.8: 138588955-138588955 on Build GRCh38
rs555293846 Chr.8: 138588980-138588980 on Build GRCh38
rs926676544 Chr.8: 138588913-138588913 on Build GRCh38
rs967821559 Chr.8: 138588908-138588908 on Build GRCh38

View Full Product Details