Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138589221-138589727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
UniGene
Hs.117169
Gene Chromosome Location
Chr.8: 138588235-138914087 on Build GRCh38
Transcripts
NM_152888.2 XM_011516883.2 XM_011516884.2 XM_011516885.2 XM_011516886.2 XM_011516887.2 XM_011516889.2 XM_017013150.1 XM_017013151.1 XM_017013152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6632834 c.4745G>T p.G1582V Chr.8: 138589389-138589389 on Build GRCh38
3715957 c.4860G>A p.P1620P Chr.8: 138589274-138589274 on Build GRCh38
6472398 c.4765C>A p.P1589T Chr.8: 138589369-138589369 on Build GRCh38
126630 c.4750C>T p.Q1584* Chr.8: 138589384-138589384 on Build GRCh38
4365248 c.4876C>T p.P1626S Chr.8: 138589258-138589258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925176206 Chr.8: 138589601-138589601 on Build GRCh38
rs920938360 Chr.8: 138589333-138589333 on Build GRCh38
rs778895890 Chr.8: 138589637-138589639 on Build GRCh38
rs751308685 Chr.8: 138589560-138589560 on Build GRCh38
rs747100169 Chr.8: 138589302-138589302 on Build GRCh38
rs564075290 Chr.8: 138589433-138589433 on Build GRCh38
rs1054960273 Chr.8: 138589406-138589406 on Build GRCh38
rs1012854092 Chr.8: 138589422-138589422 on Build GRCh38
rs971519739 Chr.8: 138589463-138589463 on Build GRCh38
rs776786087 Chr.8: 138589337-138589337 on Build GRCh38
rs780444476 Chr.8: 138589438-138589438 on Build GRCh38
rs755307992 Chr.8: 138589279-138589279 on Build GRCh38
rs368533920 Chr.8: 138589432-138589432 on Build GRCh38
rs755759145 Chr.8: 138589277-138589277 on Build GRCh38
rs759165504 Chr.8: 138589399-138589399 on Build GRCh38
rs746077831 Chr.8: 138589368-138589368 on Build GRCh38
rs777805657 Chr.8: 138589295-138589295 on Build GRCh38
rs765571677 Chr.8: 138589343-138589343 on Build GRCh38
rs1024343126 Chr.8: 138589268-138589268 on Build GRCh38
rs569808796 Chr.8: 138589513-138589513 on Build GRCh38
rs900283495 Chr.8: 138589344-138589344 on Build GRCh38
rs532823660 Chr.8: 138589494-138589494 on Build GRCh38
rs771994233 Chr.8: 138589469-138589469 on Build GRCh38
rs570799918 Chr.8: 138589621-138589621 on Build GRCh38
rs895056282 Chr.8: 138589647-138589647 on Build GRCh38
rs760161445 Chr.8: 138589388-138589388 on Build GRCh38
rs764604329 Chr.8: 138589421-138589421 on Build GRCh38
rs763332115 Chr.8: 138589420-138589420 on Build GRCh38
rs60639203 Chr.8: 138589545-138589545 on Build GRCh38
rs377129806 Chr.8: 138589307-138589307 on Build GRCh38
rs139658465 Chr.8: 138589369-138589369 on Build GRCh38
rs75622841 Chr.8: 138589634-138589634 on Build GRCh38
rs146712669 Chr.8: 138589426-138589426 on Build GRCh38
rs963012276 Chr.8: 138589300-138589300 on Build GRCh38
rs781121978 Chr.8: 138589429-138589429 on Build GRCh38
rs779673840 Chr.8: 138589657-138589657 on Build GRCh38
rs761344086 Chr.8: 138589261-138589261 on Build GRCh38
rs958666895 Chr.8: 138589257-138589257 on Build GRCh38
rs541660200 Chr.8: 138589348-138589348 on Build GRCh38
rs556228006 Chr.8: 138589674-138589674 on Build GRCh38
rs549844778 Chr.8: 138589499-138589499 on Build GRCh38
rs148594998 Chr.8: 138589275-138589275 on Build GRCh38
rs772082442 Chr.8: 138589371-138589371 on Build GRCh38
rs748648857 Chr.8: 138589293-138589293 on Build GRCh38
rs774444364 Chr.8: 138589259-138589259 on Build GRCh38
rs868014610 Chr.8: 138589276-138589276 on Build GRCh38
rs974384070 Chr.8: 138589524-138589524 on Build GRCh38
rs974691796 Chr.8: 138589332-138589332 on Build GRCh38
rs144391800 Chr.8: 138589564-138589564 on Build GRCh38
rs556825337 Chr.8: 138589586-138589586 on Build GRCh38
rs1004308309 Chr.8: 138589690-138589690 on Build GRCh38
rs116332442 Chr.8: 138589572-138589572 on Build GRCh38
rs754691503 Chr.8: 138589338-138589338 on Build GRCh38
rs556264650 Chr.8: 138589304-138589304 on Build GRCh38
rs774694947 Chr.8: 138589276-138589283 on Build GRCh38
rs575827275 Chr.8: 138589317-138589317 on Build GRCh38
rs747890983 Chr.8: 138589450-138589450 on Build GRCh38
rs758961090 Chr.8: 138589294-138589294 on Build GRCh38
rs758869409 Chr.8: 138589347-138589347 on Build GRCh38
rs894522958 Chr.8: 138589416-138589416 on Build GRCh38
rs886181463 Chr.8: 138589632-138589632 on Build GRCh38
rs763940185 Chr.8: 138589555-138589555 on Build GRCh38
rs779325657 Chr.8: 138589285-138589285 on Build GRCh38
rs375490054 Chr.8: 138589353-138589353 on Build GRCh38
rs761915984 Chr.8: 138589412-138589412 on Build GRCh38
rs548753881 Chr.8: 138589546-138589546 on Build GRCh38
rs1004683079 Chr.8: 138589472-138589472 on Build GRCh38
rs767961999 Chr.8: 138589429-138589430 on Build GRCh38
rs1013428688 Chr.8: 138589649-138589649 on Build GRCh38
rs761832409 Chr.8: 138589315-138589315 on Build GRCh38
rs764074505 Chr.8: 138589254-138589254 on Build GRCh38
rs142500152 Chr.8: 138589346-138589346 on Build GRCh38
rs749054891 Chr.8: 138589308-138589308 on Build GRCh38
rs561552719 Chr.8: 138589351-138589351 on Build GRCh38
rs760121851 Chr.8: 138589336-138589336 on Build GRCh38
rs372942802 Chr.8: 138589274-138589274 on Build GRCh38
rs957176796 Chr.8: 138589500-138589500 on Build GRCh38
rs773033974 Chr.8: 138589378-138589378 on Build GRCh38
rs762688241 Chr.8: 138589252-138589252 on Build GRCh38
rs989582265 Chr.8: 138589507-138589507 on Build GRCh38
rs772735366 Chr.8: 138589321-138589321 on Build GRCh38
rs765975398 Chr.8: 138589278-138589278 on Build GRCh38
rs903659642 Chr.8: 138589391-138589391 on Build GRCh38
rs764907766 Chr.8: 138589409-138589409 on Build GRCh38
rs150372930 Chr.8: 138589402-138589402 on Build GRCh38
rs753021910 Chr.8: 138589410-138589410 on Build GRCh38
rs145946202 Chr.8: 138589349-138589349 on Build GRCh38
rs143281223 Chr.8: 138589392-138589392 on Build GRCh38
rs781409790 Chr.8: 138589360-138589360 on Build GRCh38
rs370843003 Chr.8: 138589314-138589314 on Build GRCh38
rs750135911 Chr.8: 138589619-138589619 on Build GRCh38

View Full Product Details