Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 141144066-141144550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND3
Entrez Gene ID
22898
Gene Name
DENN domain containing 3
Gene Aliases
-
UniGene
Hs.18166
Gene Chromosome Location
Chr.8: 141128618-141195804 on Build GRCh38
Transcripts
NM_014957.2 XM_005250838.4 XM_005250839.2 XM_005250840.4 XM_005250841.3 XM_005250842.4 XM_011516933.1 XM_011516934.2 XM_011516937.1 XM_011516941.2 XM_017013241.1 XM_017013242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893294 c.384-7A>T p.? Chr.8: 141144141-141144141 on Build GRCh38
6642354 c.395T>C p.L132P Chr.8: 141144159-141144159 on Build GRCh38
7246004 c.463A>G p.I155V Chr.8: 141144227-141144227 on Build GRCh38
5885412 c.486G>A p.P162P Chr.8: 141144250-141144250 on Build GRCh38
3897214 c.439G>T p.D147Y Chr.8: 141144203-141144203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777532651 Chr.8: 141144186-141144186 on Build GRCh38
rs754378452 Chr.8: 141144120-141144120 on Build GRCh38
rs551237031 Chr.8: 141144495-141144495 on Build GRCh38
rs376229948 Chr.8: 141144119-141144119 on Build GRCh38
rs751490379 Chr.8: 141144181-141144182 on Build GRCh38
rs972342487 Chr.8: 141144118-141144118 on Build GRCh38
rs756185955 Chr.8: 141144256-141144256 on Build GRCh38
rs760885420 Chr.8: 141144209-141144209 on Build GRCh38
rs879250767 Chr.8: 141144161-141144161 on Build GRCh38
rs889103951 Chr.8: 141144315-141144315 on Build GRCh38
rs957382858 Chr.8: 141144234-141144234 on Build GRCh38
rs1014429597 Chr.8: 141144437-141144437 on Build GRCh38
rs993669733 Chr.8: 141144395-141144395 on Build GRCh38
rs867167223 Chr.8: 141144473-141144473 on Build GRCh38
rs755594245 Chr.8: 141144227-141144227 on Build GRCh38
rs988590301 Chr.8: 141144493-141144493 on Build GRCh38
rs769635872 Chr.8: 141144180-141144180 on Build GRCh38
rs750103828 Chr.8: 141144208-141144208 on Build GRCh38
rs572514677 Chr.8: 141144197-141144197 on Build GRCh38
rs778071426 Chr.8: 141144249-141144249 on Build GRCh38
rs200601782 Chr.8: 141144213-141144213 on Build GRCh38
rs965886120 Chr.8: 141144506-141144506 on Build GRCh38
rs997309371 Chr.8: 141144125-141144125 on Build GRCh38
rs533757159 Chr.8: 141144368-141144368 on Build GRCh38
rs370068043 Chr.8: 141144158-141144158 on Build GRCh38
rs116789554 Chr.8: 141144160-141144160 on Build GRCh38
rs1009013685 Chr.8: 141144347-141144347 on Build GRCh38
rs1050236339 Chr.8: 141144384-141144384 on Build GRCh38
rs934376383 Chr.8: 141144489-141144489 on Build GRCh38
rs780139045 Chr.8: 141144306-141144309 on Build GRCh38
rs756830931 Chr.8: 141144382-141144382 on Build GRCh38
rs888529898 Chr.8: 141144390-141144390 on Build GRCh38
rs112710966 Chr.8: 141144464-141144467 on Build GRCh38
rs878975915 Chr.8: 141144284-141144285 on Build GRCh38
rs957127699 Chr.8: 141144483-141144483 on Build GRCh38
rs267601799 Chr.8: 141144214-141144214 on Build GRCh38
rs769460963 Chr.8: 141144262-141144262 on Build GRCh38
rs745587771 Chr.8: 141144168-141144168 on Build GRCh38
rs767108350 Chr.8: 141144203-141144203 on Build GRCh38
rs1032772801 Chr.8: 141144233-141144233 on Build GRCh38
rs193097558 Chr.8: 141144461-141144461 on Build GRCh38
rs922569899 Chr.8: 141144379-141144379 on Build GRCh38
rs199601114 Chr.8: 141144196-141144196 on Build GRCh38
rs1014422872 Chr.8: 141144386-141144386 on Build GRCh38
rs914387105 Chr.8: 141144521-141144521 on Build GRCh38
rs763004692 Chr.8: 141144190-141144190 on Build GRCh38
rs897356671 Chr.8: 141144433-141144433 on Build GRCh38
rs888572996 Chr.8: 141144157-141144157 on Build GRCh38
rs779591535 Chr.8: 141144232-141144232 on Build GRCh38
rs971076362 Chr.8: 141144449-141144449 on Build GRCh38
rs893048834 Chr.8: 141144276-141144276 on Build GRCh38
rs307761 Chr.8: 141144192-141144192 on Build GRCh38
rs307762 Chr.8: 141144163-141144163 on Build GRCh38
rs1027789350 Chr.8: 141144416-141144416 on Build GRCh38
rs565363645 Chr.8: 141144421-141144421 on Build GRCh38
rs544062286 Chr.8: 141144294-141144294 on Build GRCh38
rs975956209 Chr.8: 141144522-141144522 on Build GRCh38
rs745822061 Chr.8: 141144482-141144482 on Build GRCh38
rs771579693 Chr.8: 141144302-141144302 on Build GRCh38
rs1020102457 Chr.8: 141144494-141144494 on Build GRCh38
rs976011765 Chr.8: 141144356-141144356 on Build GRCh38
rs932669108 Chr.8: 141144380-141144380 on Build GRCh38
rs532067603 Chr.8: 141144285-141144285 on Build GRCh38
rs779195618 Chr.8: 141144151-141144151 on Build GRCh38
rs989953915 Chr.8: 141144511-141144511 on Build GRCh38
rs111678968 Chr.8: 141144250-141144250 on Build GRCh38
rs991733624 Chr.8: 141144422-141144422 on Build GRCh38
rs941480615 Chr.8: 141144393-141144393 on Build GRCh38
rs749151175 Chr.8: 141144271-141144271 on Build GRCh38
rs750747442 Chr.8: 141144101-141144101 on Build GRCh38
rs779946069 Chr.8: 141144264-141144264 on Build GRCh38
rs944594237 Chr.8: 141144333-141144333 on Build GRCh38
rs768542057 Chr.8: 141144281-141144281 on Build GRCh38
rs551914822 Chr.8: 141144478-141144478 on Build GRCh38
rs898035291 Chr.8: 141144386-141144387 on Build GRCh38
rs369050051 Chr.8: 141144127-141144127 on Build GRCh38
rs755011734 Chr.8: 141144138-141144138 on Build GRCh38
rs756524635 Chr.8: 141144103-141144103 on Build GRCh38
rs377145402 Chr.8: 141144237-141144237 on Build GRCh38
rs552171043 Chr.8: 141144238-141144238 on Build GRCh38

View Full Product Details