Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142840324-142840832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GML
Entrez Gene ID
2765
Gene Name
glycosylphosphatidylinositol anchored molecule like
Gene Aliases
LY6DL
UniGene
Hs.661218
Gene Chromosome Location
Chr.8: 142834801-142846846 on Build GRCh38
Transcripts
NM_002066.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687330 c.67G>A p.A23T Chr.8: 142840504-142840504 on Build GRCh38
6112717 c.66C>A p.R22R Chr.8: 142840503-142840503 on Build GRCh38
1096866 c.66C>T p.R22R Chr.8: 142840503-142840503 on Build GRCh38
1455277 c.65G>A p.R22H Chr.8: 142840502-142840502 on Build GRCh38
3645945 c.34C>T p.L12F Chr.8: 142840471-142840471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888756189 Chr.8: 142840357-142840357 on Build GRCh38
rs150336506 Chr.8: 142840458-142840458 on Build GRCh38
rs1006361291 Chr.8: 142840566-142840566 on Build GRCh38
rs1028882975 Chr.8: 142840528-142840528 on Build GRCh38
rs945784584 Chr.8: 142840754-142840754 on Build GRCh38
rs569921732 Chr.8: 142840799-142840799 on Build GRCh38
rs758161442 Chr.8: 142840398-142840399 on Build GRCh38
rs1045344543 Chr.8: 142840464-142840464 on Build GRCh38
rs186330766 Chr.8: 142840563-142840563 on Build GRCh38
rs188114864 Chr.8: 142840469-142840469 on Build GRCh38
rs756917702 Chr.8: 142840519-142840519 on Build GRCh38
rs61729415 Chr.8: 142840461-142840461 on Build GRCh38
rs180916516 Chr.8: 142840545-142840545 on Build GRCh38
rs557184535 Chr.8: 142840443-142840443 on Build GRCh38
rs553529825 Chr.8: 142840382-142840382 on Build GRCh38
rs547208157 Chr.8: 142840633-142840633 on Build GRCh38
rs371868763 Chr.8: 142840805-142840805 on Build GRCh38
rs763904256 Chr.8: 142840427-142840438 on Build GRCh38
rs10104860 Chr.8: 142840761-142840761 on Build GRCh38
rs988898493 Chr.8: 142840673-142840673 on Build GRCh38
rs549351182 Chr.8: 142840570-142840570 on Build GRCh38
rs967835025 Chr.8: 142840717-142840717 on Build GRCh38
rs956258311 Chr.8: 142840665-142840665 on Build GRCh38
rs891440138 Chr.8: 142840359-142840359 on Build GRCh38
rs780572421 Chr.8: 142840470-142840470 on Build GRCh38
rs879653842 Chr.8: 142840715-142840715 on Build GRCh38
rs756903332 Chr.8: 142840463-142840463 on Build GRCh38
rs199525495 Chr.8: 142840501-142840501 on Build GRCh38
rs34441899 Chr.8: 142840466-142840466 on Build GRCh38
rs372354201 Chr.8: 142840547-142840547 on Build GRCh38
rs138131984 Chr.8: 142840503-142840503 on Build GRCh38
rs925556266 Chr.8: 142840706-142840706 on Build GRCh38
rs951882484 Chr.8: 142840426-142840426 on Build GRCh38
rs3750246 Chr.8: 142840388-142840388 on Build GRCh38
rs549369890 Chr.8: 142840525-142840525 on Build GRCh38
rs901484242 Chr.8: 142840786-142840786 on Build GRCh38
rs767986242 Chr.8: 142840598-142840598 on Build GRCh38
rs922951753 Chr.8: 142840785-142840785 on Build GRCh38
rs992195182 Chr.8: 142840450-142840450 on Build GRCh38
rs752080921 Chr.8: 142840398-142840398 on Build GRCh38
rs3750247 Chr.8: 142840396-142840396 on Build GRCh38
rs752399035 Chr.8: 142840612-142840612 on Build GRCh38
rs538481490 Chr.8: 142840375-142840375 on Build GRCh38
rs993689617 Chr.8: 142840397-142840397 on Build GRCh38
rs768828613 Chr.8: 142840477-142840477 on Build GRCh38
rs2717570 Chr.8: 142840746-142840746 on Build GRCh38
rs774540638 Chr.8: 142840482-142840482 on Build GRCh38
rs542934277 Chr.8: 142840719-142840719 on Build GRCh38
rs951851785 Chr.8: 142840541-142840541 on Build GRCh38
rs887921347 Chr.8: 142840560-142840560 on Build GRCh38
rs1006037559 Chr.8: 142840549-142840549 on Build GRCh38
rs749347372 Chr.8: 142840475-142840475 on Build GRCh38
rs759445396 Chr.8: 142840425-142840425 on Build GRCh38
rs1024935800 Chr.8: 142840617-142840617 on Build GRCh38
rs368117158 Chr.8: 142840423-142840423 on Build GRCh38
rs367753934 Chr.8: 142840389-142840389 on Build GRCh38
rs375179110 Chr.8: 142840408-142840408 on Build GRCh38
rs555744985 Chr.8: 142840422-142840422 on Build GRCh38
rs556758504 Chr.8: 142840623-142840623 on Build GRCh38
rs571037492 Chr.8: 142840610-142840610 on Build GRCh38
rs776578359 Chr.8: 142840498-142840498 on Build GRCh38
rs772406146 Chr.8: 142840495-142840495 on Build GRCh38
rs962191062 Chr.8: 142840595-142840595 on Build GRCh38
rs190477548 Chr.8: 142840749-142840749 on Build GRCh38
rs762104327 Chr.8: 142840416-142840416 on Build GRCh38
rs757878193 Chr.8: 142840452-142840452 on Build GRCh38
rs763664633 Chr.8: 142840505-142840505 on Build GRCh38
rs370445874 Chr.8: 142840465-142840465 on Build GRCh38
rs1009934862 Chr.8: 142840373-142840373 on Build GRCh38
rs200642476 Chr.8: 142840502-142840502 on Build GRCh38
rs988349686 Chr.8: 142840748-142840748 on Build GRCh38
rs992561053 Chr.8: 142840600-142840600 on Build GRCh38
rs760776634 Chr.8: 142840559-142840559 on Build GRCh38
rs762771971 Chr.8: 142840504-142840504 on Build GRCh38
rs1006413635 Chr.8: 142840423-142840425 on Build GRCh38
rs1022146607 Chr.8: 142840711-142840711 on Build GRCh38
rs373850755 Chr.8: 142840631-142840631 on Build GRCh38
rs751056563 Chr.8: 142840518-142840518 on Build GRCh38
rs962339417 Chr.8: 142840446-142840446 on Build GRCh38
rs372387223 Chr.8: 142840429-142840429 on Build GRCh38
rs1035691907 Chr.8: 142840624-142840624 on Build GRCh38
rs375561355 Chr.8: 142840430-142840430 on Build GRCh38
rs756451060 Chr.8: 142840546-142840546 on Build GRCh38

View Full Product Details