Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143581137-143581626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1D
Entrez Gene ID
1936
Gene Name
eukaryotic translation elongation factor 1 delta
Gene Aliases
EF-1D, EF1D, FP1047
UniGene
Hs.333388
Gene Chromosome Location
Chr.8: 143579697-143597675 on Build GRCh38
Transcripts
NM_001130053.2 NM_001130055.2 NM_001130056.2 NM_001130057.2 NM_001195203.1 NM_001289950.1 NM_001317743.1 NM_001960.4 NM_032378.4 XM_005250820.3 XM_005250823.1 XM_006716519.3 XM_006716520.3 XM_006716522.1 XM_006716523.1 XM_006716524.1 XM_006716525.1 XM_011516905.2 XM_011516906.2 XM_017013168.1 XM_017013169.1 XM_017013170.1 XM_017013171.1 XM_017013172.1 XM_017013173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3929416 c.1387G>A p.V463M Chr.8: 143581229-143581229 on Build GRCh38
2868415 c.1306T>C p.S436P Chr.8: 143581310-143581310 on Build GRCh38
6516726 c.1346G>T p.R449L Chr.8: 143581270-143581270 on Build GRCh38
3897448 c.1381C>T p.R461C Chr.8: 143581235-143581235 on Build GRCh38
3929417 c.1387G>A p.V463M Chr.8: 143581229-143581229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2305493 Chr.8: 143581296-143581296 on Build GRCh38
rs138340554 Chr.8: 143581463-143581464 on Build GRCh38
rs761785791 Chr.8: 143581263-143581263 on Build GRCh38
rs1048103884 Chr.8: 143581573-143581573 on Build GRCh38
rs1023575527 Chr.8: 143581604-143581604 on Build GRCh38
rs148681550 Chr.8: 143581501-143581502 on Build GRCh38
rs537569050 Chr.8: 143581601-143581601 on Build GRCh38
rs866529766 Chr.8: 143581431-143581431 on Build GRCh38
rs191973341 Chr.8: 143581191-143581191 on Build GRCh38
rs561028286 Chr.8: 143581399-143581399 on Build GRCh38
rs943918630 Chr.8: 143581542-143581542 on Build GRCh38
rs2123758 Chr.8: 143581491-143581491 on Build GRCh38
rs746295287 Chr.8: 143581375-143581375 on Build GRCh38
rs776384176 Chr.8: 143581276-143581276 on Build GRCh38
rs757481325 Chr.8: 143581232-143581232 on Build GRCh38
rs201818944 Chr.8: 143581158-143581158 on Build GRCh38
rs372287919 Chr.8: 143581553-143581553 on Build GRCh38
rs1037655072 Chr.8: 143581591-143581591 on Build GRCh38
rs188606460 Chr.8: 143581481-143581481 on Build GRCh38
rs567473547 Chr.8: 143581554-143581554 on Build GRCh38
rs141809356 Chr.8: 143581275-143581275 on Build GRCh38
rs966556801 Chr.8: 143581482-143581482 on Build GRCh38
rs772795677 Chr.8: 143581185-143581185 on Build GRCh38
rs766455968 Chr.8: 143581376-143581376 on Build GRCh38
rs369097028 Chr.8: 143581278-143581278 on Build GRCh38
rs11548167 Chr.8: 143581274-143581274 on Build GRCh38
rs759475757 Chr.8: 143581382-143581382 on Build GRCh38
rs754269237 Chr.8: 143581199-143581199 on Build GRCh38
rs367650218 Chr.8: 143581564-143581564 on Build GRCh38
rs778765331 Chr.8: 143581266-143581266 on Build GRCh38
rs546013263 Chr.8: 143581483-143581488 on Build GRCh38
rs200304612 Chr.8: 143581213-143581213 on Build GRCh38
rs534173851 Chr.8: 143581218-143581218 on Build GRCh38
rs534497314 Chr.8: 143581539-143581539 on Build GRCh38
rs1033432107 Chr.8: 143581486-143581486 on Build GRCh38
rs751523899 Chr.8: 143581230-143581230 on Build GRCh38
rs372677188 Chr.8: 143581338-143581338 on Build GRCh38
rs756331837 Chr.8: 143581375-143581375 on Build GRCh38
rs1014413253 Chr.8: 143581452-143581452 on Build GRCh38
rs776476238 Chr.8: 143581341-143581341 on Build GRCh38
rs770537200 Chr.8: 143581340-143581340 on Build GRCh38
rs767029633 Chr.8: 143581176-143581176 on Build GRCh38
rs776716763 Chr.8: 143581207-143581207 on Build GRCh38
rs761268103 Chr.8: 143581175-143581175 on Build GRCh38
rs754824124 Chr.8: 143581311-143581311 on Build GRCh38
rs368338544 Chr.8: 143581162-143581162 on Build GRCh38
rs551045944 Chr.8: 143581432-143581432 on Build GRCh38
rs370722668 Chr.8: 143581287-143581287 on Build GRCh38
rs781432843 Chr.8: 143581306-143581306 on Build GRCh38
rs888115104 Chr.8: 143581582-143581582 on Build GRCh38
rs750708671 Chr.8: 143581371-143581371 on Build GRCh38
rs750205402 Chr.8: 143581602-143581602 on Build GRCh38
rs772811624 Chr.8: 143581338-143581340 on Build GRCh38
rs201898291 Chr.8: 143581163-143581163 on Build GRCh38
rs772061508 Chr.8: 143581327-143581327 on Build GRCh38
rs764688998 Chr.8: 143581353-143581353 on Build GRCh38
rs11548164 Chr.8: 143581236-143581236 on Build GRCh38
rs555803649 Chr.8: 143581605-143581605 on Build GRCh38
rs768408455 Chr.8: 143581261-143581261 on Build GRCh38
rs921076633 Chr.8: 143581563-143581563 on Build GRCh38
rs1054478769 Chr.8: 143581498-143581498 on Build GRCh38
rs775933978 Chr.8: 143581347-143581347 on Build GRCh38
rs995190926 Chr.8: 143581390-143581390 on Build GRCh38
rs567099361 Chr.8: 143581520-143581520 on Build GRCh38
rs888096306 Chr.8: 143581401-143581401 on Build GRCh38
rs761929103 Chr.8: 143581368-143581368 on Build GRCh38
rs551746330 Chr.8: 143581169-143581169 on Build GRCh38
rs771739189 Chr.8: 143581265-143581265 on Build GRCh38
rs978846452 Chr.8: 143581485-143581485 on Build GRCh38
rs769736673 Chr.8: 143581313-143581313 on Build GRCh38
rs976612177 Chr.8: 143581545-143581545 on Build GRCh38
rs1004386508 Chr.8: 143581588-143581589 on Build GRCh38
rs749109504 Chr.8: 143581260-143581260 on Build GRCh38
rs564978810 Chr.8: 143581214-143581214 on Build GRCh38
rs568576129 Chr.8: 143581317-143581317 on Build GRCh38
rs1015761869 Chr.8: 143581447-143581447 on Build GRCh38
rs572304185 Chr.8: 143581377-143581377 on Build GRCh38
rs375083833 Chr.8: 143581160-143581160 on Build GRCh38
rs751678877 Chr.8: 143581304-143581304 on Build GRCh38
rs760372345 Chr.8: 143581188-143581188 on Build GRCh38
rs375904178 Chr.8: 143581352-143581352 on Build GRCh38
rs758460331 Chr.8: 143581219-143581219 on Build GRCh38
rs953104544 Chr.8: 143581195-143581195 on Build GRCh38
rs200810259 Chr.8: 143581159-143581159 on Build GRCh38
rs767570266 Chr.8: 143581369-143581369 on Build GRCh38
rs777523733 Chr.8: 143581336-143581336 on Build GRCh38
rs560229924 Chr.8: 143581314-143581314 on Build GRCh38
rs757283733 Chr.8: 143581305-143581305 on Build GRCh38
rs925233034 Chr.8: 143581490-143581490 on Build GRCh38
rs759251947 Chr.8: 143581345-143581345 on Build GRCh38
rs939720973 Chr.8: 143581584-143581584 on Build GRCh38
rs763175628 Chr.8: 143581350-143581350 on Build GRCh38
rs771547578 Chr.8: 143581271-143581271 on Build GRCh38
rs529426510 Chr.8: 143581404-143581404 on Build GRCh38
rs896061541 Chr.8: 143581451-143581451 on Build GRCh38
rs774685411 Chr.8: 143581360-143581360 on Build GRCh38
rs760021329 Chr.8: 143581277-143581277 on Build GRCh38
rs747638877 Chr.8: 143581156-143581157 on Build GRCh38
rs370493633 Chr.8: 143581443-143581443 on Build GRCh38
rs555518338 Chr.8: 143581234-143581234 on Build GRCh38
rs562878244 Chr.8: 143581464-143581464 on Build GRCh38
rs375110030 Chr.8: 143581343-143581343 on Build GRCh38
rs147641853 Chr.8: 143581540-143581540 on Build GRCh38
rs781339017 Chr.8: 143581244-143581244 on Build GRCh38
rs770126909 Chr.8: 143581346-143581346 on Build GRCh38
rs200657580 Chr.8: 143581310-143581310 on Build GRCh38
rs542488602 Chr.8: 143581392-143581392 on Build GRCh38
rs1036651189 Chr.8: 143581587-143581587 on Build GRCh38
rs145281112 Chr.8: 143581295-143581295 on Build GRCh38
rs148062906 Chr.8: 143581270-143581270 on Build GRCh38
rs777897534 Chr.8: 143581222-143581222 on Build GRCh38
rs916028680 Chr.8: 143581250-143581250 on Build GRCh38
rs936020361 Chr.8: 143581294-143581294 on Build GRCh38
rs1004423622 Chr.8: 143581411-143581411 on Build GRCh38

View Full Product Details