Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143866801-143867304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs375002026 Chr.8: 143867214-143867214 on Build GRCh38
rs201587092 Chr.8: 143867037-143867037 on Build GRCh38
rs895620182 Chr.8: 143867067-143867067 on Build GRCh38
rs35176503 Chr.8: 143867224-143867224 on Build GRCh38
rs781893307 Chr.8: 143867100-143867100 on Build GRCh38
rs782536099 Chr.8: 143866835-143866835 on Build GRCh38
rs528153180 Chr.8: 143867259-143867259 on Build GRCh38
rs374405420 Chr.8: 143866859-143866859 on Build GRCh38
rs977353069 Chr.8: 143867123-143867123 on Build GRCh38
rs1048567847 Chr.8: 143867056-143867056 on Build GRCh38
rs781847135 Chr.8: 143867133-143867133 on Build GRCh38
rs782643318 Chr.8: 143867026-143867026 on Build GRCh38
rs368863679 Chr.8: 143867131-143867131 on Build GRCh38
rs1015473623 Chr.8: 143866998-143866998 on Build GRCh38
rs782566173 Chr.8: 143866914-143866914 on Build GRCh38
rs200372671 Chr.8: 143866848-143866848 on Build GRCh38
rs781926085 Chr.8: 143866852-143866852 on Build GRCh38
rs545639287 Chr.8: 143867220-143867220 on Build GRCh38
rs782207878 Chr.8: 143867153-143867153 on Build GRCh38
rs200500117 Chr.8: 143867036-143867036 on Build GRCh38
rs572354463 Chr.8: 143866890-143866890 on Build GRCh38
rs782734320 Chr.8: 143867223-143867223 on Build GRCh38
rs782669606 Chr.8: 143866884-143866884 on Build GRCh38
rs781865685 Chr.8: 143867246-143867246 on Build GRCh38
rs572291032 Chr.8: 143867079-143867079 on Build GRCh38
rs782414797 Chr.8: 143866843-143866843 on Build GRCh38
rs1004039605 Chr.8: 143866989-143866989 on Build GRCh38
rs79860671 Chr.8: 143867030-143867030 on Build GRCh38
rs782042148 Chr.8: 143867170-143867170 on Build GRCh38
rs189896129 Chr.8: 143866965-143866965 on Build GRCh38
rs782809770 Chr.8: 143867184-143867184 on Build GRCh38
rs375707682 Chr.8: 143867129-143867129 on Build GRCh38
rs782125364 Chr.8: 143867134-143867134 on Build GRCh38
rs782147317 Chr.8: 143866951-143866951 on Build GRCh38
rs182185580 Chr.8: 143867113-143867113 on Build GRCh38
rs551942992 Chr.8: 143866975-143866975 on Build GRCh38
rs200700093 Chr.8: 143867070-143867070 on Build GRCh38
rs368909298 Chr.8: 143867014-143867014 on Build GRCh38
rs781868121 Chr.8: 143867105-143867105 on Build GRCh38
rs782626446 Chr.8: 143866920-143866920 on Build GRCh38
rs782405559 Chr.8: 143867212-143867212 on Build GRCh38
rs781885118 Chr.8: 143867149-143867149 on Build GRCh38
rs782050033 Chr.8: 143867222-143867222 on Build GRCh38
rs782452886 Chr.8: 143867096-143867096 on Build GRCh38
rs782124931 Chr.8: 143866861-143866861 on Build GRCh38
rs782821082 Chr.8: 143866832-143866832 on Build GRCh38
rs782446058 Chr.8: 143867254-143867254 on Build GRCh38
rs567287803 Chr.8: 143867027-143867027 on Build GRCh38
rs782149613 Chr.8: 143867279-143867279 on Build GRCh38
rs782395561 Chr.8: 143866995-143866995 on Build GRCh38
rs373165682 Chr.8: 143867260-143867260 on Build GRCh38
rs782301705 Chr.8: 143867025-143867025 on Build GRCh38
rs892750826 Chr.8: 143866836-143866836 on Build GRCh38
rs782691764 Chr.8: 143867261-143867261 on Build GRCh38
rs782110508 Chr.8: 143867053-143867053 on Build GRCh38
rs554910033 Chr.8: 143867083-143867083 on Build GRCh38
rs377207781 Chr.8: 143866851-143866851 on Build GRCh38
rs782099419 Chr.8: 143866864-143866864 on Build GRCh38
rs553724064 Chr.8: 143866876-143866876 on Build GRCh38
rs782373505 Chr.8: 143867263-143867263 on Build GRCh38
rs751723702 Chr.8: 143866842-143866842 on Build GRCh38
rs782768826 Chr.8: 143867244-143867244 on Build GRCh38
rs189688618 Chr.8: 143867157-143867157 on Build GRCh38
rs782318008 Chr.8: 143866924-143866924 on Build GRCh38
rs782182155 Chr.8: 143867115-143867115 on Build GRCh38
rs782322639 Chr.8: 143867117-143867117 on Build GRCh38
rs782374797 Chr.8: 143866935-143866935 on Build GRCh38
rs782653009 Chr.8: 143867204-143867204 on Build GRCh38
rs199828972 Chr.8: 143867280-143867280 on Build GRCh38
rs782200809 Chr.8: 143867208-143867208 on Build GRCh38
rs141900369 Chr.8: 143866913-143866913 on Build GRCh38
rs781793059 Chr.8: 143867193-143867193 on Build GRCh38
rs782107684 Chr.8: 143867172-143867172 on Build GRCh38
rs782662791 Chr.8: 143867152-143867152 on Build GRCh38
rs782042830 Chr.8: 143866858-143866858 on Build GRCh38
rs542970349 Chr.8: 143866893-143866893 on Build GRCh38
rs369404340 Chr.8: 143866822-143866822 on Build GRCh38
rs925397944 Chr.8: 143866931-143866931 on Build GRCh38
rs78008227 Chr.8: 143867191-143867191 on Build GRCh38
rs781845272 Chr.8: 143867090-143867090 on Build GRCh38
rs782481607 Chr.8: 143867187-143867187 on Build GRCh38
rs781948794 Chr.8: 143867055-143867055 on Build GRCh38
rs1020981879 Chr.8: 143867257-143867257 on Build GRCh38
rs376434440 Chr.8: 143866863-143866863 on Build GRCh38
rs782561343 Chr.8: 143867235-143867235 on Build GRCh38
rs782007848 Chr.8: 143866889-143866889 on Build GRCh38
rs755180904 Chr.8: 143866871-143866871 on Build GRCh38
rs782145665 Chr.8: 143867010-143867010 on Build GRCh38
rs782450682 Chr.8: 143866883-143866883 on Build GRCh38
rs567353491 Chr.8: 143866825-143866825 on Build GRCh38
rs782033531 Chr.8: 143867213-143867213 on Build GRCh38
rs782457896 Chr.8: 143867093-143867093 on Build GRCh38
rs782436797 Chr.8: 143867021-143867021 on Build GRCh38
rs540799146 Chr.8: 143867151-143867151 on Build GRCh38
rs553276549 Chr.8: 143866926-143866926 on Build GRCh38
rs373694823 Chr.8: 143866912-143866912 on Build GRCh38
rs782773843 Chr.8: 143866963-143866963 on Build GRCh38
rs782636871 Chr.8: 143866915-143866915 on Build GRCh38
rs782704141 Chr.8: 143866830-143866830 on Build GRCh38
rs781835809 Chr.8: 143867140-143867140 on Build GRCh38
rs536436352 Chr.8: 143866909-143866909 on Build GRCh38
rs782003100 Chr.8: 143866936-143866936 on Build GRCh38
rs7839934 Chr.8: 143867013-143867013 on Build GRCh38
rs781937384 Chr.8: 143866950-143866950 on Build GRCh38
rs1041275634 Chr.8: 143867044-143867044 on Build GRCh38
rs782097693 Chr.8: 143867057-143867057 on Build GRCh38
rs781861594 Chr.8: 143867209-143867210 on Build GRCh38
rs782379600 Chr.8: 143867174-143867174 on Build GRCh38
rs553910552 Chr.8: 143867078-143867078 on Build GRCh38
rs782639558 Chr.8: 143866844-143866844 on Build GRCh38
rs782812608 Chr.8: 143866870-143866870 on Build GRCh38
rs782525735 Chr.8: 143866875-143866875 on Build GRCh38
rs139977710 Chr.8: 143867202-143867202 on Build GRCh38
rs781871822 Chr.8: 143867065-143867065 on Build GRCh38
rs782179779 Chr.8: 143866882-143866882 on Build GRCh38
rs781812162 Chr.8: 143867004-143867004 on Build GRCh38
rs782230626 Chr.8: 143867272-143867272 on Build GRCh38
rs371109688 Chr.8: 143866866-143866866 on Build GRCh38
rs782150025 Chr.8: 143867183-143867183 on Build GRCh38
rs781856397 Chr.8: 143866957-143866957 on Build GRCh38
rs146515033 Chr.8: 143867144-143867144 on Build GRCh38
rs781797299 Chr.8: 143866889-143866890 on Build GRCh38
rs782434357 Chr.8: 143866841-143866841 on Build GRCh38
rs782281943 Chr.8: 143866930-143866930 on Build GRCh38
rs368041373 Chr.8: 143866952-143866952 on Build GRCh38
rs782357750 Chr.8: 143866948-143866948 on Build GRCh38
rs782469351 Chr.8: 143867073-143867073 on Build GRCh38
rs782512165 Chr.8: 143867015-143867015 on Build GRCh38
rs367640831 Chr.8: 143867252-143867252 on Build GRCh38
rs144123426 Chr.8: 143867061-143867061 on Build GRCh38
rs199859328 Chr.8: 143866854-143866854 on Build GRCh38
rs782182830 Chr.8: 143867071-143867071 on Build GRCh38
rs1036757233 Chr.8: 143867177-143867177 on Build GRCh38
rs536513369 Chr.8: 143867080-143867080 on Build GRCh38
rs575399844 Chr.8: 143867215-143867215 on Build GRCh38
rs782476654 Chr.8: 143866919-143866919 on Build GRCh38
rs782583923 Chr.8: 143867119-143867119 on Build GRCh38
rs782108580 Chr.8: 143866999-143866999 on Build GRCh38
rs577724900 Chr.8: 143866849-143866849 on Build GRCh38
rs560287631 Chr.8: 143867155-143867155 on Build GRCh38
rs782256924 Chr.8: 143866969-143866969 on Build GRCh38
rs532157702 Chr.8: 143866968-143866968 on Build GRCh38
rs782031234 Chr.8: 143867050-143867050 on Build GRCh38
rs782181155 Chr.8: 143866978-143866978 on Build GRCh38
rs375927494 Chr.8: 143867181-143867181 on Build GRCh38
rs782411839 Chr.8: 143867164-143867164 on Build GRCh38
rs782103340 Chr.8: 143866834-143866834 on Build GRCh38
rs782696825 Chr.8: 143867169-143867169 on Build GRCh38
rs782510861 Chr.8: 143867203-143867203 on Build GRCh38
rs185044374 Chr.8: 143866962-143866962 on Build GRCh38
rs781958045 Chr.8: 143866996-143866996 on Build GRCh38
rs1019316844 Chr.8: 143866847-143866847 on Build GRCh38
rs782245728 Chr.8: 143867122-143867122 on Build GRCh38
rs782195794 Chr.8: 143867211-143867211 on Build GRCh38
rs573169771 Chr.8: 143866949-143866949 on Build GRCh38
rs782381962 Chr.8: 143866888-143866888 on Build GRCh38
rs782820648 Chr.8: 143867132-143867132 on Build GRCh38
rs992722786 Chr.8: 143867008-143867008 on Build GRCh38
rs782819737 Chr.8: 143867082-143867082 on Build GRCh38
rs782711807 Chr.8: 143867097-143867097 on Build GRCh38
rs782724618 Chr.8: 143867136-143867136 on Build GRCh38
rs782461729 Chr.8: 143867199-143867199 on Build GRCh38
rs781951419 Chr.8: 143867266-143867266 on Build GRCh38
rs527896611 Chr.8: 143867156-143867156 on Build GRCh38
rs782111553 Chr.8: 143866986-143866986 on Build GRCh38
rs782777055 Chr.8: 143866908-143866908 on Build GRCh38
rs373841264 Chr.8: 143866942-143866942 on Build GRCh38
rs548952593 Chr.8: 143866823-143866823 on Build GRCh38
rs781808161 Chr.8: 143866831-143866831 on Build GRCh38
rs201160651 Chr.8: 143866857-143866857 on Build GRCh38
rs782181719 Chr.8: 143866921-143866921 on Build GRCh38
rs139137378 Chr.8: 143866985-143866985 on Build GRCh38
rs571579347 Chr.8: 143867216-143867216 on Build GRCh38
rs781980733 Chr.8: 143867003-143867003 on Build GRCh38
rs782061475 Chr.8: 143866899-143866899 on Build GRCh38
rs782318551 Chr.8: 143866820-143866820 on Build GRCh38
rs370081725 Chr.8: 143867062-143867062 on Build GRCh38
rs782390496 Chr.8: 143867125-143867125 on Build GRCh38
rs782764819 Chr.8: 143867063-143867063 on Build GRCh38
rs533349686 Chr.8: 143867029-143867029 on Build GRCh38
rs782686212 Chr.8: 143867058-143867058 on Build GRCh38
rs186320706 Chr.8: 143867118-143867118 on Build GRCh38
rs369610998 Chr.8: 143867034-143867034 on Build GRCh38
rs782089709 Chr.8: 143867178-143867178 on Build GRCh38
rs12681478 Chr.8: 143867251-143867251 on Build GRCh38
rs781923229 Chr.8: 143867032-143867032 on Build GRCh38
rs781914422 Chr.8: 143866897-143866897 on Build GRCh38
rs782364626 Chr.8: 143867210-143867210 on Build GRCh38
rs372460806 Chr.8: 143867102-143867102 on Build GRCh38
rs376941147 Chr.8: 143867278-143867278 on Build GRCh38
rs782789068 Chr.8: 143866904-143866904 on Build GRCh38
rs370476982 Chr.8: 143866916-143866916 on Build GRCh38
rs925364651 Chr.8: 143867024-143867024 on Build GRCh38
rs373346250 Chr.8: 143867054-143867054 on Build GRCh38

View Full Product Details