Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143867193-143867689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs949761822 Chr.8: 143867465-143867465 on Build GRCh38
rs782510299 Chr.8: 143867618-143867618 on Build GRCh38
rs375002026 Chr.8: 143867214-143867214 on Build GRCh38
rs377003169 Chr.8: 143867474-143867474 on Build GRCh38
rs368596970 Chr.8: 143867340-143867340 on Build GRCh38
rs35176503 Chr.8: 143867224-143867224 on Build GRCh38
rs528153180 Chr.8: 143867259-143867259 on Build GRCh38
rs781881308 Chr.8: 143867473-143867473 on Build GRCh38
rs782125978 Chr.8: 143867343-143867343 on Build GRCh38
rs375397416 Chr.8: 143867570-143867570 on Build GRCh38
rs370750586 Chr.8: 143867512-143867512 on Build GRCh38
rs561179913 Chr.8: 143867456-143867456 on Build GRCh38
rs782021058 Chr.8: 143867620-143867620 on Build GRCh38
rs782248529 Chr.8: 143867326-143867326 on Build GRCh38
rs1009516137 Chr.8: 143867390-143867390 on Build GRCh38
rs782406601 Chr.8: 143867320-143867320 on Build GRCh38
rs201996221 Chr.8: 143867633-143867633 on Build GRCh38
rs201495006 Chr.8: 143867361-143867361 on Build GRCh38
rs373806117 Chr.8: 143867461-143867461 on Build GRCh38
rs145468420 Chr.8: 143867431-143867431 on Build GRCh38
rs545639287 Chr.8: 143867220-143867220 on Build GRCh38
rs782648397 Chr.8: 143867368-143867368 on Build GRCh38
rs200778068 Chr.8: 143867486-143867486 on Build GRCh38
rs782230626 Chr.8: 143867272-143867272 on Build GRCh38
rs782092695 Chr.8: 143867335-143867335 on Build GRCh38
rs114128539 Chr.8: 143867417-143867417 on Build GRCh38
rs917528015 Chr.8: 143867581-143867581 on Build GRCh38
rs377560588 Chr.8: 143867310-143867310 on Build GRCh38
rs782722315 Chr.8: 143867299-143867299 on Build GRCh38
rs782210559 Chr.8: 143867440-143867440 on Build GRCh38
rs782734320 Chr.8: 143867223-143867223 on Build GRCh38
rs781792099 Chr.8: 143867349-143867349 on Build GRCh38
rs781865685 Chr.8: 143867246-143867246 on Build GRCh38
rs369677374 Chr.8: 143867658-143867658 on Build GRCh38
rs954328784 Chr.8: 143867446-143867446 on Build GRCh38
rs200739597 Chr.8: 143867566-143867566 on Build GRCh38
rs372006372 Chr.8: 143867659-143867659 on Build GRCh38
rs541086730 Chr.8: 143867442-143867442 on Build GRCh38
rs371299411 Chr.8: 143867600-143867600 on Build GRCh38
rs374869706 Chr.8: 143867434-143867434 on Build GRCh38
rs782069995 Chr.8: 143867565-143867565 on Build GRCh38
rs782548581 Chr.8: 143867503-143867503 on Build GRCh38
rs372536695 Chr.8: 143867291-143867291 on Build GRCh38
rs971453097 Chr.8: 143867595-143867595 on Build GRCh38
rs181215404 Chr.8: 143867491-143867491 on Build GRCh38
rs372213876 Chr.8: 143867634-143867634 on Build GRCh38
rs200834681 Chr.8: 143867555-143867555 on Build GRCh38
rs782308067 Chr.8: 143867338-143867338 on Build GRCh38
rs781882643 Chr.8: 143867405-143867405 on Build GRCh38
rs775059124 Chr.8: 143867554-143867554 on Build GRCh38
rs782495364 Chr.8: 143867648-143867648 on Build GRCh38
rs374236103 Chr.8: 143867545-143867545 on Build GRCh38
rs565248760 Chr.8: 143867546-143867546 on Build GRCh38
rs367640831 Chr.8: 143867252-143867252 on Build GRCh38
rs781905726 Chr.8: 143867575-143867575 on Build GRCh38
rs782560259 Chr.8: 143867308-143867308 on Build GRCh38
rs782470665 Chr.8: 143867411-143867411 on Build GRCh38
rs576788732 Chr.8: 143867428-143867428 on Build GRCh38
rs377058942 Chr.8: 143867296-143867296 on Build GRCh38
rs782087018 Chr.8: 143867628-143867628 on Build GRCh38
rs782200151 Chr.8: 143867316-143867316 on Build GRCh38
rs782487282 Chr.8: 143867312-143867312 on Build GRCh38
rs782460294 Chr.8: 143867418-143867418 on Build GRCh38
rs575399844 Chr.8: 143867215-143867215 on Build GRCh38
rs782405559 Chr.8: 143867212-143867212 on Build GRCh38
rs782728039 Chr.8: 143867557-143867557 on Build GRCh38
rs782399459 Chr.8: 143867599-143867599 on Build GRCh38
rs530256095 Chr.8: 143867626-143867626 on Build GRCh38
rs35785899 Chr.8: 143867571-143867571 on Build GRCh38
rs554849814 Chr.8: 143867370-143867370 on Build GRCh38
rs782050033 Chr.8: 143867222-143867222 on Build GRCh38
rs782456492 Chr.8: 143867353-143867353 on Build GRCh38
rs569958202 Chr.8: 143867383-143867383 on Build GRCh38
rs782429963 Chr.8: 143867550-143867550 on Build GRCh38
rs369357888 Chr.8: 143867443-143867443 on Build GRCh38
rs782393717 Chr.8: 143867376-143867376 on Build GRCh38
rs782399863 Chr.8: 143867327-143867327 on Build GRCh38
rs782238212 Chr.8: 143867375-143867375 on Build GRCh38
rs782446058 Chr.8: 143867254-143867254 on Build GRCh38
rs782195591 Chr.8: 143867372-143867372 on Build GRCh38
rs782575319 Chr.8: 143867584-143867584 on Build GRCh38
rs782149613 Chr.8: 143867279-143867279 on Build GRCh38
rs375347181 Chr.8: 143867314-143867314 on Build GRCh38
rs564150761 Chr.8: 143867550-143867550 on Build GRCh38
rs545377325 Chr.8: 143867528-143867529 on Build GRCh38
rs373165682 Chr.8: 143867260-143867260 on Build GRCh38
rs782677884 Chr.8: 143867506-143867506 on Build GRCh38
rs782559655 Chr.8: 143867650-143867650 on Build GRCh38
rs375302155 Chr.8: 143867640-143867640 on Build GRCh38
rs782691764 Chr.8: 143867261-143867261 on Build GRCh38
rs782205604 Chr.8: 143867643-143867643 on Build GRCh38
rs782238764 Chr.8: 143867605-143867605 on Build GRCh38
rs199879536 Chr.8: 143867387-143867387 on Build GRCh38
rs782191594 Chr.8: 143867596-143867596 on Build GRCh38
rs782700219 Chr.8: 143867469-143867469 on Build GRCh38
rs781901196 Chr.8: 143867637-143867637 on Build GRCh38
rs367617111 Chr.8: 143867371-143867371 on Build GRCh38
rs782373505 Chr.8: 143867263-143867263 on Build GRCh38
rs781951419 Chr.8: 143867266-143867266 on Build GRCh38
rs781837101 Chr.8: 143867483-143867483 on Build GRCh38
rs782768826 Chr.8: 143867244-143867244 on Build GRCh38
rs965408907 Chr.8: 143867422-143867422 on Build GRCh38
rs782694610 Chr.8: 143867322-143867322 on Build GRCh38
rs199828972 Chr.8: 143867280-143867280 on Build GRCh38
rs532408718 Chr.8: 143867547-143867547 on Build GRCh38
rs376560450 Chr.8: 143867449-143867449 on Build GRCh38
rs976943937 Chr.8: 143867285-143867285 on Build GRCh38
rs782767924 Chr.8: 143867394-143867394 on Build GRCh38
rs782661898 Chr.8: 143867424-143867424 on Build GRCh38
rs368673995 Chr.8: 143867580-143867580 on Build GRCh38
rs782718501 Chr.8: 143867388-143867388 on Build GRCh38
rs782763036 Chr.8: 143867346-143867346 on Build GRCh38
rs782609773 Chr.8: 143867543-143867543 on Build GRCh38
rs571579347 Chr.8: 143867216-143867216 on Build GRCh38
rs370847653 Chr.8: 143867311-143867311 on Build GRCh38
rs781931058 Chr.8: 143867339-143867339 on Build GRCh38
rs782395289 Chr.8: 143867517-143867517 on Build GRCh38
rs781953027 Chr.8: 143867615-143867615 on Build GRCh38
rs782655371 Chr.8: 143867641-143867641 on Build GRCh38
rs782020110 Chr.8: 143867391-143867391 on Build GRCh38
rs782616789 Chr.8: 143867496-143867496 on Build GRCh38
rs1020981879 Chr.8: 143867257-143867257 on Build GRCh38
rs782561343 Chr.8: 143867235-143867235 on Build GRCh38
rs572259751 Chr.8: 143867445-143867445 on Build GRCh38
rs781839108 Chr.8: 143867564-143867564 on Build GRCh38
rs12681478 Chr.8: 143867251-143867251 on Build GRCh38
rs375138049 Chr.8: 143867306-143867306 on Build GRCh38
rs559701261 Chr.8: 143867336-143867336 on Build GRCh38
rs1044515215 Chr.8: 143867468-143867468 on Build GRCh38
rs782779716 Chr.8: 143867624-143867624 on Build GRCh38
rs782312316 Chr.8: 143867619-143867619 on Build GRCh38
rs782033531 Chr.8: 143867213-143867213 on Build GRCh38
rs368672062 Chr.8: 143867477-143867477 on Build GRCh38
rs376941147 Chr.8: 143867278-143867278 on Build GRCh38
rs782493859 Chr.8: 143867583-143867583 on Build GRCh38
rs371636220 Chr.8: 143867290-143867290 on Build GRCh38
rs371992082 Chr.8: 143867448-143867448 on Build GRCh38
rs781854550 Chr.8: 143867341-143867341 on Build GRCh38
rs781948091 Chr.8: 143867328-143867328 on Build GRCh38
rs369842241 Chr.8: 143867455-143867455 on Build GRCh38

View Full Product Details