Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 16411769-16412281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC2
Entrez Gene ID
54796
Gene Name
basonuclin 2
Gene Aliases
BSN2
UniGene
Hs.656581
Gene Chromosome Location
Chr.9: 16409503-16870838 on Build GRCh38
Transcripts
NM_001317939.1 NM_001317940.1 NM_017637.5 XM_011517924.1 XM_017014816.1 XM_017014817.1 XM_017014818.1 XM_017014820.1 XM_017014822.1 XM_017014823.1 XM_017014824.1 XM_017014825.1 XM_017014829.1

SNP Detail

SNP ID Chromosome Location
rs757611383 Chr.9: 16412214-16412214 on Build GRCh38
rs1012061305 Chr.9: 16411931-16411931 on Build GRCh38
rs940651258 Chr.9: 16412022-16412022 on Build GRCh38
rs549795903 Chr.9: 16411875-16411875 on Build GRCh38
rs1019269369 Chr.9: 16412207-16412207 on Build GRCh38
rs535474710 Chr.9: 16411946-16411946 on Build GRCh38
rs1002949695 Chr.9: 16412111-16412111 on Build GRCh38
rs538337182 Chr.9: 16411912-16411912 on Build GRCh38
rs182933689 Chr.9: 16412187-16412187 on Build GRCh38
rs1036828148 Chr.9: 16411800-16411800 on Build GRCh38
rs566106404 Chr.9: 16412073-16412073 on Build GRCh38
rs953498215 Chr.9: 16412249-16412249 on Build GRCh38
rs1023436057 Chr.9: 16411975-16411975 on Build GRCh38
rs576946131 Chr.9: 16412228-16412228 on Build GRCh38
rs908534545 Chr.9: 16412219-16412219 on Build GRCh38
rs963801720 Chr.9: 16412182-16412182 on Build GRCh38
rs1047466191 Chr.9: 16411894-16411894 on Build GRCh38
rs942870301 Chr.9: 16411878-16411878 on Build GRCh38
rs1021268220 Chr.9: 16411880-16411880 on Build GRCh38
rs111436809 Chr.9: 16411856-16411856 on Build GRCh38
rs369663348 Chr.9: 16412084-16412084 on Build GRCh38
rs912274305 Chr.9: 16411906-16411906 on Build GRCh38
rs1011444382 Chr.9: 16411876-16411876 on Build GRCh38
rs180699594 Chr.9: 16411910-16411910 on Build GRCh38
rs1006040298 Chr.9: 16411913-16411913 on Build GRCh38
rs1012959283 Chr.9: 16412244-16412244 on Build GRCh38
rs941392545 Chr.9: 16412224-16412224 on Build GRCh38
rs922092717 Chr.9: 16411928-16411928 on Build GRCh38
rs1044844299 Chr.9: 16412245-16412245 on Build GRCh38
rs932246153 Chr.9: 16411949-16411949 on Build GRCh38
rs909227523 Chr.9: 16412008-16412008 on Build GRCh38
rs894533089 Chr.9: 16412232-16412232 on Build GRCh38
rs187389867 Chr.9: 16411924-16411924 on Build GRCh38
rs1047342843 Chr.9: 16412218-16412218 on Build GRCh38
rs1003472529 Chr.9: 16411992-16411992 on Build GRCh38
rs1049331111 Chr.9: 16411972-16411972 on Build GRCh38
rs748701469 Chr.9: 16412240-16412240 on Build GRCh38
rs76971402 Chr.9: 16412206-16412206 on Build GRCh38
rs956439634 Chr.9: 16412113-16412113 on Build GRCh38
rs560868800 Chr.9: 16411808-16411808 on Build GRCh38
rs779083473 Chr.9: 16412236-16412236 on Build GRCh38
rs909426957 Chr.9: 16412252-16412252 on Build GRCh38
rs907055018 Chr.9: 16411841-16411841 on Build GRCh38
rs190777189 Chr.9: 16412089-16412089 on Build GRCh38
rs1021467651 Chr.9: 16412185-16412185 on Build GRCh38
rs775315376 Chr.9: 16411955-16411955 on Build GRCh38
rs765391732 Chr.9: 16412231-16412231 on Build GRCh38
rs531380103 Chr.9: 16411869-16411869 on Build GRCh38
rs554797223 Chr.9: 16412135-16412135 on Build GRCh38
rs1036454209 Chr.9: 16412032-16412032 on Build GRCh38
rs749417258 Chr.9: 16412045-16412045 on Build GRCh38
rs922820650 Chr.9: 16411881-16411881 on Build GRCh38
rs985060796 Chr.9: 16412251-16412251 on Build GRCh38
rs967034762 Chr.9: 16412195-16412195 on Build GRCh38
rs760670979 Chr.9: 16411966-16411966 on Build GRCh38
rs1056070491 Chr.9: 16412116-16412116 on Build GRCh38
rs771631736 Chr.9: 16411813-16411816 on Build GRCh38
rs975555554 Chr.9: 16412197-16412197 on Build GRCh38
rs145195024 Chr.9: 16412161-16412161 on Build GRCh38
rs970581765 Chr.9: 16411989-16411989 on Build GRCh38
rs922768332 Chr.9: 16412203-16412203 on Build GRCh38
rs977211130 Chr.9: 16411892-16411892 on Build GRCh38
rs569743931 Chr.9: 16411891-16411891 on Build GRCh38
rs189408752 Chr.9: 16411799-16411799 on Build GRCh38

View Full Product Details