Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 16412587-16413100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC2
Entrez Gene ID
54796
Gene Name
basonuclin 2
Gene Aliases
BSN2
UniGene
Hs.656581
Gene Chromosome Location
Chr.9: 16409503-16870838 on Build GRCh38
Transcripts
NM_001317939.1 NM_001317940.1 NM_017637.5 XM_011517924.1 XM_017014816.1 XM_017014817.1 XM_017014818.1 XM_017014820.1 XM_017014822.1 XM_017014823.1 XM_017014824.1 XM_017014825.1 XM_017014829.1

SNP Detail

SNP ID Chromosome Location
rs377205832 Chr.9: 16412668-16412669 on Build GRCh38
rs879220603 Chr.9: 16412721-16412726 on Build GRCh38
rs763706694 Chr.9: 16412737-16412746 on Build GRCh38
rs899480256 Chr.9: 16412728-16412728 on Build GRCh38
rs16934619 Chr.9: 16412870-16412870 on Build GRCh38
rs574864912 Chr.9: 16413056-16413056 on Build GRCh38
rs946608470 Chr.9: 16412718-16412718 on Build GRCh38
rs1035921796 Chr.9: 16412681-16412681 on Build GRCh38
rs548357502 Chr.9: 16412867-16412867 on Build GRCh38
rs914688361 Chr.9: 16413067-16413067 on Build GRCh38
rs554130237 Chr.9: 16413008-16413008 on Build GRCh38
rs771533583 Chr.9: 16412986-16412986 on Build GRCh38
rs775036340 Chr.9: 16413017-16413017 on Build GRCh38
rs922422724 Chr.9: 16412999-16412999 on Build GRCh38
rs530278920 Chr.9: 16412764-16412764 on Build GRCh38
rs113241027 Chr.9: 16412744-16412744 on Build GRCh38
rs934486775 Chr.9: 16412746-16412746 on Build GRCh38
rs968798014 Chr.9: 16412719-16412719 on Build GRCh38
rs922508265 Chr.9: 16412751-16412751 on Build GRCh38
rs1057323905 Chr.9: 16413073-16413073 on Build GRCh38
rs1035687047 Chr.9: 16413040-16413040 on Build GRCh38
rs537319646 Chr.9: 16412885-16412885 on Build GRCh38
rs796299670 Chr.9: 16412662-16412663 on Build GRCh38
rs890854363 Chr.9: 16412761-16412761 on Build GRCh38
rs991415513 Chr.9: 16412714-16412714 on Build GRCh38
rs978802947 Chr.9: 16412720-16412720 on Build GRCh38
rs928629369 Chr.9: 16412833-16412833 on Build GRCh38
rs374746041 Chr.9: 16412721-16412730 on Build GRCh38
rs200034269 Chr.9: 16412721-16412746 on Build GRCh38
rs962892670 Chr.9: 16412781-16412782 on Build GRCh38
rs1029753819 Chr.9: 16412761-16412768 on Build GRCh38
rs879147664 Chr.9: 16412734-16412734 on Build GRCh38
rs924678687 Chr.9: 16412956-16412956 on Build GRCh38
rs909563011 Chr.9: 16413019-16413019 on Build GRCh38
rs932573119 Chr.9: 16412943-16412943 on Build GRCh38
rs981480076 Chr.9: 16412661-16412661 on Build GRCh38
rs921152529 Chr.9: 16412876-16412876 on Build GRCh38
rs940983821 Chr.9: 16413030-16413030 on Build GRCh38
rs148600867 Chr.9: 16412732-16412732 on Build GRCh38
rs1003696961 Chr.9: 16413065-16413065 on Build GRCh38
rs888234634 Chr.9: 16412626-16412626 on Build GRCh38
rs915911092 Chr.9: 16412637-16412637 on Build GRCh38
rs981385411 Chr.9: 16412829-16412829 on Build GRCh38
rs1005300569 Chr.9: 16412627-16412627 on Build GRCh38
rs898007796 Chr.9: 16412775-16412775 on Build GRCh38
rs996541953 Chr.9: 16412746-16412746 on Build GRCh38
rs60922728 Chr.9: 16412755-16412766 on Build GRCh38
rs978832396 Chr.9: 16412902-16412902 on Build GRCh38
rs892302137 Chr.9: 16412692-16412692 on Build GRCh38
rs970298421 Chr.9: 16412821-16412821 on Build GRCh38
rs976644717 Chr.9: 16412998-16412998 on Build GRCh38
rs532249895 Chr.9: 16412660-16412660 on Build GRCh38
rs1050741642 Chr.9: 16412973-16412974 on Build GRCh38
rs528650308 Chr.9: 16412620-16412620 on Build GRCh38
rs1023108776 Chr.9: 16413004-16413004 on Build GRCh38
rs200175163 Chr.9: 16412711-16412711 on Build GRCh38
rs535119022 Chr.9: 16413002-16413002 on Build GRCh38
rs58174243 Chr.9: 16412749-16412766 on Build GRCh38
rs144799488 Chr.9: 16412906-16412906 on Build GRCh38
rs993642871 Chr.9: 16412844-16412844 on Build GRCh38
rs1005336409 Chr.9: 16412767-16412767 on Build GRCh38
rs1037192190 Chr.9: 16413047-16413047 on Build GRCh38
rs569020425 Chr.9: 16412721-16412732 on Build GRCh38
rs915909494 Chr.9: 16412717-16412717 on Build GRCh38
rs956273289 Chr.9: 16413045-16413045 on Build GRCh38
rs1036362145 Chr.9: 16412862-16412862 on Build GRCh38
rs932545505 Chr.9: 16412756-16412756 on Build GRCh38
rs962221997 Chr.9: 16412663-16412663 on Build GRCh38
rs1036755109 Chr.9: 16412768-16412768 on Build GRCh38
rs570713010 Chr.9: 16412918-16412918 on Build GRCh38
rs989334382 Chr.9: 16412858-16412858 on Build GRCh38
rs970244596 Chr.9: 16413018-16413018 on Build GRCh38
rs974280003 Chr.9: 16412809-16412809 on Build GRCh38
rs375875977 Chr.9: 16412689-16412689 on Build GRCh38
rs542079492 Chr.9: 16412866-16412866 on Build GRCh38
rs1041360642 Chr.9: 16412748-16412748 on Build GRCh38
rs147956947 Chr.9: 16412746-16412747 on Build GRCh38
rs192720739 Chr.9: 16413052-16413052 on Build GRCh38
rs566267625 Chr.9: 16412763-16412763 on Build GRCh38
rs775428835 Chr.9: 16412735-16412746 on Build GRCh38
rs996488468 Chr.9: 16412617-16412617 on Build GRCh38
rs890791602 Chr.9: 16412976-16412976 on Build GRCh38
rs73415403 Chr.9: 16412670-16412670 on Build GRCh38
rs148705430 Chr.9: 16412985-16412987 on Build GRCh38
rs1003966101 Chr.9: 16412983-16412983 on Build GRCh38

View Full Product Details